Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 152994186-152994664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MTRF1L
Entrez Gene ID
54516
Gene Name
mitochondrial translational release factor 1 like
Gene Aliases
HMRF1L, MRF1L, mtRF1a
UniGene
Hs.225836
Gene Chromosome Location
Chr.6: 152987349-153007301 on Build GRCh38
Transcripts
NM_001114184.2 NM_001301047.2 NM_001301870.1 NM_001301871.1 NM_019041.6 NR_126056.1 XM_017010959.1 XM_017010960.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3994694 c.621G>C p.K207N Chr.6: 152994579-152994579 on Build GRCh38
3994695 c.213G>C p.K71N Chr.6: 152994579-152994579 on Build GRCh38
6870291 c.577A>G p.M193V Chr.6: 152994623-152994623 on Build GRCh38
3761599 c.232G>A p.V78I Chr.6: 152994560-152994560 on Build GRCh38
3859787 c.638G>A p.R213H Chr.6: 152994562-152994562 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs890917532 Chr.6: 152994237-152994237 on Build GRCh38
rs1007390952 Chr.6: 152994243-152994243 on Build GRCh38
rs763763647 Chr.6: 152994472-152994472 on Build GRCh38
rs754139707 Chr.6: 152994634-152994634 on Build GRCh38
rs3799135 Chr.6: 152994482-152994482 on Build GRCh38
rs750610688 Chr.6: 152994334-152994334 on Build GRCh38
rs1035868324 Chr.6: 152994368-152994368 on Build GRCh38
rs986607190 Chr.6: 152994541-152994541 on Build GRCh38
rs3192723 Chr.6: 152994560-152994560 on Build GRCh38
rs200658267 Chr.6: 152994556-152994556 on Build GRCh38
rs745385150 Chr.6: 152994544-152994544 on Build GRCh38
rs772706234 Chr.6: 152994532-152994532 on Build GRCh38
rs79968572 Chr.6: 152994244-152994244 on Build GRCh38
rs1035987927 Chr.6: 152994590-152994591 on Build GRCh38
rs913436100 Chr.6: 152994597-152994597 on Build GRCh38
rs1054681303 Chr.6: 152994593-152994593 on Build GRCh38
rs771511415 Chr.6: 152994307-152994310 on Build GRCh38
rs750502550 Chr.6: 152994483-152994483 on Build GRCh38
rs749428920 Chr.6: 152994511-152994511 on Build GRCh38
rs757173098 Chr.6: 152994481-152994481 on Build GRCh38
rs1054649077 Chr.6: 152994309-152994309 on Build GRCh38
rs779401000 Chr.6: 152994644-152994644 on Build GRCh38
rs542420466 Chr.6: 152994501-152994501 on Build GRCh38
rs755413374 Chr.6: 152994513-152994513 on Build GRCh38
rs748970349 Chr.6: 152994319-152994319 on Build GRCh38
rs774949029 Chr.6: 152994467-152994467 on Build GRCh38
rs1003088452 Chr.6: 152994357-152994357 on Build GRCh38
rs373567855 Chr.6: 152994480-152994480 on Build GRCh38
rs544807555 Chr.6: 152994443-152994443 on Build GRCh38
rs57819885 Chr.6: 152994609-152994609 on Build GRCh38
rs575123074 Chr.6: 152994268-152994270 on Build GRCh38
rs967840882 Chr.6: 152994629-152994629 on Build GRCh38
rs979109947 Chr.6: 152994640-152994640 on Build GRCh38
rs375981966 Chr.6: 152994562-152994562 on Build GRCh38
rs779127530 Chr.6: 152994525-152994525 on Build GRCh38
rs369828849 Chr.6: 152994558-152994558 on Build GRCh38
rs1039923499 Chr.6: 152994552-152994552 on Build GRCh38
rs907223395 Chr.6: 152994363-152994363 on Build GRCh38
rs766335720 Chr.6: 152994612-152994612 on Build GRCh38
rs774011789 Chr.6: 152994563-152994563 on Build GRCh38
rs1046228921 Chr.6: 152994324-152994324 on Build GRCh38
rs958936169 Chr.6: 152994392-152994392 on Build GRCh38
rs772683410 Chr.6: 152994607-152994609 on Build GRCh38
rs778357906 Chr.6: 152994539-152994539 on Build GRCh38
rs191229290 Chr.6: 152994460-152994460 on Build GRCh38
rs761530340 Chr.6: 152994575-152994575 on Build GRCh38
rs762831078 Chr.6: 152994471-152994471 on Build GRCh38
rs1034281812 Chr.6: 152994423-152994423 on Build GRCh38
rs552710702 Chr.6: 152994304-152994304 on Build GRCh38
rs992067089 Chr.6: 152994397-152994397 on Build GRCh38
rs1001926119 Chr.6: 152994398-152994398 on Build GRCh38
rs1010330277 Chr.6: 152994321-152994321 on Build GRCh38
rs1002867557 Chr.6: 152994571-152994571 on Build GRCh38
rs748727246 Chr.6: 152994526-152994526 on Build GRCh38
rs372750968 Chr.6: 152994561-152994561 on Build GRCh38
rs916358081 Chr.6: 152994409-152994409 on Build GRCh38
rs3799136 Chr.6: 152994426-152994426 on Build GRCh38
rs755142691 Chr.6: 152994643-152994643 on Build GRCh38
rs767451675 Chr.6: 152994588-152994588 on Build GRCh38
rs764986177 Chr.6: 152994465-152994465 on Build GRCh38
rs995543297 Chr.6: 152994278-152994278 on Build GRCh38
rs573582962 Chr.6: 152994218-152994218 on Build GRCh38
rs576731150 Chr.6: 152994608-152994608 on Build GRCh38
rs951280668 Chr.6: 152994508-152994508 on Build GRCh38
rs34086858 Chr.6: 152994489-152994490 on Build GRCh38

View Full Product Details