Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 132600150-132600709 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACAD11
Entrez Gene ID
84129
Gene Name
acyl-CoA dehydrogenase family member 11
Gene Aliases
ACAD-11
UniGene
Hs.441378
Gene Chromosome Location
Chr.3: 132558138-132660131 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Gene Symbol
ACKR4
Entrez Gene ID
51554
Gene Name
atypical chemokine receptor 4
Gene Aliases
CC-CKR-11, CCBP2, CCR-11, CCR10, CCR11, CCRL1, CCX CKR, CCX-CKR, CKR-11, PPR1, VSHK1
UniGene
Hs.310512
Gene Chromosome Location
Chr.3: 132597237-132602644 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Gene Symbol
NPHP3-ACAD11
Entrez Gene ID
100532724
Gene Name
NPHP3-ACAD11 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.511991
Gene Chromosome Location
Chr.3: 132558138-132722459 on Build GRCh38
Transcripts
NM_032169.4 NR_132426.1 NR_132427.1 NR_132428.1 NM_016557.3 NM_178445.2 NR_037804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3373019 c.14A>C p.Q5P Chr.3: 132600411-132600411 on Build GRCh38
7416663 c.175A>G p.N59D Chr.3: 132600572-132600572 on Build GRCh38
7353958 c.293C>T p.A98V Chr.3: 132600690-132600690 on Build GRCh38
3774550 c.169G>A p.A57T Chr.3: 132600566-132600566 on Build GRCh38
3587596 c.156C>T p.F52F Chr.3: 132600553-132600553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758894103 Chr.3: 132600464-132600464 on Build GRCh38
rs750226016 Chr.3: 132600370-132600374 on Build GRCh38
rs776936636 Chr.3: 132600365-132600365 on Build GRCh38
rs764614922 Chr.3: 132600590-132600590 on Build GRCh38
rs977081381 Chr.3: 132600311-132600311 on Build GRCh38
rs1013560779 Chr.3: 132600501-132600501 on Build GRCh38
rs1002305250 Chr.3: 132600254-132600254 on Build GRCh38
rs565084078 Chr.3: 132600295-132600295 on Build GRCh38
rs769995139 Chr.3: 132600486-132600486 on Build GRCh38
rs774366222 Chr.3: 132600527-132600527 on Build GRCh38
rs965429226 Chr.3: 132600661-132600661 on Build GRCh38
rs746138637 Chr.3: 132600482-132600482 on Build GRCh38
rs543904253 Chr.3: 132600294-132600294 on Build GRCh38
rs56119026 Chr.3: 132600556-132600556 on Build GRCh38
rs981858059 Chr.3: 132600323-132600323 on Build GRCh38
rs755793042 Chr.3: 132600436-132600436 on Build GRCh38
rs932489634 Chr.3: 132600332-132600332 on Build GRCh38
rs141021878 Chr.3: 132600554-132600554 on Build GRCh38
rs761684366 Chr.3: 132600425-132600425 on Build GRCh38
rs574430457 Chr.3: 132600227-132600227 on Build GRCh38
rs760462327 Chr.3: 132600549-132600549 on Build GRCh38
rs768129807 Chr.3: 132600185-132600185 on Build GRCh38
rs146640019 Chr.3: 132600536-132600536 on Build GRCh38
rs921161555 Chr.3: 132600320-132600320 on Build GRCh38
rs925770820 Chr.3: 132600456-132600456 on Build GRCh38
rs937418623 Chr.3: 132600372-132600372 on Build GRCh38
rs200765869 Chr.3: 132600496-132600496 on Build GRCh38
rs769901948 Chr.3: 132600384-132600384 on Build GRCh38
rs747427067 Chr.3: 132600466-132600466 on Build GRCh38
rs773115352 Chr.3: 132600546-132600546 on Build GRCh38
rs773207367 Chr.3: 132600686-132600686 on Build GRCh38
rs908243151 Chr.3: 132600282-132600282 on Build GRCh38
rs761613249 Chr.3: 132600529-132600529 on Build GRCh38
rs1026812655 Chr.3: 132600221-132600221 on Build GRCh38
rs139827468 Chr.3: 132600475-132600475 on Build GRCh38
rs766124398 Chr.3: 132600553-132600553 on Build GRCh38
rs754527548 Chr.3: 132600558-132600558 on Build GRCh38
rs61748105 Chr.3: 132600517-132600517 on Build GRCh38
rs781594676 Chr.3: 132600613-132600613 on Build GRCh38
rs532559426 Chr.3: 132600393-132600393 on Build GRCh38
rs757673404 Chr.3: 132600469-132600469 on Build GRCh38
rs759715558 Chr.3: 132600380-132600380 on Build GRCh38
rs764092132 Chr.3: 132600395-132600395 on Build GRCh38
rs768810205 Chr.3: 132600676-132600676 on Build GRCh38
rs772121242 Chr.3: 132600682-132600682 on Build GRCh38
rs1027143313 Chr.3: 132600570-132600570 on Build GRCh38
rs752150433 Chr.3: 132600598-132600598 on Build GRCh38
rs774091672 Chr.3: 132600399-132600399 on Build GRCh38
rs756407396 Chr.3: 132600626-132600626 on Build GRCh38
rs147449210 Chr.3: 132600629-132600629 on Build GRCh38
rs370395498 Chr.3: 132600457-132600457 on Build GRCh38
rs757683408 Chr.3: 132600600-132600600 on Build GRCh38
rs908473851 Chr.3: 132600271-132600271 on Build GRCh38
rs750127744 Chr.3: 132600426-132600426 on Build GRCh38
rs749424889 Chr.3: 132600665-132600665 on Build GRCh38
rs375285193 Chr.3: 132600582-132600582 on Build GRCh38
rs755172967 Chr.3: 132600665-132600667 on Build GRCh38
rs1027091624 Chr.3: 132600257-132600257 on Build GRCh38
rs746230914 Chr.3: 132600615-132600615 on Build GRCh38
rs982702990 Chr.3: 132600269-132600269 on Build GRCh38
rs774261094 Chr.3: 132600680-132600680 on Build GRCh38
rs777999661 Chr.3: 132600463-132600463 on Build GRCh38
rs1042931661 Chr.3: 132600424-132600424 on Build GRCh38
rs771132633 Chr.3: 132600349-132600349 on Build GRCh38
rs775525024 Chr.3: 132600386-132600386 on Build GRCh38
rs540931804 Chr.3: 132600465-132600465 on Build GRCh38

View Full Product Details