Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 19404187-19404758 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSGALNACT1
Entrez Gene ID
55790
Gene Name
chondroitin sulfate N-acetylgalactosaminyltransferase 1
Gene Aliases
CSGalNAcT-1, ChGn, beta4GalNAcT
UniGene
Hs.613729
Gene Chromosome Location
Chr.8: 19404161-19757940 on Build GRCh38
Transcripts
NM_001130518.1 NM_018371.4 NR_024040.1 XM_006716358.2 XM_006716359.2 XM_006716360.2 XM_006716361.2 XM_006716362.2 XM_006716363.1 XM_006716364.3 XM_011544578.1 XM_011544579.1 XM_011544580.1 XM_011544582.1 XM_011544583.1 XM_011544584.1 XM_011544585.2 XM_017013623.1 XM_017013624.1 XM_017013625.1 XM_017013626.1 XM_017013627.1 XM_017013628.1 XM_017013629.1 XM_017013630.1 XM_017013631.1 XM_017013632.1 XM_017013633.1 XM_017013634.1 XM_017013635.1 XM_017013636.1 XM_017013637.1 XM_017013638.1 XM_017013639.1

SNP Detail

SNP ID Chromosome Location
rs34024297 Chr.8: 19404608-19404609 on Build GRCh38
rs1004552901 Chr.8: 19404315-19404315 on Build GRCh38
rs540162753 Chr.8: 19404348-19404351 on Build GRCh38
rs796962635 Chr.8: 19404623-19404624 on Build GRCh38
rs553039039 Chr.8: 19404304-19404304 on Build GRCh38
rs562936755 Chr.8: 19404333-19404333 on Build GRCh38
rs977849024 Chr.8: 19404719-19404719 on Build GRCh38
rs775010744 Chr.8: 19404670-19404670 on Build GRCh38
rs530234302 Chr.8: 19404450-19404450 on Build GRCh38
rs750940895 Chr.8: 19404419-19404419 on Build GRCh38
rs1024763312 Chr.8: 19404251-19404251 on Build GRCh38
rs772072131 Chr.8: 19404628-19404628 on Build GRCh38
rs572456370 Chr.8: 19404379-19404379 on Build GRCh38
rs35704904 Chr.8: 19404624-19404625 on Build GRCh38
rs368106336 Chr.8: 19404375-19404376 on Build GRCh38
rs940307128 Chr.8: 19404491-19404491 on Build GRCh38
rs747590058 Chr.8: 19404462-19404462 on Build GRCh38
rs757717411 Chr.8: 19404606-19404607 on Build GRCh38
rs779017204 Chr.8: 19404646-19404646 on Build GRCh38
rs559907777 Chr.8: 19404658-19404658 on Build GRCh38
rs532321542 Chr.8: 19404215-19404215 on Build GRCh38
rs758304801 Chr.8: 19404617-19404618 on Build GRCh38
rs1029493551 Chr.8: 19404688-19404688 on Build GRCh38
rs1003336312 Chr.8: 19404620-19404620 on Build GRCh38
rs968849810 Chr.8: 19404412-19404412 on Build GRCh38
rs66526055 Chr.8: 19404624-19404624 on Build GRCh38
rs552349198 Chr.8: 19404635-19404635 on Build GRCh38
rs752824226 Chr.8: 19404308-19404308 on Build GRCh38
rs760789514 Chr.8: 19404674-19404674 on Build GRCh38
rs1021651922 Chr.8: 19404667-19404667 on Build GRCh38
rs764356650 Chr.8: 19404366-19404366 on Build GRCh38
rs754253829 Chr.8: 19404368-19404368 on Build GRCh38
rs561215350 Chr.8: 19404499-19404499 on Build GRCh38
rs764442353 Chr.8: 19404700-19404700 on Build GRCh38
rs947856492 Chr.8: 19404577-19404577 on Build GRCh38
rs898890790 Chr.8: 19404502-19404502 on Build GRCh38
rs767766982 Chr.8: 19404306-19404306 on Build GRCh38
rs548241999 Chr.8: 19404622-19404622 on Build GRCh38
rs987682608 Chr.8: 19404488-19404488 on Build GRCh38
rs796235789 Chr.8: 19404358-19404358 on Build GRCh38
rs1037821766 Chr.8: 19404326-19404326 on Build GRCh38
rs34647465 Chr.8: 19404623-19404624 on Build GRCh38
rs758930870 Chr.8: 19404442-19404442 on Build GRCh38
rs769447265 Chr.8: 19404466-19404466 on Build GRCh38
rs780902186 Chr.8: 19404623-19404626 on Build GRCh38
rs762157206 Chr.8: 19404448-19404448 on Build GRCh38
rs775728881 Chr.8: 19404645-19404645 on Build GRCh38
rs57082500 Chr.8: 19404624-19404624 on Build GRCh38
rs969195944 Chr.8: 19404668-19404668 on Build GRCh38
rs7001231 Chr.8: 19404219-19404219 on Build GRCh38
rs766710235 Chr.8: 19404625-19404626 on Build GRCh38
rs566512247 Chr.8: 19404414-19404415 on Build GRCh38
rs1038079788 Chr.8: 19404607-19404607 on Build GRCh38
rs757524016 Chr.8: 19404396-19404396 on Build GRCh38
rs777352677 Chr.8: 19404477-19404477 on Build GRCh38
rs78627047 Chr.8: 19404459-19404459 on Build GRCh38
rs547308937 Chr.8: 19404374-19404375 on Build GRCh38
rs936555705 Chr.8: 19404376-19404376 on Build GRCh38
rs1023489580 Chr.8: 19404348-19404349 on Build GRCh38
rs760984516 Chr.8: 19404359-19404359 on Build GRCh38
rs1020465146 Chr.8: 19404694-19404694 on Build GRCh38
rs532782845 Chr.8: 19404626-19404626 on Build GRCh38
rs763429382 Chr.8: 19404273-19404273 on Build GRCh38
rs1014396444 Chr.8: 19404670-19404670 on Build GRCh38
rs779306539 Chr.8: 19404402-19404402 on Build GRCh38
rs1035653759 Chr.8: 19404365-19404365 on Build GRCh38
rs886486345 Chr.8: 19404547-19404547 on Build GRCh38
rs746903152 Chr.8: 19404622-19404623 on Build GRCh38
rs544124721 Chr.8: 19404601-19404601 on Build GRCh38
rs3840719 Chr.8: 19404607-19404608 on Build GRCh38
rs908132467 Chr.8: 19404490-19404490 on Build GRCh38
rs7005528 Chr.8: 19404595-19404595 on Build GRCh38
rs116759812 Chr.8: 19404702-19404702 on Build GRCh38
rs1051555335 Chr.8: 19404485-19404485 on Build GRCh38
rs924989741 Chr.8: 19404343-19404343 on Build GRCh38

View Full Product Details