Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 70093425-70094007 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFY2
Entrez Gene ID
55506
Gene Name
H2A histone family member Y2
Gene Aliases
macroH2A2
UniGene
Hs.499953
Gene Chromosome Location
Chr.10: 70052601-70112284 on Build GRCh38
Transcripts
NM_018649.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3440062 c.485C>T p.P162L Chr.10: 70093742-70093742 on Build GRCh38
4976398 c.510T>G p.T170T Chr.10: 70093767-70093767 on Build GRCh38
6990635 c.519C>G p.S173S Chr.10: 70093776-70093776 on Build GRCh38
3867698 c.588G>A p.K196K Chr.10: 70093845-70093845 on Build GRCh38
4643471 c.493A>G p.S165G Chr.10: 70093750-70093750 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1034756358 Chr.10: 70093714-70093714 on Build GRCh38
rs962567845 Chr.10: 70093693-70093693 on Build GRCh38
rs764489328 Chr.10: 70093684-70093684 on Build GRCh38
rs748849388 Chr.10: 70093689-70093690 on Build GRCh38
rs556508031 Chr.10: 70093581-70093581 on Build GRCh38
rs149601898 Chr.10: 70093815-70093815 on Build GRCh38
rs115694077 Chr.10: 70093782-70093782 on Build GRCh38
rs776962995 Chr.10: 70093647-70093647 on Build GRCh38
rs1003222715 Chr.10: 70093678-70093678 on Build GRCh38
rs999295759 Chr.10: 70093608-70093608 on Build GRCh38
rs752613211 Chr.10: 70093566-70093566 on Build GRCh38
rs761072448 Chr.10: 70093472-70093472 on Build GRCh38
rs903303934 Chr.10: 70093577-70093577 on Build GRCh38
rs765216015 Chr.10: 70093677-70093677 on Build GRCh38
rs764579018 Chr.10: 70093831-70093831 on Build GRCh38
rs146404483 Chr.10: 70093753-70093753 on Build GRCh38
rs574755723 Chr.10: 70093641-70093641 on Build GRCh38
rs1054475359 Chr.10: 70093916-70093916 on Build GRCh38
rs757510295 Chr.10: 70093692-70093692 on Build GRCh38
rs10999133 Chr.10: 70093551-70093551 on Build GRCh38
rs758515975 Chr.10: 70093895-70093895 on Build GRCh38
rs946588612 Chr.10: 70093945-70093945 on Build GRCh38
rs564494641 Chr.10: 70093861-70093861 on Build GRCh38
rs544375651 Chr.10: 70093563-70093563 on Build GRCh38
rs796184534 Chr.10: 70093607-70093607 on Build GRCh38
rs374912544 Chr.10: 70093720-70093720 on Build GRCh38
rs893344080 Chr.10: 70093492-70093492 on Build GRCh38
rs193036230 Chr.10: 70093730-70093730 on Build GRCh38
rs910511504 Chr.10: 70093510-70093510 on Build GRCh38
rs952256954 Chr.10: 70093768-70093768 on Build GRCh38
rs116983359 Chr.10: 70093873-70093873 on Build GRCh38
rs748961382 Chr.10: 70093752-70093752 on Build GRCh38
rs147123101 Chr.10: 70093824-70093824 on Build GRCh38
rs1039122931 Chr.10: 70093552-70093552 on Build GRCh38
rs531795922 Chr.10: 70093810-70093810 on Build GRCh38
rs755906354 Chr.10: 70093746-70093746 on Build GRCh38
rs533551376 Chr.10: 70093923-70093923 on Build GRCh38
rs375251282 Chr.10: 70093850-70093850 on Build GRCh38
rs1004165753 Chr.10: 70093672-70093672 on Build GRCh38
rs577157357 Chr.10: 70093562-70093562 on Build GRCh38
rs370312833 Chr.10: 70093691-70093691 on Build GRCh38
rs550582562 Chr.10: 70093462-70093462 on Build GRCh38
rs572730923 Chr.10: 70093511-70093511 on Build GRCh38
rs148926673 Chr.10: 70093557-70093557 on Build GRCh38
rs756016089 Chr.10: 70093682-70093682 on Build GRCh38
rs754146722 Chr.10: 70093690-70093690 on Build GRCh38
rs767427903 Chr.10: 70093729-70093729 on Build GRCh38
rs893713450 Chr.10: 70093657-70093657 on Build GRCh38
rs779187429 Chr.10: 70093716-70093716 on Build GRCh38
rs973916611 Chr.10: 70093694-70093694 on Build GRCh38
rs114997689 Chr.10: 70093834-70093834 on Build GRCh38
rs934778139 Chr.10: 70093902-70093902 on Build GRCh38
rs777502925 Chr.10: 70093748-70093748 on Build GRCh38
rs1022700140 Chr.10: 70093697-70093697 on Build GRCh38
rs560568526 Chr.10: 70093661-70093661 on Build GRCh38
rs370708667 Chr.10: 70093658-70093658 on Build GRCh38
rs536989478 Chr.10: 70093659-70093659 on Build GRCh38
rs771427114 Chr.10: 70093813-70093813 on Build GRCh38
rs111936676 Chr.10: 70093612-70093612 on Build GRCh38
rs758411497 Chr.10: 70093721-70093721 on Build GRCh38

View Full Product Details