Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 67705181-67705774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GNG12
Entrez Gene ID
55970
Gene Name
G protein subunit gamma 12
Gene Aliases
-
UniGene
Hs.431101
Gene Chromosome Location
Chr.1: 67701466-67833779 on Build GRCh38
Transcripts
NM_018841.5 XM_017001809.1 XM_017001810.1 XM_017001811.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3985212 c.203C>G p.T68S Chr.1: 67705467-67705467 on Build GRCh38
4009667 c.137A>G p.H46R Chr.1: 67705533-67705533 on Build GRCh38
4009668 c.111G>A p.A37A Chr.1: 67705559-67705559 on Build GRCh38
4009669 c.99G>A p.S33S Chr.1: 67705571-67705571 on Build GRCh38
4938379 c.94-1G>C p.? Chr.1: 67705577-67705577 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs948498528 Chr.1: 67705248-67705248 on Build GRCh38
rs113817847 Chr.1: 67705417-67705417 on Build GRCh38
rs768444205 Chr.1: 67705531-67705531 on Build GRCh38
rs188229809 Chr.1: 67705206-67705206 on Build GRCh38
rs548359378 Chr.1: 67705264-67705264 on Build GRCh38
rs573708614 Chr.1: 67705422-67705422 on Build GRCh38
rs764681398 Chr.1: 67705572-67705572 on Build GRCh38
rs17130202 Chr.1: 67705532-67705532 on Build GRCh38
rs991298660 Chr.1: 67705582-67705582 on Build GRCh38
rs146253708 Chr.1: 67705534-67705534 on Build GRCh38
rs917092199 Chr.1: 67705584-67705584 on Build GRCh38
rs190580980 Chr.1: 67705348-67705348 on Build GRCh38
rs770955679 Chr.1: 67705285-67705285 on Build GRCh38
rs149098644 Chr.1: 67705727-67705727 on Build GRCh38
rs368515270 Chr.1: 67705434-67705434 on Build GRCh38
rs917126176 Chr.1: 67705212-67705212 on Build GRCh38
rs200225732 Chr.1: 67705540-67705540 on Build GRCh38
rs773725682 Chr.1: 67705286-67705286 on Build GRCh38
rs1046790622 Chr.1: 67705316-67705316 on Build GRCh38
rs751759729 Chr.1: 67705589-67705589 on Build GRCh38
rs778832059 Chr.1: 67705463-67705463 on Build GRCh38
rs781564846 Chr.1: 67705473-67705473 on Build GRCh38
rs554320263 Chr.1: 67705561-67705561 on Build GRCh38
rs949973868 Chr.1: 67705608-67705608 on Build GRCh38
rs765968908 Chr.1: 67705231-67705231 on Build GRCh38
rs376613159 Chr.1: 67705623-67705623 on Build GRCh38
rs80227853 Chr.1: 67705724-67705724 on Build GRCh38
rs760251491 Chr.1: 67705225-67705225 on Build GRCh38
rs111445182 Chr.1: 67705442-67705442 on Build GRCh38
rs909692593 Chr.1: 67705635-67705635 on Build GRCh38
rs543094266 Chr.1: 67705643-67705643 on Build GRCh38
rs753541054 Chr.1: 67705413-67705413 on Build GRCh38
rs751663804 Chr.1: 67705466-67705466 on Build GRCh38
rs369365588 Chr.1: 67705559-67705559 on Build GRCh38
rs373833781 Chr.1: 67705412-67705412 on Build GRCh38
rs560988139 Chr.1: 67705636-67705636 on Build GRCh38
rs778036855 Chr.1: 67705449-67705449 on Build GRCh38
rs765594443 Chr.1: 67705560-67705560 on Build GRCh38
rs571208080 Chr.1: 67705370-67705370 on Build GRCh38
rs942944514 Chr.1: 67705739-67705739 on Build GRCh38
rs918267184 Chr.1: 67705660-67705660 on Build GRCh38
rs372297619 Chr.1: 67705420-67705420 on Build GRCh38
rs187892406 Chr.1: 67705610-67705610 on Build GRCh38
rs568464156 Chr.1: 67705342-67705342 on Build GRCh38
rs542263823 Chr.1: 67705616-67705616 on Build GRCh38
rs554125849 Chr.1: 67705565-67705565 on Build GRCh38
rs536998272 Chr.1: 67705343-67705343 on Build GRCh38
rs950016282 Chr.1: 67705222-67705222 on Build GRCh38
rs749045743 Chr.1: 67705478-67705478 on Build GRCh38
rs917089160 Chr.1: 67705247-67705247 on Build GRCh38
rs924564719 Chr.1: 67705639-67705639 on Build GRCh38
rs1050988063 Chr.1: 67705685-67705685 on Build GRCh38
rs753197245 Chr.1: 67705429-67705429 on Build GRCh38
rs932085209 Chr.1: 67705675-67705675 on Build GRCh38
rs147603186 Chr.1: 67705552-67705552 on Build GRCh38
rs973103746 Chr.1: 67705542-67705542 on Build GRCh38
rs370289695 Chr.1: 67705656-67705656 on Build GRCh38
rs374150257 Chr.1: 67705428-67705428 on Build GRCh38
rs139638442 Chr.1: 67705533-67705533 on Build GRCh38
rs1022402829 Chr.1: 67705467-67705468 on Build GRCh38
rs893861483 Chr.1: 67705665-67705665 on Build GRCh38
rs994490691 Chr.1: 67705744-67705744 on Build GRCh38
rs11802819 Chr.1: 67705740-67705740 on Build GRCh38
rs890635567 Chr.1: 67705707-67705707 on Build GRCh38
rs757607138 Chr.1: 67705468-67705468 on Build GRCh38
rs757343160 Chr.1: 67705602-67705602 on Build GRCh38
rs370081170 Chr.1: 67705405-67705405 on Build GRCh38
rs890675821 Chr.1: 67705441-67705441 on Build GRCh38
rs763599665 Chr.1: 67705571-67705571 on Build GRCh38

View Full Product Details