Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 232770695-232771294 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIGYF2
Entrez Gene ID
26058
Gene Name
GRB10 interacting GYF protein 2
Gene Aliases
GYF2, PARK11, PERQ2, PERQ3, TNRC15
UniGene
Hs.565319
Gene Chromosome Location
Chr.2: 232697305-232860577 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Gene Symbol
KCNJ13
Entrez Gene ID
3769
Gene Name
potassium voltage-gated channel subfamily J member 13
Gene Aliases
KIR1.4, KIR7.1, LCA16, SVD
UniGene
Hs.467338
Gene Chromosome Location
Chr.2: 232765802-232776565 on Build GRCh38
Transcripts
NM_001103146.1 NM_001103147.1 NM_001103148.1 NM_015575.3 NR_103492.1 NM_001172416.1 NM_001172417.1 NM_002242.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6691129 c.154C>T p.R52C Chr.2: 232771209-232771209 on Build GRCh38
1017864 c.390C>T p.D130D Chr.2: 232770973-232770973 on Build GRCh38
4128215 c.126A>T p.R42R Chr.2: 232771237-232771237 on Build GRCh38
6691131 c.296G>C p.C99S Chr.2: 232771067-232771067 on Build GRCh38
1017865 c.222C>A p.L74L Chr.2: 232771141-232771141 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749946210 Chr.2: 232771155-232771155 on Build GRCh38
rs951275023 Chr.2: 232770773-232770773 on Build GRCh38
rs200415013 Chr.2: 232771239-232771239 on Build GRCh38
rs749097137 Chr.2: 232770957-232770957 on Build GRCh38
rs770352640 Chr.2: 232771222-232771222 on Build GRCh38
rs942118177 Chr.2: 232770719-232770719 on Build GRCh38
rs869320631 Chr.2: 232771205-232771205 on Build GRCh38
rs777396981 Chr.2: 232770889-232770889 on Build GRCh38
rs780804477 Chr.2: 232770883-232770883 on Build GRCh38
rs1026757774 Chr.2: 232771264-232771264 on Build GRCh38
rs1003135933 Chr.2: 232771159-232771159 on Build GRCh38
rs763508215 Chr.2: 232771258-232771258 on Build GRCh38
rs907341776 Chr.2: 232771093-232771093 on Build GRCh38
rs59336151 Chr.2: 232770728-232770729 on Build GRCh38
rs756066841 Chr.2: 232771156-232771156 on Build GRCh38
rs183766886 Chr.2: 232771094-232771094 on Build GRCh38
rs776705101 Chr.2: 232771058-232771058 on Build GRCh38
rs963732090 Chr.2: 232771162-232771162 on Build GRCh38
rs760000696 Chr.2: 232770874-232770875 on Build GRCh38
rs753616368 Chr.2: 232771170-232771170 on Build GRCh38
rs763335927 Chr.2: 232770858-232770869 on Build GRCh38
rs113733422 Chr.2: 232770728-232770728 on Build GRCh38
rs1020473832 Chr.2: 232770882-232770882 on Build GRCh38
rs747308840 Chr.2: 232770811-232770811 on Build GRCh38
rs570372184 Chr.2: 232770780-232770780 on Build GRCh38
rs769160785 Chr.2: 232771263-232771263 on Build GRCh38
rs767870732 Chr.2: 232770875-232770875 on Build GRCh38
rs778621880 Chr.2: 232771201-232771201 on Build GRCh38
rs77818131 Chr.2: 232771149-232771149 on Build GRCh38
rs763812532 Chr.2: 232770874-232770874 on Build GRCh38
rs761501599 Chr.2: 232771145-232771145 on Build GRCh38
rs539369632 Chr.2: 232771270-232771270 on Build GRCh38
rs1042298486 Chr.2: 232771075-232771075 on Build GRCh38
rs1050969759 Chr.2: 232770770-232770770 on Build GRCh38
rs760027526 Chr.2: 232771071-232771071 on Build GRCh38
rs113524903 Chr.2: 232770796-232770796 on Build GRCh38
rs765523438 Chr.2: 232770852-232770852 on Build GRCh38
rs748561212 Chr.2: 232770747-232770747 on Build GRCh38
rs534763826 Chr.2: 232770880-232770880 on Build GRCh38
rs755553780 Chr.2: 232770883-232770883 on Build GRCh38
rs772329921 Chr.2: 232770958-232770958 on Build GRCh38
rs1035865983 Chr.2: 232770949-232770949 on Build GRCh38
rs754789407 Chr.2: 232771192-232771192 on Build GRCh38
rs149725357 Chr.2: 232770925-232770925 on Build GRCh38
rs763341185 Chr.2: 232771115-232771115 on Build GRCh38
rs753283107 Chr.2: 232770883-232770884 on Build GRCh38
rs138305098 Chr.2: 232770815-232770815 on Build GRCh38
rs770239517 Chr.2: 232771246-232771246 on Build GRCh38
rs779781039 Chr.2: 232770956-232770956 on Build GRCh38
rs771136440 Chr.2: 232771217-232771217 on Build GRCh38
rs991734668 Chr.2: 232771092-232771092 on Build GRCh38
rs573097228 Chr.2: 232770846-232770847 on Build GRCh38
rs761628223 Chr.2: 232771273-232771273 on Build GRCh38
rs929791413 Chr.2: 232771245-232771245 on Build GRCh38
rs753425666 Chr.2: 232770921-232770921 on Build GRCh38
rs112079468 Chr.2: 232771019-232771019 on Build GRCh38
rs918408003 Chr.2: 232770743-232770743 on Build GRCh38
rs1054582760 Chr.2: 232770964-232770964 on Build GRCh38
rs528902715 Chr.2: 232770746-232770746 on Build GRCh38
rs745935897 Chr.2: 232771223-232771223 on Build GRCh38
rs747234040 Chr.2: 232771212-232771212 on Build GRCh38
rs972148230 Chr.2: 232771163-232771163 on Build GRCh38
rs777940941 Chr.2: 232770971-232770971 on Build GRCh38
rs751914348 Chr.2: 232770862-232770862 on Build GRCh38
rs766711095 Chr.2: 232770858-232770863 on Build GRCh38
rs753098916 Chr.2: 232770862-232770862 on Build GRCh38
rs371849984 Chr.2: 232771168-232771168 on Build GRCh38
rs747184742 Chr.2: 232770997-232770997 on Build GRCh38
rs764509761 Chr.2: 232771118-232771118 on Build GRCh38
rs946261593 Chr.2: 232771005-232771005 on Build GRCh38
rs863224884 Chr.2: 232770905-232770905 on Build GRCh38
rs397975119 Chr.2: 232770736-232770737 on Build GRCh38
rs758041173 Chr.2: 232770868-232770868 on Build GRCh38
rs894185751 Chr.2: 232771226-232771226 on Build GRCh38
rs891055056 Chr.2: 232770782-232770782 on Build GRCh38
rs1011309347 Chr.2: 232770823-232770823 on Build GRCh38
rs771011732 Chr.2: 232771048-232771048 on Build GRCh38
rs929815557 Chr.2: 232770745-232770745 on Build GRCh38
rs751152038 Chr.2: 232770878-232770878 on Build GRCh38
rs374810770 Chr.2: 232771125-232771125 on Build GRCh38
rs756015481 Chr.2: 232770934-232770934 on Build GRCh38
rs1027142931 Chr.2: 232770740-232770740 on Build GRCh38
rs983002935 Chr.2: 232770777-232770777 on Build GRCh38
rs965276572 Chr.2: 232770784-232770784 on Build GRCh38
rs975265480 Chr.2: 232770793-232770793 on Build GRCh38
rs903322709 Chr.2: 232770896-232770896 on Build GRCh38
rs371256707 Chr.2: 232771121-232771121 on Build GRCh38
rs786205550 Chr.2: 232771004-232771004 on Build GRCh38
rs776091698 Chr.2: 232771251-232771251 on Build GRCh38
rs777633027 Chr.2: 232770790-232770790 on Build GRCh38

View Full Product Details