Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 21325901-21326499 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRN2
Entrez Gene ID
22803
Gene Name
5'-3' exoribonuclease 2
Gene Aliases
-
UniGene
Hs.255932
Gene Chromosome Location
Chr.20: 21303284-21389825 on Build GRCh38
Transcripts
NM_001317960.1 NM_012255.4 XM_017027722.1 XM_017027723.1 XM_017027724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7240486 c.204-8C>G p.? Chr.20: 21326482-21326482 on Build GRCh38
1025088 c.116G>A p.S39N Chr.20: 21326319-21326319 on Build GRCh38
6381197 c.132T>A p.N44K Chr.20: 21326335-21326335 on Build GRCh38
4741760 c.164T>C p.M55T Chr.20: 21326367-21326367 on Build GRCh38
1025089 c.200A>G p.D67G Chr.20: 21326403-21326403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs991023733 Chr.20: 21326235-21326235 on Build GRCh38
rs370240383 Chr.20: 21326109-21326109 on Build GRCh38
rs745512832 Chr.20: 21326382-21326382 on Build GRCh38
rs976824143 Chr.20: 21326200-21326200 on Build GRCh38
rs759752650 Chr.20: 21326454-21326454 on Build GRCh38
rs375675074 Chr.20: 21326145-21326147 on Build GRCh38
rs773357284 Chr.20: 21326242-21326242 on Build GRCh38
rs369080895 Chr.20: 21326257-21326257 on Build GRCh38
rs548727737 Chr.20: 21326198-21326198 on Build GRCh38
rs773612171 Chr.20: 21326147-21326147 on Build GRCh38
rs766974620 Chr.20: 21325972-21325973 on Build GRCh38
rs1032888589 Chr.20: 21326161-21326161 on Build GRCh38
rs551757662 Chr.20: 21326468-21326468 on Build GRCh38
rs770038508 Chr.20: 21326065-21326065 on Build GRCh38
rs961722692 Chr.20: 21326091-21326091 on Build GRCh38
rs904813717 Chr.20: 21326048-21326048 on Build GRCh38
rs925954912 Chr.20: 21326410-21326410 on Build GRCh38
rs547166916 Chr.20: 21326102-21326102 on Build GRCh38
rs759714778 Chr.20: 21326265-21326265 on Build GRCh38
rs771701387 Chr.20: 21326032-21326032 on Build GRCh38
rs756791504 Chr.20: 21326326-21326326 on Build GRCh38
rs78094536 Chr.20: 21326166-21326166 on Build GRCh38
rs958221753 Chr.20: 21326216-21326216 on Build GRCh38
rs370308584 Chr.20: 21326462-21326462 on Build GRCh38
rs780608516 Chr.20: 21326375-21326375 on Build GRCh38
rs765385322 Chr.20: 21326456-21326456 on Build GRCh38
rs898332100 Chr.20: 21325945-21325945 on Build GRCh38
rs773841644 Chr.20: 21326430-21326430 on Build GRCh38
rs775211380 Chr.20: 21326457-21326457 on Build GRCh38
rs528603061 Chr.20: 21326017-21326017 on Build GRCh38
rs374750927 Chr.20: 21326418-21326418 on Build GRCh38
rs751559463 Chr.20: 21326295-21326295 on Build GRCh38
rs894638013 Chr.20: 21326065-21326066 on Build GRCh38
rs760850691 Chr.20: 21326253-21326253 on Build GRCh38
rs753353121 Chr.20: 21326249-21326249 on Build GRCh38
rs189255798 Chr.20: 21326428-21326428 on Build GRCh38
rs752508946 Chr.20: 21326281-21326281 on Build GRCh38
rs950220040 Chr.20: 21326446-21326446 on Build GRCh38
rs764036284 Chr.20: 21326292-21326292 on Build GRCh38
rs35542728 Chr.20: 21325992-21325993 on Build GRCh38
rs373315919 Chr.20: 21326289-21326289 on Build GRCh38
rs776903908 Chr.20: 21326258-21326258 on Build GRCh38
rs752360814 Chr.20: 21326452-21326453 on Build GRCh38
rs993133352 Chr.20: 21326106-21326106 on Build GRCh38
rs2296728 Chr.20: 21326469-21326469 on Build GRCh38
rs899281284 Chr.20: 21326466-21326466 on Build GRCh38
rs776620358 Chr.20: 21326435-21326435 on Build GRCh38
rs754499506 Chr.20: 21326267-21326267 on Build GRCh38
rs1037249598 Chr.20: 21325930-21325930 on Build GRCh38
rs889739934 Chr.20: 21326055-21326055 on Build GRCh38
rs779383651 Chr.20: 21326417-21326417 on Build GRCh38
rs547728016 Chr.20: 21326434-21326434 on Build GRCh38
rs1024019890 Chr.20: 21326082-21326082 on Build GRCh38
rs202127993 Chr.20: 21326463-21326463 on Build GRCh38
rs949647991 Chr.20: 21326333-21326333 on Build GRCh38
rs965866405 Chr.20: 21326153-21326153 on Build GRCh38
rs765056802 Chr.20: 21326266-21326266 on Build GRCh38
rs760439820 Chr.20: 21326064-21326064 on Build GRCh38
rs922660689 Chr.20: 21326211-21326211 on Build GRCh38
rs376549093 Chr.20: 21326416-21326416 on Build GRCh38
rs764915313 Chr.20: 21326447-21326451 on Build GRCh38
rs775006405 Chr.20: 21326056-21326056 on Build GRCh38
rs531716074 Chr.20: 21326148-21326148 on Build GRCh38
rs995796862 Chr.20: 21325988-21325988 on Build GRCh38
rs79960785 Chr.20: 21326058-21326058 on Build GRCh38
rs940733192 Chr.20: 21326451-21326451 on Build GRCh38
rs1028189216 Chr.20: 21326026-21326026 on Build GRCh38
rs376927991 Chr.20: 21326433-21326433 on Build GRCh38
rs142962022 Chr.20: 21326090-21326090 on Build GRCh38

View Full Product Details