Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 25732224-25732728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NGLY1
Entrez Gene ID
55768
Gene Name
N-glycanase 1
Gene Aliases
CDDG, CDG1V, PNG1, PNGase
UniGene
Hs.368960
Gene Chromosome Location
Chr.3: 25718944-25790039 on Build GRCh38
Transcripts
NM_001145293.1 NM_001145294.1 NM_001145295.1 NM_018297.3 XM_005265316.1 XM_005265317.1 XM_011533944.1 XM_017006839.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2948852 c.1333A>C p.I445L Chr.3: 25732411-25732411 on Build GRCh38
6048884 c.1398A>T p.R466S Chr.3: 25732346-25732346 on Build GRCh38
3696011 c.1279A>C p.I427L Chr.3: 25732411-25732411 on Build GRCh38
3714518 c.1307G>T p.R436M Chr.3: 25732437-25732437 on Build GRCh38
4465880 c.1404C>T p.A468A Chr.3: 25732340-25732340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054776263 Chr.3: 25732702-25732702 on Build GRCh38
rs748956391 Chr.3: 25732391-25732391 on Build GRCh38
rs369598638 Chr.3: 25732476-25732476 on Build GRCh38
rs372323207 Chr.3: 25732440-25732440 on Build GRCh38
rs909705419 Chr.3: 25732564-25732564 on Build GRCh38
rs770565316 Chr.3: 25732511-25732511 on Build GRCh38
rs900192738 Chr.3: 25732604-25732604 on Build GRCh38
rs750639430 Chr.3: 25732469-25732469 on Build GRCh38
rs748868283 Chr.3: 25732498-25732498 on Build GRCh38
rs927838172 Chr.3: 25732661-25732661 on Build GRCh38
rs927617505 Chr.3: 25732281-25732281 on Build GRCh38
rs770653213 Chr.3: 25732402-25732402 on Build GRCh38
rs754714582 Chr.3: 25732380-25732380 on Build GRCh38
rs543901983 Chr.3: 25732426-25732426 on Build GRCh38
rs984344472 Chr.3: 25732248-25732248 on Build GRCh38
rs745483470 Chr.3: 25732524-25732524 on Build GRCh38
rs762647602 Chr.3: 25732303-25732303 on Build GRCh38
rs939017219 Chr.3: 25732700-25732700 on Build GRCh38
rs201859775 Chr.3: 25732531-25732531 on Build GRCh38
rs995835831 Chr.3: 25732616-25732616 on Build GRCh38
rs112241790 Chr.3: 25732703-25732703 on Build GRCh38
rs759978728 Chr.3: 25732335-25732335 on Build GRCh38
rs939925055 Chr.3: 25732254-25732254 on Build GRCh38
rs777326126 Chr.3: 25732534-25732534 on Build GRCh38
rs199898452 Chr.3: 25732474-25732474 on Build GRCh38
rs370304224 Chr.3: 25732532-25732532 on Build GRCh38
rs375966958 Chr.3: 25732381-25732381 on Build GRCh38
rs111544055 Chr.3: 25732328-25732328 on Build GRCh38
rs768131676 Chr.3: 25732339-25732339 on Build GRCh38
rs899066280 Chr.3: 25732514-25732514 on Build GRCh38
rs774184276 Chr.3: 25732512-25732512 on Build GRCh38
rs879185450 Chr.3: 25732629-25732629 on Build GRCh38
rs192062614 Chr.3: 25732494-25732494 on Build GRCh38
rs938966609 Chr.3: 25732361-25732361 on Build GRCh38
rs761443165 Chr.3: 25732290-25732290 on Build GRCh38
rs761035118 Chr.3: 25732444-25732444 on Build GRCh38
rs577617186 Chr.3: 25732667-25732667 on Build GRCh38
rs563624865 Chr.3: 25732633-25732635 on Build GRCh38
rs984008061 Chr.3: 25732553-25732553 on Build GRCh38
rs754079493 Chr.3: 25732356-25732356 on Build GRCh38
rs766881984 Chr.3: 25732325-25732325 on Build GRCh38
rs772810347 Chr.3: 25732390-25732390 on Build GRCh38
rs772506281 Chr.3: 25732526-25732526 on Build GRCh38
rs756409700 Chr.3: 25732517-25732517 on Build GRCh38
rs776016452 Chr.3: 25732278-25732278 on Build GRCh38
rs765500224 Chr.3: 25732467-25732467 on Build GRCh38
rs1043604336 Chr.3: 25732441-25732441 on Build GRCh38
rs764056387 Chr.3: 25732354-25732354 on Build GRCh38
rs753137266 Chr.3: 25732344-25732344 on Build GRCh38
rs767512465 Chr.3: 25732396-25732397 on Build GRCh38
rs201442228 Chr.3: 25732455-25732455 on Build GRCh38
rs769156478 Chr.3: 25732295-25732295 on Build GRCh38
rs768571298 Chr.3: 25732272-25732272 on Build GRCh38
rs773050159 Chr.3: 25732299-25732299 on Build GRCh38
rs779826640 Chr.3: 25732369-25732369 on Build GRCh38
rs752759931 Chr.3: 25732501-25732502 on Build GRCh38
rs767739104 Chr.3: 25732439-25732439 on Build GRCh38
rs558929301 Chr.3: 25732582-25732582 on Build GRCh38
rs144144643 Chr.3: 25732411-25732411 on Build GRCh38
rs753847475 Chr.3: 25732447-25732447 on Build GRCh38
rs972655047 Chr.3: 25732627-25732627 on Build GRCh38
rs187892679 Chr.3: 25732389-25732389 on Build GRCh38
rs999701051 Chr.3: 25732682-25732682 on Build GRCh38
rs79372214 Chr.3: 25732269-25732269 on Build GRCh38
rs746748010 Chr.3: 25732378-25732378 on Build GRCh38
rs992161646 Chr.3: 25732450-25732450 on Build GRCh38
rs751908554 Chr.3: 25732334-25732334 on Build GRCh38
rs760016381 Chr.3: 25732423-25732423 on Build GRCh38
rs998119300 Chr.3: 25732502-25732502 on Build GRCh38
rs587777265 Chr.3: 25732374-25732375 on Build GRCh38
rs755940005 Chr.3: 25732477-25732477 on Build GRCh38
rs757430160 Chr.3: 25732364-25732364 on Build GRCh38
rs930499233 Chr.3: 25732520-25732520 on Build GRCh38
rs754852315 Chr.3: 25732473-25732473 on Build GRCh38
rs909832253 Chr.3: 25732249-25732249 on Build GRCh38
rs756254761 Chr.3: 25732350-25732350 on Build GRCh38

View Full Product Details