Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 127986569-127987118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAP130
Entrez Gene ID
79595
Gene Name
Sin3A associated protein 130
Gene Aliases
-
UniGene
Hs.32995
Gene Chromosome Location
Chr.2: 127941217-128028093 on Build GRCh38
Transcripts
NM_001145928.1 NM_024545.3 XM_005263767.3 XM_005263768.3 XM_005263769.3 XM_005263770.3 XM_006712748.3 XM_006712749.3 XM_017004903.1 XM_017004904.1 XM_017004905.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6771818 c.1994C>T p.S665L Chr.2: 127986827-127986827 on Build GRCh38
5394304 c.1891G>A p.A631T Chr.2: 127986930-127986930 on Build GRCh38
6046765 c.1870G>A p.A624T Chr.2: 127986951-127986951 on Build GRCh38
4416305 c.1938C>T p.T646T Chr.2: 127986883-127986883 on Build GRCh38
6771810 c.1939G>A p.V647M Chr.2: 127986882-127986882 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760801637 Chr.2: 127986992-127986992 on Build GRCh38
rs747959672 Chr.2: 127986761-127986761 on Build GRCh38
rs545472828 Chr.2: 127986879-127986879 on Build GRCh38
rs867544407 Chr.2: 127986768-127986768 on Build GRCh38
rs1030909924 Chr.2: 127987025-127987025 on Build GRCh38
rs370863669 Chr.2: 127986974-127986974 on Build GRCh38
rs768049712 Chr.2: 127986802-127986802 on Build GRCh38
rs756500380 Chr.2: 127986829-127986829 on Build GRCh38
rs775476794 Chr.2: 127986987-127986987 on Build GRCh38
rs752275112 Chr.2: 127986950-127986950 on Build GRCh38
rs746901110 Chr.2: 127986599-127986599 on Build GRCh38
rs575486225 Chr.2: 127986728-127986728 on Build GRCh38
rs772996183 Chr.2: 127986774-127986774 on Build GRCh38
rs749113764 Chr.2: 127986882-127986882 on Build GRCh38
rs772113827 Chr.2: 127986901-127986901 on Build GRCh38
rs754662337 Chr.2: 127986939-127986939 on Build GRCh38
rs146525786 Chr.2: 127986796-127986796 on Build GRCh38
rs777703108 Chr.2: 127986596-127986596 on Build GRCh38
rs758675517 Chr.2: 127986713-127986714 on Build GRCh38
rs72841716 Chr.2: 127986906-127986906 on Build GRCh38
rs764491200 Chr.2: 127986830-127986830 on Build GRCh38
rs559114703 Chr.2: 127986646-127986646 on Build GRCh38
rs368027557 Chr.2: 127986966-127986966 on Build GRCh38
rs937273151 Chr.2: 127986622-127986622 on Build GRCh38
rs776479486 Chr.2: 127986757-127986757 on Build GRCh38
rs759651122 Chr.2: 127986897-127986897 on Build GRCh38
rs559677024 Chr.2: 127986843-127986843 on Build GRCh38
rs746529480 Chr.2: 127986744-127986744 on Build GRCh38
rs71420815 Chr.2: 127987043-127987043 on Build GRCh38
rs753205036 Chr.2: 127986827-127986827 on Build GRCh38
rs754273324 Chr.2: 127986840-127986840 on Build GRCh38
rs1055355487 Chr.2: 127986644-127986644 on Build GRCh38
rs781184042 Chr.2: 127986857-127986857 on Build GRCh38
rs200023786 Chr.2: 127986766-127986766 on Build GRCh38
rs267598865 Chr.2: 127986912-127986912 on Build GRCh38
rs75485150 Chr.2: 127986740-127986740 on Build GRCh38
rs3732196 Chr.2: 127986883-127986883 on Build GRCh38
rs550940612 Chr.2: 127986946-127986946 on Build GRCh38
rs751235591 Chr.2: 127986935-127986935 on Build GRCh38
rs371015231 Chr.2: 127986969-127986969 on Build GRCh38
rs374888723 Chr.2: 127986926-127986926 on Build GRCh38
rs762366012 Chr.2: 127987036-127987036 on Build GRCh38
rs764726128 Chr.2: 127986921-127986921 on Build GRCh38
rs896837630 Chr.2: 127986673-127986673 on Build GRCh38
rs1036620115 Chr.2: 127986651-127986651 on Build GRCh38
rs777496589 Chr.2: 127986881-127986881 on Build GRCh38
rs960646831 Chr.2: 127987049-127987049 on Build GRCh38
rs997795755 Chr.2: 127986960-127986960 on Build GRCh38
rs182681063 Chr.2: 127986847-127986847 on Build GRCh38
rs953960272 Chr.2: 127986731-127986731 on Build GRCh38
rs746139735 Chr.2: 127986973-127986973 on Build GRCh38
rs769542706 Chr.2: 127986763-127986763 on Build GRCh38
rs372670192 Chr.2: 127986743-127986743 on Build GRCh38
rs143177451 Chr.2: 127986877-127986877 on Build GRCh38
rs1022589834 Chr.2: 127986742-127986742 on Build GRCh38
rs113833801 Chr.2: 127986767-127986767 on Build GRCh38
rs763104740 Chr.2: 127986776-127986776 on Build GRCh38
rs756006581 Chr.2: 127986959-127986959 on Build GRCh38
rs992169360 Chr.2: 127987095-127987095 on Build GRCh38
rs759576173 Chr.2: 127986799-127986799 on Build GRCh38
rs990584204 Chr.2: 127986869-127986869 on Build GRCh38
rs751056874 Chr.2: 127986846-127986846 on Build GRCh38
rs200408275 Chr.2: 127986914-127986914 on Build GRCh38
rs765567183 Chr.2: 127986868-127986868 on Build GRCh38
rs754220405 Chr.2: 127986925-127986925 on Build GRCh38
rs766611393 Chr.2: 127986793-127986793 on Build GRCh38
rs753499224 Chr.2: 127987056-127987056 on Build GRCh38
rs34467505 Chr.2: 127986755-127986755 on Build GRCh38
rs2948068 Chr.2: 127987006-127987006 on Build GRCh38
rs373598411 Chr.2: 127986597-127986597 on Build GRCh38
rs758081189 Chr.2: 127986841-127986841 on Build GRCh38
rs766114286 Chr.2: 127986933-127986933 on Build GRCh38
rs941694296 Chr.2: 127987088-127987088 on Build GRCh38
rs751866039 Chr.2: 127986869-127986874 on Build GRCh38
rs937222204 Chr.2: 127987003-127987003 on Build GRCh38
rs768042998 Chr.2: 127986711-127986711 on Build GRCh38
rs888032540 Chr.2: 127986593-127986593 on Build GRCh38

View Full Product Details