Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 189450707-189451255 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WDR75
Entrez Gene ID
84128
Gene Name
WD repeat domain 75
Gene Aliases
NET16, UTP17
UniGene
Hs.399984
Gene Chromosome Location
Chr.2: 189441433-189475562 on Build GRCh38
Transcripts
NM_001303096.1 NM_032168.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1186074 c.255G>A p.W85* Chr.2: 189450941-189450941 on Build GRCh38
6089764 c.273C>T p.I91I Chr.2: 189450959-189450959 on Build GRCh38
1162804 c.268G>A p.G90S Chr.2: 189450954-189450954 on Build GRCh38
7423872 c.218T>G p.L73R Chr.2: 189450904-189450904 on Build GRCh38
1013989 c.221A>G p.Y74C Chr.2: 189450907-189450907 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745333053 Chr.2: 189450887-189450887 on Build GRCh38
rs756969134 Chr.2: 189451013-189451013 on Build GRCh38
rs775136502 Chr.2: 189451079-189451079 on Build GRCh38
rs778401644 Chr.2: 189451015-189451015 on Build GRCh38
rs1026975818 Chr.2: 189450804-189450804 on Build GRCh38
rs774743436 Chr.2: 189451003-189451004 on Build GRCh38
rs988917308 Chr.2: 189450899-189450899 on Build GRCh38
rs570840418 Chr.2: 189451040-189451040 on Build GRCh38
rs950345487 Chr.2: 189450877-189450877 on Build GRCh38
rs569059516 Chr.2: 189451213-189451213 on Build GRCh38
rs547299676 Chr.2: 189451135-189451135 on Build GRCh38
rs373310728 Chr.2: 189450975-189450975 on Build GRCh38
rs759510371 Chr.2: 189450993-189450993 on Build GRCh38
rs565945538 Chr.2: 189450885-189450885 on Build GRCh38
rs12988354 Chr.2: 189450845-189450845 on Build GRCh38
rs901244680 Chr.2: 189451071-189451071 on Build GRCh38
rs748940818 Chr.2: 189450873-189450873 on Build GRCh38
rs535688660 Chr.2: 189451186-189451186 on Build GRCh38
rs371624897 Chr.2: 189450936-189450936 on Build GRCh38
rs762434495 Chr.2: 189450948-189450948 on Build GRCh38
rs10178969 Chr.2: 189450997-189450997 on Build GRCh38
rs13012233 Chr.2: 189450811-189450811 on Build GRCh38
rs774529200 Chr.2: 189450982-189450982 on Build GRCh38
rs1006974732 Chr.2: 189450860-189450860 on Build GRCh38
rs770279398 Chr.2: 189450949-189450949 on Build GRCh38
rs572718447 Chr.2: 189450932-189450932 on Build GRCh38
rs76793461 Chr.2: 189451016-189451016 on Build GRCh38
rs200268180 Chr.2: 189450919-189450919 on Build GRCh38
rs13016910 Chr.2: 189450957-189450957 on Build GRCh38
rs373151228 Chr.2: 189450863-189450863 on Build GRCh38
rs563820335 Chr.2: 189451005-189451005 on Build GRCh38
rs968428038 Chr.2: 189451226-189451226 on Build GRCh38
rs540544797 Chr.2: 189450938-189450938 on Build GRCh38
rs1018376415 Chr.2: 189451064-189451064 on Build GRCh38
rs867981996 Chr.2: 189450961-189450961 on Build GRCh38
rs952943522 Chr.2: 189450817-189450817 on Build GRCh38
rs12988115 Chr.2: 189450747-189450747 on Build GRCh38
rs547737562 Chr.2: 189450883-189450883 on Build GRCh38
rs10202780 Chr.2: 189450994-189450994 on Build GRCh38
rs956990172 Chr.2: 189451151-189451151 on Build GRCh38
rs544410873 Chr.2: 189451003-189451003 on Build GRCh38
rs1011530337 Chr.2: 189451200-189451200 on Build GRCh38
rs115130066 Chr.2: 189451027-189451027 on Build GRCh38
rs557127925 Chr.2: 189451210-189451210 on Build GRCh38
rs976863180 Chr.2: 189450876-189450876 on Build GRCh38
rs947003033 Chr.2: 189450937-189450937 on Build GRCh38
rs758396443 Chr.2: 189450822-189450822 on Build GRCh38
rs12993393 Chr.2: 189450884-189450884 on Build GRCh38
rs777466052 Chr.2: 189450871-189450871 on Build GRCh38
rs74537192 Chr.2: 189450882-189450882 on Build GRCh38
rs1035811818 Chr.2: 189451228-189451228 on Build GRCh38
rs115060734 Chr.2: 189451046-189451046 on Build GRCh38
rs773645303 Chr.2: 189450968-189450968 on Build GRCh38
rs766470637 Chr.2: 189450981-189450981 on Build GRCh38
rs1022892446 Chr.2: 189451205-189451205 on Build GRCh38
rs905667871 Chr.2: 189451209-189451209 on Build GRCh38
rs539164673 Chr.2: 189451227-189451227 on Build GRCh38
rs10202907 Chr.2: 189451146-189451146 on Build GRCh38

View Full Product Details