Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 208238748-208239223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IDH1
Entrez Gene ID
3417
Gene Name
isocitrate dehydrogenase (NADP(+)) 1, cytosolic
Gene Aliases
HEL-216, HEL-S-26, IDCD, IDH, IDP, IDPC, PICD
UniGene
Hs.593422
Gene Chromosome Location
Chr.2: 208236227-208255143 on Build GRCh38
Transcripts
NM_001282386.1 NM_001282387.1 NM_005896.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
325527 c.1138A>T p.I380F Chr.2: 208239087-208239087 on Build GRCh38
6959401 c.1016G>A p.G339E Chr.2: 208239209-208239209 on Build GRCh38
6926764 c.1013G>A p.R338K Chr.2: 208239212-208239212 on Build GRCh38
6979528 c.1046A>G p.N349S Chr.2: 208239179-208239179 on Build GRCh38
6958798 c.1082_1084delAAG p.E361delE Chr.2: 208239141-208239143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs917070025 Chr.2: 208238804-208238804 on Build GRCh38
rs9646838 Chr.2: 208238771-208238771 on Build GRCh38
rs765282142 Chr.2: 208239110-208239110 on Build GRCh38
rs201267625 Chr.2: 208239105-208239105 on Build GRCh38
rs974630737 Chr.2: 208238803-208238803 on Build GRCh38
rs775623666 Chr.2: 208239072-208239072 on Build GRCh38
rs752187592 Chr.2: 208239119-208239119 on Build GRCh38
rs777025307 Chr.2: 208239060-208239060 on Build GRCh38
rs760003909 Chr.2: 208239062-208239062 on Build GRCh38
rs200718328 Chr.2: 208239181-208239181 on Build GRCh38
rs1019914547 Chr.2: 208238802-208238802 on Build GRCh38
rs186944298 Chr.2: 208239057-208239057 on Build GRCh38
rs564930424 Chr.2: 208238769-208238769 on Build GRCh38
rs371837057 Chr.2: 208239044-208239044 on Build GRCh38
rs746615557 Chr.2: 208239127-208239127 on Build GRCh38
rs965361225 Chr.2: 208238906-208238906 on Build GRCh38
rs775893896 Chr.2: 208239149-208239149 on Build GRCh38
rs939533739 Chr.2: 208239007-208239007 on Build GRCh38
rs756940135 Chr.2: 208239130-208239130 on Build GRCh38
rs529117793 Chr.2: 208238969-208238969 on Build GRCh38
rs766246232 Chr.2: 208239092-208239092 on Build GRCh38
rs182206511 Chr.2: 208238829-208238829 on Build GRCh38
rs1042867599 Chr.2: 208238979-208238979 on Build GRCh38
rs934520634 Chr.2: 208239016-208239016 on Build GRCh38
rs762867621 Chr.2: 208239150-208239150 on Build GRCh38
rs191864560 Chr.2: 208238801-208238801 on Build GRCh38
rs772278481 Chr.2: 208238820-208238820 on Build GRCh38
rs764338499 Chr.2: 208239074-208239074 on Build GRCh38
rs775128585 Chr.2: 208239096-208239096 on Build GRCh38
rs770226824 Chr.2: 208239138-208239138 on Build GRCh38
rs776513405 Chr.2: 208239173-208239173 on Build GRCh38
rs777431979 Chr.2: 208239126-208239126 on Build GRCh38
rs773506444 Chr.2: 208238835-208238835 on Build GRCh38
rs766547920 Chr.2: 208239153-208239153 on Build GRCh38
rs200591678 Chr.2: 208239125-208239125 on Build GRCh38
rs995012431 Chr.2: 208238787-208238787 on Build GRCh38
rs762143982 Chr.2: 208239084-208239084 on Build GRCh38
rs369234651 Chr.2: 208239106-208239106 on Build GRCh38
rs777467520 Chr.2: 208238807-208238807 on Build GRCh38
rs989492384 Chr.2: 208238856-208238856 on Build GRCh38
rs935939155 Chr.2: 208238848-208238848 on Build GRCh38
rs536614679 Chr.2: 208239066-208239066 on Build GRCh38
rs975375815 Chr.2: 208238927-208238927 on Build GRCh38
rs372711897 Chr.2: 208239135-208239135 on Build GRCh38
rs566023900 Chr.2: 208239004-208239004 on Build GRCh38
rs199743726 Chr.2: 208239054-208239054 on Build GRCh38
rs762954264 Chr.2: 208239073-208239073 on Build GRCh38
rs141895800 Chr.2: 208239076-208239076 on Build GRCh38
rs536880332 Chr.2: 208239165-208239165 on Build GRCh38
rs112625205 Chr.2: 208239024-208239024 on Build GRCh38
rs921265831 Chr.2: 208238954-208238954 on Build GRCh38
rs771701369 Chr.2: 208239046-208239046 on Build GRCh38
rs754957038 Chr.2: 208239107-208239107 on Build GRCh38
rs763941915 Chr.2: 208239028-208239029 on Build GRCh38
rs759413310 Chr.2: 208239188-208239188 on Build GRCh38
rs375389985 Chr.2: 208239065-208239065 on Build GRCh38
rs750457328 Chr.2: 208239064-208239065 on Build GRCh38
rs1018752106 Chr.2: 208238788-208238788 on Build GRCh38
rs748230942 Chr.2: 208238799-208238799 on Build GRCh38
rs768596300 Chr.2: 208239158-208239158 on Build GRCh38
rs781533021 Chr.2: 208239131-208239131 on Build GRCh38
rs765179388 Chr.2: 208239203-208239203 on Build GRCh38
rs763043045 Chr.2: 208239081-208239082 on Build GRCh38
rs367959618 Chr.2: 208239030-208239030 on Build GRCh38
rs548881825 Chr.2: 208238975-208238975 on Build GRCh38
rs951659793 Chr.2: 208238793-208238793 on Build GRCh38
rs376666493 Chr.2: 208239045-208239045 on Build GRCh38

View Full Product Details