Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 97465127-97465690 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAP2A
Entrez Gene ID
5911
Gene Name
RAP2A, member of RAS oncogene family
Gene Aliases
K-REV, KREV, RAP2, RbBP-30
UniGene
Hs.508480
Gene Chromosome Location
Chr.13: 97434221-97467998 on Build GRCh38
Transcripts
NM_021033.6

SNP Detail

SNP ID Chromosome Location
rs1010546675 Chr.13: 97465352-97465352 on Build GRCh38
rs911700490 Chr.13: 97465510-97465510 on Build GRCh38
rs983410836 Chr.13: 97465508-97465508 on Build GRCh38
rs1032255867 Chr.13: 97465408-97465408 on Build GRCh38
rs775892934 Chr.13: 97465532-97465532 on Build GRCh38
rs900265065 Chr.13: 97465549-97465549 on Build GRCh38
rs929204314 Chr.13: 97465507-97465507 on Build GRCh38
rs574516483 Chr.13: 97465197-97465197 on Build GRCh38
rs973408129 Chr.13: 97465481-97465481 on Build GRCh38
rs930981206 Chr.13: 97465446-97465446 on Build GRCh38
rs914818817 Chr.13: 97465365-97465365 on Build GRCh38
rs932830917 Chr.13: 97465367-97465367 on Build GRCh38
rs1031649910 Chr.13: 97465290-97465290 on Build GRCh38
rs896839121 Chr.13: 97465201-97465201 on Build GRCh38
rs1012064806 Chr.13: 97465488-97465488 on Build GRCh38
rs749324737 Chr.13: 97465353-97465353 on Build GRCh38
rs948710886 Chr.13: 97465198-97465198 on Build GRCh38
rs1036758658 Chr.13: 97465428-97465428 on Build GRCh38
rs1021794108 Chr.13: 97465363-97465363 on Build GRCh38
rs57876065 Chr.13: 97465523-97465523 on Build GRCh38
rs771065812 Chr.13: 97465379-97465379 on Build GRCh38
rs906379270 Chr.13: 97465513-97465513 on Build GRCh38
rs560054682 Chr.13: 97465463-97465463 on Build GRCh38
rs540327897 Chr.13: 97465439-97465439 on Build GRCh38
rs920513247 Chr.13: 97465233-97465233 on Build GRCh38
rs981428600 Chr.13: 97465282-97465282 on Build GRCh38
rs946352374 Chr.13: 97465568-97465571 on Build GRCh38
rs57530477 Chr.13: 97465518-97465518 on Build GRCh38
rs995411840 Chr.13: 97465206-97465206 on Build GRCh38
rs895095163 Chr.13: 97465457-97465457 on Build GRCh38
rs550430297 Chr.13: 97465195-97465195 on Build GRCh38
rs1012529310 Chr.13: 97465248-97465248 on Build GRCh38
rs563349730 Chr.13: 97465456-97465456 on Build GRCh38
rs777725275 Chr.13: 97465356-97465356 on Build GRCh38
rs765979212 Chr.13: 97465590-97465591 on Build GRCh38
rs1009044493 Chr.13: 97465275-97465275 on Build GRCh38
rs1031765949 Chr.13: 97465603-97465603 on Build GRCh38
rs553860765 Chr.13: 97465630-97465630 on Build GRCh38
rs749153769 Chr.13: 97465429-97465429 on Build GRCh38
rs929088173 Chr.13: 97465235-97465235 on Build GRCh38
rs1031954504 Chr.13: 97465633-97465633 on Build GRCh38
rs867288525 Chr.13: 97465199-97465199 on Build GRCh38
rs1023053364 Chr.13: 97465252-97465252 on Build GRCh38
rs746428535 Chr.13: 97465426-97465426 on Build GRCh38
rs956044897 Chr.13: 97465299-97465299 on Build GRCh38
rs772305889 Chr.13: 97465490-97465490 on Build GRCh38
rs574442402 Chr.13: 97465399-97465399 on Build GRCh38
rs890333663 Chr.13: 97465260-97465260 on Build GRCh38
rs902227951 Chr.13: 97465627-97465627 on Build GRCh38
rs894944082 Chr.13: 97465223-97465223 on Build GRCh38
rs900148953 Chr.13: 97465332-97465332 on Build GRCh38
rs1048012588 Chr.13: 97465250-97465250 on Build GRCh38
rs774683131 Chr.13: 97465382-97465382 on Build GRCh38
rs971496125 Chr.13: 97465263-97465263 on Build GRCh38
rs1027319708 Chr.13: 97465215-97465215 on Build GRCh38
rs1043367006 Chr.13: 97465575-97465575 on Build GRCh38
rs113677896 Chr.13: 97465535-97465535 on Build GRCh38

View Full Product Details