Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77105563-77106008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIPC
Entrez Gene ID
85457
Gene Name
CLOCK interacting pacemaker
Gene Aliases
KIAA1737
UniGene
Hs.709066
Gene Chromosome Location
Chr.14: 77098235-77117287 on Build GRCh38
Transcripts
NM_033426.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5419277 c.16C>T p.P6S Chr.14: 77105724-77105724 on Build GRCh38
5648708 c.86G>C p.R29P Chr.14: 77105794-77105794 on Build GRCh38
6694445 c.94G>A p.G32R Chr.14: 77105802-77105802 on Build GRCh38
236769 c.45_46insT p.A16fs*22 Chr.14: 77105753-77105754 on Build GRCh38
699318 c.44C>A p.S15Y Chr.14: 77105752-77105752 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11552006 Chr.14: 77105799-77105799 on Build GRCh38
rs1053237000 Chr.14: 77105606-77105606 on Build GRCh38
rs759899870 Chr.14: 77105699-77105699 on Build GRCh38
rs1004813212 Chr.14: 77105657-77105657 on Build GRCh38
rs376431401 Chr.14: 77105673-77105673 on Build GRCh38
rs747016504 Chr.14: 77105794-77105794 on Build GRCh38
rs746333424 Chr.14: 77105797-77105797 on Build GRCh38
rs923818316 Chr.14: 77105884-77105884 on Build GRCh38
rs749932413 Chr.14: 77105736-77105736 on Build GRCh38
rs7492304 Chr.14: 77105898-77105898 on Build GRCh38
rs761349795 Chr.14: 77105717-77105717 on Build GRCh38
rs772315030 Chr.14: 77105659-77105659 on Build GRCh38
rs778063251 Chr.14: 77105783-77105783 on Build GRCh38
rs571921022 Chr.14: 77105778-77105778 on Build GRCh38
rs766333104 Chr.14: 77105858-77105858 on Build GRCh38
rs576066281 Chr.14: 77105654-77105654 on Build GRCh38
rs184833962 Chr.14: 77105682-77105682 on Build GRCh38
rs964218840 Chr.14: 77105864-77105864 on Build GRCh38
rs967903089 Chr.14: 77105810-77105810 on Build GRCh38
rs770246405 Chr.14: 77105804-77105804 on Build GRCh38
rs766245014 Chr.14: 77105748-77105748 on Build GRCh38
rs765134581 Chr.14: 77105886-77105886 on Build GRCh38
rs913140111 Chr.14: 77105653-77105653 on Build GRCh38
rs916432364 Chr.14: 77105807-77105807 on Build GRCh38
rs760612809 Chr.14: 77105741-77105741 on Build GRCh38
rs374494028 Chr.14: 77105827-77105827 on Build GRCh38
rs781697967 Chr.14: 77105795-77105795 on Build GRCh38
rs147179239 Chr.14: 77105727-77105727 on Build GRCh38
rs561538254 Chr.14: 77105716-77105716 on Build GRCh38
rs370987526 Chr.14: 77105822-77105822 on Build GRCh38
rs750716051 Chr.14: 77105866-77105867 on Build GRCh38
rs950728889 Chr.14: 77105615-77105615 on Build GRCh38
rs377615689 Chr.14: 77105725-77105725 on Build GRCh38
rs1050725579 Chr.14: 77105934-77105934 on Build GRCh38
rs774615691 Chr.14: 77105793-77105793 on Build GRCh38
rs778540808 Chr.14: 77105764-77105764 on Build GRCh38
rs768228196 Chr.14: 77105710-77105710 on Build GRCh38
rs779238816 Chr.14: 77105702-77105703 on Build GRCh38
rs747901822 Chr.14: 77105816-77105816 on Build GRCh38
rs758677409 Chr.14: 77105779-77105779 on Build GRCh38
rs113899918 Chr.14: 77105891-77105891 on Build GRCh38
rs912211570 Chr.14: 77105944-77105944 on Build GRCh38
rs1057155222 Chr.14: 77105690-77105690 on Build GRCh38
rs375619876 Chr.14: 77105689-77105689 on Build GRCh38
rs749447750 Chr.14: 77105669-77105670 on Build GRCh38
rs990904130 Chr.14: 77105753-77105753 on Build GRCh38
rs771647337 Chr.14: 77105819-77105819 on Build GRCh38
rs375362528 Chr.14: 77105892-77105892 on Build GRCh38
rs757665312 Chr.14: 77105689-77105689 on Build GRCh38
rs759365469 Chr.14: 77105875-77105875 on Build GRCh38
rs961101648 Chr.14: 77105692-77105692 on Build GRCh38
rs767073303 Chr.14: 77105730-77105730 on Build GRCh38
rs776826656 Chr.14: 77105966-77105966 on Build GRCh38
rs754762197 Chr.14: 77105755-77105755 on Build GRCh38
rs776670026 Chr.14: 77105862-77105862 on Build GRCh38
rs879817605 Chr.14: 77105617-77105617 on Build GRCh38
rs775703910 Chr.14: 77105805-77105805 on Build GRCh38

View Full Product Details