Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94045931-94046463 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTUB2
Entrez Gene ID
78990
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 2
Gene Aliases
C14orf137, OTB2, OTU2
UniGene
Hs.278815
Gene Chromosome Location
Chr.14: 94016083-94048933 on Build GRCh38
Transcripts
NM_023112.3 XM_005268055.3 XM_011537148.2

SNP Detail

SNP ID Chromosome Location
rs190214508 Chr.14: 94046070-94046070 on Build GRCh38
rs917494709 Chr.14: 94046429-94046429 on Build GRCh38
rs76277259 Chr.14: 94046124-94046124 on Build GRCh38
rs756324408 Chr.14: 94045954-94045954 on Build GRCh38
rs993642087 Chr.14: 94046051-94046051 on Build GRCh38
rs928911669 Chr.14: 94046382-94046382 on Build GRCh38
rs549624835 Chr.14: 94046185-94046185 on Build GRCh38
rs746051138 Chr.14: 94045950-94045950 on Build GRCh38
rs771674839 Chr.14: 94046106-94046106 on Build GRCh38
rs372804232 Chr.14: 94046188-94046188 on Build GRCh38
rs1006014164 Chr.14: 94046342-94046343 on Build GRCh38
rs1007448794 Chr.14: 94046178-94046178 on Build GRCh38
rs1045681668 Chr.14: 94046362-94046362 on Build GRCh38
rs901198371 Chr.14: 94046246-94046246 on Build GRCh38
rs531472514 Chr.14: 94046395-94046395 on Build GRCh38
rs768172962 Chr.14: 94045986-94045986 on Build GRCh38
rs1054220474 Chr.14: 94046167-94046167 on Build GRCh38
rs998186381 Chr.14: 94046252-94046252 on Build GRCh38
rs562041661 Chr.14: 94046437-94046437 on Build GRCh38
rs951664348 Chr.14: 94046047-94046047 on Build GRCh38
rs929522685 Chr.14: 94046387-94046387 on Build GRCh38
rs773131366 Chr.14: 94045969-94045969 on Build GRCh38
rs958953409 Chr.14: 94046064-94046064 on Build GRCh38
rs915935944 Chr.14: 94046200-94046200 on Build GRCh38
rs768728247 Chr.14: 94045957-94045957 on Build GRCh38
rs576615442 Chr.14: 94046061-94046061 on Build GRCh38
rs1029597469 Chr.14: 94046146-94046146 on Build GRCh38
rs886865076 Chr.14: 94046060-94046060 on Build GRCh38
rs949732253 Chr.14: 94046086-94046086 on Build GRCh38
rs1054467301 Chr.14: 94045987-94045987 on Build GRCh38
rs887936506 Chr.14: 94046434-94046434 on Build GRCh38
rs879768913 Chr.14: 94046322-94046322 on Build GRCh38
rs780162254 Chr.14: 94045955-94045955 on Build GRCh38
rs74589634 Chr.14: 94046076-94046076 on Build GRCh38
rs949035753 Chr.14: 94046018-94046018 on Build GRCh38
rs564408365 Chr.14: 94046343-94046343 on Build GRCh38
rs907410867 Chr.14: 94046045-94046045 on Build GRCh38
rs1026186061 Chr.14: 94046289-94046289 on Build GRCh38
rs186137185 Chr.14: 94045984-94045984 on Build GRCh38
rs182340920 Chr.14: 94046306-94046306 on Build GRCh38
rs957509760 Chr.14: 94046182-94046182 on Build GRCh38
rs142155442 Chr.14: 94046413-94046413 on Build GRCh38
rs551987593 Chr.14: 94046041-94046041 on Build GRCh38
rs991766792 Chr.14: 94046412-94046412 on Build GRCh38
rs937619663 Chr.14: 94046052-94046052 on Build GRCh38
rs888918682 Chr.14: 94046171-94046171 on Build GRCh38
rs924707412 Chr.14: 94045972-94045972 on Build GRCh38
rs45535344 Chr.14: 94046150-94046150 on Build GRCh38
rs990634941 Chr.14: 94046195-94046195 on Build GRCh38
rs571898534 Chr.14: 94046156-94046156 on Build GRCh38
rs3827900 Chr.14: 94046439-94046439 on Build GRCh38
rs8021300 Chr.14: 94046069-94046069 on Build GRCh38
rs915887409 Chr.14: 94045989-94045989 on Build GRCh38
rs749333318 Chr.14: 94045956-94045956 on Build GRCh38
rs1047892989 Chr.14: 94046426-94046426 on Build GRCh38
rs977255945 Chr.14: 94046155-94046155 on Build GRCh38
rs759452280 Chr.14: 94046440-94046440 on Build GRCh38
rs977222882 Chr.14: 94045965-94045965 on Build GRCh38
rs1021084071 Chr.14: 94046305-94046305 on Build GRCh38
rs958839173 Chr.14: 94046392-94046392 on Build GRCh38

View Full Product Details