Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68056051-68056484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUS2
Entrez Gene ID
54920
Gene Name
dihydrouridine synthase 2
Gene Aliases
DUS2L, SMM1, URLC8
UniGene
Hs.534460
Gene Chromosome Location
Chr.16: 68022944-68079323 on Build GRCh38
Transcripts
NM_001271762.1 NM_001271763.1 NM_017803.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6650263 c.311A>G p.E104G Chr.16: 68056366-68056366 on Build GRCh38
1204704 c.362C>T p.S121F Chr.16: 68056417-68056417 on Build GRCh38
703796 c.313A>T p.N105Y Chr.16: 68056368-68056368 on Build GRCh38
5645996 c.315T>G p.N105K Chr.16: 68056370-68056370 on Build GRCh38
6650258 c.316G>T p.D106Y Chr.16: 68056371-68056371 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs939218041 Chr.16: 68056250-68056250 on Build GRCh38
rs1006621275 Chr.16: 68056440-68056440 on Build GRCh38
rs1018887331 Chr.16: 68056449-68056449 on Build GRCh38
rs746065586 Chr.16: 68056421-68056421 on Build GRCh38
rs765474996 Chr.16: 68056371-68056371 on Build GRCh38
rs1020095120 Chr.16: 68056283-68056283 on Build GRCh38
rs556635429 Chr.16: 68056142-68056142 on Build GRCh38
rs913268039 Chr.16: 68056172-68056172 on Build GRCh38
rs759669195 Chr.16: 68056339-68056339 on Build GRCh38
rs77183187 Chr.16: 68056188-68056188 on Build GRCh38
rs986670947 Chr.16: 68056202-68056202 on Build GRCh38
rs536455619 Chr.16: 68056197-68056197 on Build GRCh38
rs201046877 Chr.16: 68056454-68056454 on Build GRCh38
rs371513033 Chr.16: 68056331-68056331 on Build GRCh38
rs763904725 Chr.16: 68056212-68056212 on Build GRCh38
rs925907245 Chr.16: 68056365-68056365 on Build GRCh38
rs774131194 Chr.16: 68056242-68056242 on Build GRCh38
rs540502826 Chr.16: 68056338-68056338 on Build GRCh38
rs755966518 Chr.16: 68056423-68056423 on Build GRCh38
rs150676760 Chr.16: 68056309-68056309 on Build GRCh38
rs761112033 Chr.16: 68056330-68056330 on Build GRCh38
rs957526579 Chr.16: 68056457-68056457 on Build GRCh38
rs773379916 Chr.16: 68056328-68056328 on Build GRCh38
rs369846772 Chr.16: 68056450-68056450 on Build GRCh38
rs144136215 Chr.16: 68056381-68056381 on Build GRCh38
rs988206620 Chr.16: 68056252-68056252 on Build GRCh38
rs990348928 Chr.16: 68056122-68056122 on Build GRCh38
rs371502499 Chr.16: 68056249-68056249 on Build GRCh38
rs779541209 Chr.16: 68056431-68056431 on Build GRCh38
rs750134756 Chr.16: 68056382-68056382 on Build GRCh38
rs953975473 Chr.16: 68056193-68056193 on Build GRCh38
rs928683219 Chr.16: 68056233-68056233 on Build GRCh38
rs9933500 Chr.16: 68056082-68056082 on Build GRCh38
rs993798066 Chr.16: 68056410-68056410 on Build GRCh38
rs1038292119 Chr.16: 68056336-68056336 on Build GRCh38
rs894968393 Chr.16: 68056329-68056329 on Build GRCh38
rs749018294 Chr.16: 68056437-68056437 on Build GRCh38
rs751538505 Chr.16: 68056372-68056372 on Build GRCh38
rs554420886 Chr.16: 68056240-68056240 on Build GRCh38
rs1035023389 Chr.16: 68056222-68056222 on Build GRCh38
rs995105366 Chr.16: 68056070-68056070 on Build GRCh38
rs377294409 Chr.16: 68056444-68056444 on Build GRCh38

View Full Product Details