Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 59697484-59697979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTRH2
Entrez Gene ID
51651
Gene Name
peptidyl-tRNA hydrolase 2
Gene Aliases
BIT1, CFAP37, CGI-147, IMNEPD, PTH, PTH 2, PTH2
UniGene
Hs.12677
Gene Chromosome Location
Chr.17: 59697306-59707598 on Build GRCh38
Transcripts
NM_001015509.2 NM_016077.4 XM_011524887.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5853123 c.461G>A p.G154D Chr.17: 59697518-59697518 on Build GRCh38
5712816 c.43A>G p.S15G Chr.17: 59697936-59697936 on Build GRCh38
5669974 c.357delA p.A120fs*7 Chr.17: 59697622-59697622 on Build GRCh38
5579441 c.127A>G p.S43G Chr.17: 59697852-59697852 on Build GRCh38
5464126 c.203T>G p.I68S Chr.17: 59697776-59697776 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs573168821 Chr.17: 59697930-59697930 on Build GRCh38
rs763966443 Chr.17: 59697764-59697764 on Build GRCh38
rs765174064 Chr.17: 59697891-59697891 on Build GRCh38
rs769066334 Chr.17: 59697702-59697702 on Build GRCh38
rs895769216 Chr.17: 59697630-59697630 on Build GRCh38
rs771032527 Chr.17: 59697536-59697536 on Build GRCh38
rs777064652 Chr.17: 59697851-59697852 on Build GRCh38
rs753776429 Chr.17: 59697884-59697884 on Build GRCh38
rs151008341 Chr.17: 59697761-59697761 on Build GRCh38
rs142925288 Chr.17: 59697811-59697811 on Build GRCh38
rs771731005 Chr.17: 59697844-59697844 on Build GRCh38
rs757015560 Chr.17: 59697772-59697772 on Build GRCh38
rs770212661 Chr.17: 59697828-59697828 on Build GRCh38
rs540235333 Chr.17: 59697936-59697936 on Build GRCh38
rs1044400035 Chr.17: 59697750-59697750 on Build GRCh38
rs747601962 Chr.17: 59697695-59697695 on Build GRCh38
rs11267 Chr.17: 59697612-59697612 on Build GRCh38
rs985248778 Chr.17: 59697678-59697680 on Build GRCh38
rs373761025 Chr.17: 59697892-59697892 on Build GRCh38
rs760336557 Chr.17: 59697657-59697658 on Build GRCh38
rs777157358 Chr.17: 59697719-59697719 on Build GRCh38
rs145242271 Chr.17: 59697510-59697510 on Build GRCh38
rs913374207 Chr.17: 59697845-59697845 on Build GRCh38
rs1037221821 Chr.17: 59697793-59697793 on Build GRCh38
rs748645426 Chr.17: 59697927-59697927 on Build GRCh38
rs773538437 Chr.17: 59697841-59697841 on Build GRCh38
rs777422769 Chr.17: 59697530-59697530 on Build GRCh38
rs990602167 Chr.17: 59697664-59697664 on Build GRCh38
rs199524996 Chr.17: 59697905-59697905 on Build GRCh38
rs768716197 Chr.17: 59697809-59697809 on Build GRCh38
rs778836130 Chr.17: 59697774-59697774 on Build GRCh38
rs758673450 Chr.17: 59697784-59697784 on Build GRCh38
rs751148771 Chr.17: 59697600-59697600 on Build GRCh38
rs758228273 Chr.17: 59697900-59697900 on Build GRCh38
rs730882234 Chr.17: 59697725-59697725 on Build GRCh38
rs755075133 Chr.17: 59697913-59697913 on Build GRCh38
rs1035605110 Chr.17: 59697603-59697603 on Build GRCh38
rs766131601 Chr.17: 59697557-59697557 on Build GRCh38
rs938886094 Chr.17: 59697789-59697789 on Build GRCh38
rs974808666 Chr.17: 59697757-59697757 on Build GRCh38
rs786201017 Chr.17: 59697709-59697710 on Build GRCh38
rs11547273 Chr.17: 59697939-59697939 on Build GRCh38
rs180743750 Chr.17: 59697732-59697732 on Build GRCh38
rs760107286 Chr.17: 59697874-59697874 on Build GRCh38
rs557093317 Chr.17: 59697890-59697890 on Build GRCh38
rs1001232841 Chr.17: 59697583-59697583 on Build GRCh38
rs762188817 Chr.17: 59697595-59697595 on Build GRCh38
rs369388812 Chr.17: 59697643-59697643 on Build GRCh38
rs200433515 Chr.17: 59697689-59697689 on Build GRCh38
rs751750580 Chr.17: 59697909-59697909 on Build GRCh38
rs368765158 Chr.17: 59697931-59697931 on Build GRCh38
rs200466198 Chr.17: 59697604-59697604 on Build GRCh38
rs750740618 Chr.17: 59697775-59697775 on Build GRCh38
rs748679121 Chr.17: 59697822-59697822 on Build GRCh38
rs770091619 Chr.17: 59697728-59697728 on Build GRCh38
rs567829475 Chr.17: 59697527-59697527 on Build GRCh38
rs554318196 Chr.17: 59697613-59697613 on Build GRCh38
rs1017975829 Chr.17: 59697555-59697555 on Build GRCh38
rs775164978 Chr.17: 59697858-59697858 on Build GRCh38
rs749561616 Chr.17: 59697531-59697531 on Build GRCh38
rs535044946 Chr.17: 59697658-59697658 on Build GRCh38
rs765979326 Chr.17: 59697599-59697599 on Build GRCh38
rs774051081 Chr.17: 59697731-59697731 on Build GRCh38
rs566455259 Chr.17: 59697624-59697624 on Build GRCh38
rs975919661 Chr.17: 59697566-59697566 on Build GRCh38
rs952593557 Chr.17: 59697671-59697671 on Build GRCh38
rs76310651 Chr.17: 59697918-59697918 on Build GRCh38
rs776839594 Chr.17: 59697585-59697585 on Build GRCh38
rs905514104 Chr.17: 59697567-59697567 on Build GRCh38
rs35719333 Chr.17: 59697575-59697576 on Build GRCh38
rs761360230 Chr.17: 59697942-59697942 on Build GRCh38
rs748023305 Chr.17: 59697522-59697522 on Build GRCh38
rs377445665 Chr.17: 59697896-59697896 on Build GRCh38
rs754500972 Chr.17: 59697601-59697601 on Build GRCh38
rs769153566 Chr.17: 59697560-59697561 on Build GRCh38
rs746659595 Chr.17: 59697934-59697934 on Build GRCh38
rs145040441 Chr.17: 59697938-59697938 on Build GRCh38
rs994059740 Chr.17: 59697958-59697958 on Build GRCh38
rs376426220 Chr.17: 59697945-59697945 on Build GRCh38
rs780377118 Chr.17: 59697787-59697787 on Build GRCh38
rs985804677 Chr.17: 59697551-59697551 on Build GRCh38
rs747089911 Chr.17: 59697792-59697792 on Build GRCh38
rs538467110 Chr.17: 59697537-59697537 on Build GRCh38
rs933536913 Chr.17: 59697800-59697800 on Build GRCh38
rs752177984 Chr.17: 59697743-59697743 on Build GRCh38
rs375293191 Chr.17: 59697579-59697579 on Build GRCh38
rs765298738 Chr.17: 59697943-59697943 on Build GRCh38
rs759188112 Chr.17: 59697739-59697739 on Build GRCh38
rs749056029 Chr.17: 59697639-59697639 on Build GRCh38
rs147437736 Chr.17: 59697928-59697928 on Build GRCh38
rs776151941 Chr.17: 59697561-59697561 on Build GRCh38
rs762093483 Chr.17: 59697726-59697726 on Build GRCh38
rs752694989 Chr.17: 59697605-59697605 on Build GRCh38
rs772335469 Chr.17: 59697667-59697667 on Build GRCh38
rs763600816 Chr.17: 59697878-59697878 on Build GRCh38
rs753741114 Chr.17: 59697765-59697765 on Build GRCh38
rs771215869 Chr.17: 59697932-59697932 on Build GRCh38
rs779281770 Chr.17: 59697640-59697640 on Build GRCh38
rs1016376790 Chr.17: 59697717-59697717 on Build GRCh38
rs923283708 Chr.17: 59697791-59697791 on Build GRCh38

View Full Product Details