Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48970414-48970986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GYS1
Entrez Gene ID
2997
Gene Name
glycogen synthase 1
Gene Aliases
GSY, GYS
UniGene
Hs.386225
Gene Chromosome Location
Chr.19: 48968125-48993353 on Build GRCh38
Transcripts
NM_001161587.1 NM_002103.4 NR_027763.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3536716 c.1666C>G p.R556G Chr.19: 48970689-48970689 on Build GRCh38
4452213 c.1682A>G p.D561G Chr.19: 48970673-48970673 on Build GRCh38
3536717 c.1599C>T p.G533G Chr.19: 48970974-48970974 on Build GRCh38
3536715 c.1779C>T p.S593S Chr.19: 48970576-48970576 on Build GRCh38
6210206 c.1735C>T p.R579W Chr.19: 48970620-48970620 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372542706 Chr.19: 48970576-48970576 on Build GRCh38
rs2270938 Chr.19: 48970770-48970770 on Build GRCh38
rs934776008 Chr.19: 48970812-48970813 on Build GRCh38
rs375027132 Chr.19: 48970673-48970673 on Build GRCh38
rs759561434 Chr.19: 48970934-48970934 on Build GRCh38
rs769155028 Chr.19: 48970651-48970652 on Build GRCh38
rs758915459 Chr.19: 48970469-48970469 on Build GRCh38
rs775569633 Chr.19: 48970450-48970450 on Build GRCh38
rs768029465 Chr.19: 48970949-48970949 on Build GRCh38
rs747293986 Chr.19: 48970474-48970474 on Build GRCh38
rs377063513 Chr.19: 48970593-48970593 on Build GRCh38
rs747526091 Chr.19: 48970583-48970583 on Build GRCh38
rs781568202 Chr.19: 48970686-48970686 on Build GRCh38
rs561646250 Chr.19: 48970958-48970958 on Build GRCh38
rs762467120 Chr.19: 48970521-48970521 on Build GRCh38
rs753013962 Chr.19: 48970691-48970691 on Build GRCh38
rs772645668 Chr.19: 48970520-48970520 on Build GRCh38
rs770398662 Chr.19: 48970611-48970611 on Build GRCh38
rs572026305 Chr.19: 48970788-48970788 on Build GRCh38
rs757609406 Chr.19: 48970462-48970462 on Build GRCh38
rs749307856 Chr.19: 48970632-48970633 on Build GRCh38
rs575792567 Chr.19: 48970814-48970814 on Build GRCh38
rs540051635 Chr.19: 48970497-48970497 on Build GRCh38
rs750481025 Chr.19: 48970539-48970539 on Build GRCh38
rs751255988 Chr.19: 48970436-48970436 on Build GRCh38
rs370992600 Chr.19: 48970683-48970683 on Build GRCh38
rs773784707 Chr.19: 48970615-48970615 on Build GRCh38
rs764351612 Chr.19: 48970928-48970928 on Build GRCh38
rs754061582 Chr.19: 48970451-48970451 on Build GRCh38
rs867792741 Chr.19: 48970661-48970661 on Build GRCh38
rs993164113 Chr.19: 48970718-48970718 on Build GRCh38
rs952292874 Chr.19: 48970748-48970748 on Build GRCh38
rs368201475 Chr.19: 48970907-48970907 on Build GRCh38
rs766257309 Chr.19: 48970523-48970523 on Build GRCh38
rs545857502 Chr.19: 48970802-48970802 on Build GRCh38
rs781618580 Chr.19: 48970600-48970600 on Build GRCh38
rs760730560 Chr.19: 48970886-48970886 on Build GRCh38
rs968964775 Chr.19: 48970682-48970682 on Build GRCh38
rs767323550 Chr.19: 48970526-48970526 on Build GRCh38
rs763313450 Chr.19: 48970677-48970677 on Build GRCh38
rs914537376 Chr.19: 48970818-48970818 on Build GRCh38
rs775296584 Chr.19: 48970647-48970647 on Build GRCh38
rs144530380 Chr.19: 48970753-48970755 on Build GRCh38
rs754825881 Chr.19: 48970440-48970440 on Build GRCh38
rs745662040 Chr.19: 48970679-48970679 on Build GRCh38
rs767898665 Chr.19: 48970888-48970899 on Build GRCh38
rs1012232014 Chr.19: 48970817-48970817 on Build GRCh38
rs758577682 Chr.19: 48970552-48970552 on Build GRCh38
rs752499733 Chr.19: 48970528-48970528 on Build GRCh38
rs771907219 Chr.19: 48970638-48970638 on Build GRCh38
rs778151650 Chr.19: 48970713-48970713 on Build GRCh38
rs1006130978 Chr.19: 48970450-48970450 on Build GRCh38
rs768545442 Chr.19: 48970892-48970892 on Build GRCh38
rs574419440 Chr.19: 48970746-48970746 on Build GRCh38
rs143040823 Chr.19: 48970547-48970547 on Build GRCh38
rs947377748 Chr.19: 48970831-48970831 on Build GRCh38
rs1044348744 Chr.19: 48970866-48970866 on Build GRCh38
rs771996063 Chr.19: 48970751-48970751 on Build GRCh38
rs888431613 Chr.19: 48970810-48970810 on Build GRCh38
rs146698792 Chr.19: 48970606-48970606 on Build GRCh38
rs745944767 Chr.19: 48970467-48970467 on Build GRCh38
rs192877287 Chr.19: 48970513-48970513 on Build GRCh38
rs764183447 Chr.19: 48970444-48970444 on Build GRCh38
rs760568725 Chr.19: 48970651-48970651 on Build GRCh38
rs374209527 Chr.19: 48970524-48970524 on Build GRCh38
rs1000684948 Chr.19: 48970439-48970439 on Build GRCh38
rs765569128 Chr.19: 48970533-48970533 on Build GRCh38
rs1032583213 Chr.19: 48970542-48970542 on Build GRCh38
rs748471233 Chr.19: 48970601-48970601 on Build GRCh38
rs1003122096 Chr.19: 48970908-48970908 on Build GRCh38
rs756503222 Chr.19: 48970706-48970706 on Build GRCh38
rs770548301 Chr.19: 48970735-48970735 on Build GRCh38
rs988809717 Chr.19: 48970813-48970813 on Build GRCh38
rs566032681 Chr.19: 48970478-48970478 on Build GRCh38
rs753171319 Chr.19: 48970952-48970952 on Build GRCh38
rs112732511 Chr.19: 48970785-48970785 on Build GRCh38
rs759512073 Chr.19: 48970619-48970619 on Build GRCh38
rs148812621 Chr.19: 48970743-48970743 on Build GRCh38
rs370798288 Chr.19: 48970504-48970504 on Build GRCh38
rs1002459436 Chr.19: 48970639-48970639 on Build GRCh38
rs779130126 Chr.19: 48970463-48970463 on Build GRCh38
rs563959255 Chr.19: 48970803-48970803 on Build GRCh38
rs751603575 Chr.19: 48970929-48970929 on Build GRCh38
rs766465529 Chr.19: 48970678-48970678 on Build GRCh38
rs775386568 Chr.19: 48970787-48970787 on Build GRCh38
rs774833368 Chr.19: 48970683-48970685 on Build GRCh38
rs748918644 Chr.19: 48970719-48970719 on Build GRCh38
rs765512016 Chr.19: 48970652-48970652 on Build GRCh38
rs1000551471 Chr.19: 48970646-48970647 on Build GRCh38
rs547918969 Chr.19: 48970471-48970471 on Build GRCh38
rs370779569 Chr.19: 48970927-48970927 on Build GRCh38
rs543315812 Chr.19: 48970846-48970846 on Build GRCh38
rs573017660 Chr.19: 48970805-48970805 on Build GRCh38
rs756664869 Chr.19: 48970957-48970957 on Build GRCh38
rs748757141 Chr.19: 48970608-48970608 on Build GRCh38
rs757485951 Chr.19: 48970530-48970530 on Build GRCh38

View Full Product Details