Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 83956025-83956620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf64
Entrez Gene ID
84267
Gene Name
chromosome 9 open reading frame 64
Gene Aliases
-
UniGene
Hs.208914
Gene Chromosome Location
Chr.9: 83938311-83956986 on Build GRCh38
Transcripts
NM_001317997.1 NM_032307.4 XM_005252271.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110635 c.76G>A p.G26R Chr.9: 83956425-83956425 on Build GRCh38
3848982 c.330C>T p.L110L Chr.9: 83956171-83956171 on Build GRCh38
1314979 c.306G>T p.L102L Chr.9: 83956195-83956195 on Build GRCh38
6221715 c.53G>T p.S18I Chr.9: 83956448-83956448 on Build GRCh38
3908548 c.331G>A p.D111N Chr.9: 83956170-83956170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371097502 Chr.9: 83956420-83956420 on Build GRCh38
rs183493508 Chr.9: 83956211-83956211 on Build GRCh38
rs761051038 Chr.9: 83956223-83956223 on Build GRCh38
rs777751135 Chr.9: 83956265-83956265 on Build GRCh38
rs1007846417 Chr.9: 83956251-83956251 on Build GRCh38
rs367948518 Chr.9: 83956426-83956426 on Build GRCh38
rs375775610 Chr.9: 83956523-83956523 on Build GRCh38
rs908460119 Chr.9: 83956431-83956431 on Build GRCh38
rs748013764 Chr.9: 83956143-83956143 on Build GRCh38
rs201179129 Chr.9: 83956451-83956451 on Build GRCh38
rs1056635951 Chr.9: 83956536-83956536 on Build GRCh38
rs530116332 Chr.9: 83956463-83956463 on Build GRCh38
rs765623164 Chr.9: 83956241-83956241 on Build GRCh38
rs577282500 Chr.9: 83956146-83956146 on Build GRCh38
rs771018653 Chr.9: 83956482-83956482 on Build GRCh38
rs767358358 Chr.9: 83956308-83956308 on Build GRCh38
rs773221993 Chr.9: 83956343-83956343 on Build GRCh38
rs763018033 Chr.9: 83956307-83956307 on Build GRCh38
rs938653740 Chr.9: 83956068-83956068 on Build GRCh38
rs376334570 Chr.9: 83956213-83956213 on Build GRCh38
rs370900126 Chr.9: 83956334-83956334 on Build GRCh38
rs969264555 Chr.9: 83956311-83956311 on Build GRCh38
rs770627224 Chr.9: 83956172-83956172 on Build GRCh38
rs1056649474 Chr.9: 83956163-83956163 on Build GRCh38
rs748863152 Chr.9: 83956270-83956270 on Build GRCh38
rs751733671 Chr.9: 83956534-83956534 on Build GRCh38
rs757924826 Chr.9: 83956446-83956446 on Build GRCh38
rs970434016 Chr.9: 83956542-83956542 on Build GRCh38
rs754380740 Chr.9: 83956321-83956321 on Build GRCh38
rs755931578 Chr.9: 83956327-83956327 on Build GRCh38
rs541714184 Chr.9: 83956281-83956281 on Build GRCh38
rs992853800 Chr.9: 83956071-83956071 on Build GRCh38
rs747402482 Chr.9: 83956358-83956358 on Build GRCh38
rs758660665 Chr.9: 83956492-83956492 on Build GRCh38
rs777328770 Chr.9: 83956364-83956364 on Build GRCh38
rs766575120 Chr.9: 83956410-83956410 on Build GRCh38
rs755946493 Chr.9: 83956258-83956258 on Build GRCh38
rs751204627 Chr.9: 83956529-83956529 on Build GRCh38
rs761973673 Chr.9: 83956230-83956230 on Build GRCh38
rs200452634 Chr.9: 83956376-83956376 on Build GRCh38
rs201517382 Chr.9: 83956432-83956432 on Build GRCh38
rs930878306 Chr.9: 83956518-83956518 on Build GRCh38
rs1049205396 Chr.9: 83956094-83956094 on Build GRCh38
rs983508162 Chr.9: 83956588-83956588 on Build GRCh38
rs773592058 Chr.9: 83956305-83956305 on Build GRCh38
rs776356197 Chr.9: 83956507-83956507 on Build GRCh38
rs375718696 Chr.9: 83956338-83956338 on Build GRCh38
rs759759170 Chr.9: 83956504-83956504 on Build GRCh38
rs899517382 Chr.9: 83956264-83956264 on Build GRCh38
rs896242357 Chr.9: 83956173-83956173 on Build GRCh38
rs764242518 Chr.9: 83956224-83956224 on Build GRCh38
rs566767297 Chr.9: 83956213-83956214 on Build GRCh38
rs139237715 Chr.9: 83956545-83956545 on Build GRCh38
rs765386439 Chr.9: 83956304-83956304 on Build GRCh38
rs375868534 Chr.9: 83956139-83956139 on Build GRCh38
rs769035210 Chr.9: 83956361-83956361 on Build GRCh38
rs371835249 Chr.9: 83956438-83956438 on Build GRCh38
rs754612568 Chr.9: 83956531-83956531 on Build GRCh38
rs759932091 Chr.9: 83956197-83956197 on Build GRCh38
rs771774324 Chr.9: 83956214-83956216 on Build GRCh38
rs201215719 Chr.9: 83956275-83956275 on Build GRCh38
rs374444713 Chr.9: 83956289-83956289 on Build GRCh38
rs903562469 Chr.9: 83956569-83956569 on Build GRCh38
rs749750517 Chr.9: 83956549-83956549 on Build GRCh38
rs775654653 Chr.9: 83956294-83956294 on Build GRCh38
rs756778571 Chr.9: 83956336-83956336 on Build GRCh38
rs781656702 Chr.9: 83956455-83956455 on Build GRCh38
rs199825781 Chr.9: 83956207-83956207 on Build GRCh38
rs752982297 Chr.9: 83956471-83956471 on Build GRCh38
rs920701994 Chr.9: 83956372-83956372 on Build GRCh38
rs763074430 Chr.9: 83956391-83956391 on Build GRCh38
rs1031912571 Chr.9: 83956329-83956329 on Build GRCh38
rs774600765 Chr.9: 83956411-83956411 on Build GRCh38
rs760224547 Chr.9: 83956484-83956484 on Build GRCh38
rs781068797 Chr.9: 83956584-83956584 on Build GRCh38
rs763825858 Chr.9: 83956421-83956421 on Build GRCh38
rs567193928 Chr.9: 83956589-83956589 on Build GRCh38
rs753914428 Chr.9: 83956228-83956228 on Build GRCh38
rs79211911 Chr.9: 83956442-83956442 on Build GRCh38
rs769104960 Chr.9: 83956300-83956300 on Build GRCh38
rs748676572 Chr.9: 83956461-83956461 on Build GRCh38
rs898282719 Chr.9: 83956170-83956170 on Build GRCh38
rs908345925 Chr.9: 83956594-83956594 on Build GRCh38
rs763045476 Chr.9: 83956059-83956059 on Build GRCh38
rs777080747 Chr.9: 83956153-83956153 on Build GRCh38
rs571879459 Chr.9: 83956385-83956385 on Build GRCh38
rs748951208 Chr.9: 83956166-83956166 on Build GRCh38
rs201966216 Chr.9: 83956332-83956332 on Build GRCh38
rs773986048 Chr.9: 83956178-83956178 on Build GRCh38
rs749365822 Chr.9: 83956478-83956478 on Build GRCh38
rs777275368 Chr.9: 83956301-83956301 on Build GRCh38
rs187991757 Chr.9: 83956505-83956505 on Build GRCh38
rs1023239910 Chr.9: 83956540-83956540 on Build GRCh38
rs765227800 Chr.9: 83956521-83956521 on Build GRCh38
rs948814553 Chr.9: 83956145-83956145 on Build GRCh38
rs560017514 Chr.9: 83956333-83956333 on Build GRCh38
rs773367311 Chr.9: 83956387-83956387 on Build GRCh38
rs778004244 Chr.9: 83956516-83956516 on Build GRCh38
rs574541651 Chr.9: 83956243-83956243 on Build GRCh38
rs558982640 Chr.9: 83956051-83956051 on Build GRCh38
rs779614748 Chr.9: 83956448-83956448 on Build GRCh38
rs1019273749 Chr.9: 83956261-83956261 on Build GRCh38
rs765864766 Chr.9: 83956527-83956527 on Build GRCh38
rs200023711 Chr.9: 83956244-83956244 on Build GRCh38
rs766293578 Chr.9: 83956547-83956548 on Build GRCh38
rs967946479 Chr.9: 83956229-83956229 on Build GRCh38
rs368545737 Chr.9: 83956324-83956324 on Build GRCh38
rs751460571 Chr.9: 83956116-83956116 on Build GRCh38
rs779745846 Chr.9: 83956349-83956349 on Build GRCh38
rs770394274 Chr.9: 83956271-83956271 on Build GRCh38
rs371799329 Chr.9: 83956522-83956522 on Build GRCh38
rs752684023 Chr.9: 83956250-83956250 on Build GRCh38
rs112891509 Chr.9: 83956078-83956078 on Build GRCh38
rs375670830 Chr.9: 83956553-83956553 on Build GRCh38
rs778583165 Chr.9: 83956285-83956285 on Build GRCh38
rs745469947 Chr.9: 83956287-83956287 on Build GRCh38
rs1000777385 Chr.9: 83956575-83956575 on Build GRCh38
rs41282419 Chr.9: 83956539-83956539 on Build GRCh38
rs745543273 Chr.9: 83956342-83956342 on Build GRCh38
rs761587641 Chr.9: 83956515-83956515 on Build GRCh38
rs773762916 Chr.9: 83956476-83956476 on Build GRCh38
rs367562680 Chr.9: 83956452-83956452 on Build GRCh38
rs754818909 Chr.9: 83956119-83956119 on Build GRCh38

View Full Product Details