Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133349384-133349983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL7A
Entrez Gene ID
6130
Gene Name
ribosomal protein L7a
Gene Aliases
L7A, SURF3, TRUP
UniGene
Hs.499839
Gene Chromosome Location
Chr.9: 133348214-133351425 on Build GRCh38
Transcripts
NM_000972.2 NR_002447.1

Gene Symbol
SNORD24
Entrez Gene ID
26820
Gene Name
small nucleolar RNA, C/D box 24
Gene Aliases
RNU24, U24
UniGene
Gene Chromosome Location
Chr.9: 133349396-133349470 on Build GRCh38
Transcripts
NM_000972.2 NR_002447.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996414 c.208C>A p.L70I Chr.9: 133349634-133349634 on Build GRCh38
7327497 c.130G>T p.D44Y Chr.9: 133349556-133349556 on Build GRCh38
3929892 c.203C>G p.A68G Chr.9: 133349629-133349629 on Build GRCh38
5662579 c.354A>T p.T118T Chr.9: 133349699-133349699 on Build GRCh38
5571631 c.245T>A p.V82E Chr.9: 133349590-133349590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782719130 Chr.9: 133349415-133349415 on Build GRCh38
rs991697881 Chr.9: 133349461-133349461 on Build GRCh38
rs781903336 Chr.9: 133349489-133349489 on Build GRCh38
rs782447963 Chr.9: 133349716-133349716 on Build GRCh38
rs782253602 Chr.9: 133349744-133349744 on Build GRCh38
rs782670277 Chr.9: 133349509-133349509 on Build GRCh38
rs199816702 Chr.9: 133349957-133349957 on Build GRCh38
rs782312134 Chr.9: 133349638-133349638 on Build GRCh38
rs941959380 Chr.9: 133349462-133349462 on Build GRCh38
rs781920200 Chr.9: 133349530-133349530 on Build GRCh38
rs782518136 Chr.9: 133349722-133349722 on Build GRCh38
rs1046851787 Chr.9: 133349861-133349861 on Build GRCh38
rs368462361 Chr.9: 133349703-133349703 on Build GRCh38
rs782163866 Chr.9: 133349532-133349532 on Build GRCh38
rs782483182 Chr.9: 133349426-133349427 on Build GRCh38
rs782032987 Chr.9: 133349866-133349866 on Build GRCh38
rs782036352 Chr.9: 133349416-133349416 on Build GRCh38
rs781834095 Chr.9: 133349576-133349576 on Build GRCh38
rs878888328 Chr.9: 133349761-133349761 on Build GRCh38
rs781983846 Chr.9: 133349681-133349681 on Build GRCh38
rs781949484 Chr.9: 133349477-133349477 on Build GRCh38
rs772306147 Chr.9: 133349433-133349433 on Build GRCh38
rs199584544 Chr.9: 133349628-133349628 on Build GRCh38
rs782361515 Chr.9: 133349412-133349412 on Build GRCh38
rs367994158 Chr.9: 133349463-133349463 on Build GRCh38
rs782430796 Chr.9: 133349434-133349434 on Build GRCh38
rs587606803 Chr.9: 133349508-133349508 on Build GRCh38
rs956733027 Chr.9: 133349812-133349812 on Build GRCh38
rs782618951 Chr.9: 133349452-133349452 on Build GRCh38
rs991090227 Chr.9: 133349669-133349669 on Build GRCh38
rs782689880 Chr.9: 133349865-133349865 on Build GRCh38
rs368935283 Chr.9: 133349908-133349908 on Build GRCh38
rs782322028 Chr.9: 133349529-133349529 on Build GRCh38
rs1012257176 Chr.9: 133349579-133349579 on Build GRCh38
rs782786546 Chr.9: 133349723-133349723 on Build GRCh38
rs782665736 Chr.9: 133349606-133349606 on Build GRCh38
rs958569236 Chr.9: 133349459-133349459 on Build GRCh38
rs782321642 Chr.9: 133349932-133349932 on Build GRCh38
rs116631717 Chr.9: 133349495-133349495 on Build GRCh38
rs782258169 Chr.9: 133349486-133349486 on Build GRCh38
rs113336790 Chr.9: 133349899-133349899 on Build GRCh38
rs781964119 Chr.9: 133349774-133349774 on Build GRCh38
rs782097382 Chr.9: 133349541-133349541 on Build GRCh38
rs782784920 Chr.9: 133349663-133349663 on Build GRCh38
rs782593406 Chr.9: 133349600-133349600 on Build GRCh38
rs781787675 Chr.9: 133349421-133349424 on Build GRCh38
rs782515361 Chr.9: 133349447-133349447 on Build GRCh38
rs967952299 Chr.9: 133349611-133349611 on Build GRCh38
rs372369045 Chr.9: 133349725-133349725 on Build GRCh38
rs117469434 Chr.9: 133349486-133349486 on Build GRCh38
rs1021467607 Chr.9: 133349909-133349909 on Build GRCh38
rs958906413 Chr.9: 133349765-133349766 on Build GRCh38
rs782295784 Chr.9: 133349946-133349946 on Build GRCh38
rs782102788 Chr.9: 133349828-133349828 on Build GRCh38
rs782687155 Chr.9: 133349438-133349438 on Build GRCh38
rs782096662 Chr.9: 133349691-133349691 on Build GRCh38
rs782666407 Chr.9: 133349887-133349887 on Build GRCh38
rs587693152 Chr.9: 133349802-133349802 on Build GRCh38
rs782580335 Chr.9: 133349730-133349730 on Build GRCh38
rs782597297 Chr.9: 133349427-133349428 on Build GRCh38
rs968510254 Chr.9: 133349793-133349793 on Build GRCh38
rs782465096 Chr.9: 133349499-133349499 on Build GRCh38
rs891199656 Chr.9: 133349780-133349780 on Build GRCh38
rs931447576 Chr.9: 133349854-133349854 on Build GRCh38
rs587754227 Chr.9: 133349750-133349750 on Build GRCh38
rs782303694 Chr.9: 133349457-133349457 on Build GRCh38
rs11549051 Chr.9: 133349943-133349943 on Build GRCh38
rs781987872 Chr.9: 133349538-133349538 on Build GRCh38
rs891248278 Chr.9: 133349492-133349492 on Build GRCh38
rs782621407 Chr.9: 133349922-133349922 on Build GRCh38
rs782374019 Chr.9: 133349522-133349522 on Build GRCh38
rs963369936 Chr.9: 133349953-133349953 on Build GRCh38
rs1018313351 Chr.9: 133349536-133349536 on Build GRCh38
rs1035620544 Chr.9: 133349764-133349764 on Build GRCh38
rs915574810 Chr.9: 133349692-133349692 on Build GRCh38
rs979499291 Chr.9: 133349801-133349801 on Build GRCh38
rs587766503 Chr.9: 133349619-133349619 on Build GRCh38
rs781883413 Chr.9: 133349719-133349719 on Build GRCh38
rs1021762007 Chr.9: 133349605-133349605 on Build GRCh38
rs114365767 Chr.9: 133349745-133349745 on Build GRCh38
rs782417715 Chr.9: 133349950-133349950 on Build GRCh38
rs782148443 Chr.9: 133349659-133349659 on Build GRCh38
rs587683734 Chr.9: 133349893-133349893 on Build GRCh38
rs782802831 Chr.9: 133349718-133349718 on Build GRCh38
rs35152611 Chr.9: 133349795-133349796 on Build GRCh38
rs782592452 Chr.9: 133349500-133349500 on Build GRCh38
rs782479235 Chr.9: 133349426-133349426 on Build GRCh38
rs782501709 Chr.9: 133349920-133349920 on Build GRCh38
rs587729202 Chr.9: 133349830-133349830 on Build GRCh38
rs372984922 Chr.9: 133349952-133349952 on Build GRCh38
rs782261441 Chr.9: 133349442-133349442 on Build GRCh38
rs782350563 Chr.9: 133349474-133349474 on Build GRCh38
rs1022293807 Chr.9: 133349852-133349852 on Build GRCh38
rs782249615 Chr.9: 133349610-133349610 on Build GRCh38
rs782756159 Chr.9: 133349498-133349498 on Build GRCh38
rs782526834 Chr.9: 133349570-133349570 on Build GRCh38
rs201422311 Chr.9: 133349637-133349637 on Build GRCh38
rs782041231 Chr.9: 133349652-133349652 on Build GRCh38
rs782458861 Chr.9: 133349577-133349577 on Build GRCh38
rs968539203 Chr.9: 133349912-133349912 on Build GRCh38
rs782202685 Chr.9: 133349528-133349528 on Build GRCh38
rs782264683 Chr.9: 133349915-133349915 on Build GRCh38
rs1051367757 Chr.9: 133349526-133349526 on Build GRCh38
rs782721794 Chr.9: 133349698-133349698 on Build GRCh38
rs782282159 Chr.9: 133349900-133349901 on Build GRCh38
rs142552750 Chr.9: 133349662-133349662 on Build GRCh38
rs11549052 Chr.9: 133349935-133349935 on Build GRCh38
rs148435243 Chr.9: 133349647-133349647 on Build GRCh38
rs782497750 Chr.9: 133349504-133349515 on Build GRCh38
rs370799672 Chr.9: 133349521-133349521 on Build GRCh38
rs782057652 Chr.9: 133349561-133349561 on Build GRCh38
rs1051627872 Chr.9: 133349759-133349759 on Build GRCh38
rs200424390 Chr.9: 133349904-133349904 on Build GRCh38
rs587620997 Chr.9: 133349874-133349874 on Build GRCh38
rs998959778 Chr.9: 133349879-133349879 on Build GRCh38
rs587716326 Chr.9: 133349455-133349455 on Build GRCh38
rs201387355 Chr.9: 133349469-133349469 on Build GRCh38
rs782764194 Chr.9: 133349423-133349423 on Build GRCh38
rs987169815 Chr.9: 133349822-133349822 on Build GRCh38
rs782074524 Chr.9: 133349414-133349414 on Build GRCh38
rs1012834150 Chr.9: 133349766-133349766 on Build GRCh38
rs959805519 Chr.9: 133349664-133349664 on Build GRCh38
rs190442630 Chr.9: 133349537-133349537 on Build GRCh38
rs782563156 Chr.9: 133349939-133349939 on Build GRCh38
rs782417066 Chr.9: 133349602-133349602 on Build GRCh38
rs781938514 Chr.9: 133349762-133349762 on Build GRCh38
rs782344757 Chr.9: 133349849-133349849 on Build GRCh38
rs782211217 Chr.9: 133349923-133349923 on Build GRCh38
rs782406454 Chr.9: 133349460-133349460 on Build GRCh38
rs899839472 Chr.9: 133349829-133349829 on Build GRCh38
rs781838964 Chr.9: 133349887-133349887 on Build GRCh38
rs782067009 Chr.9: 133349485-133349485 on Build GRCh38
rs782452509 Chr.9: 133349728-133349728 on Build GRCh38
rs1040307618 Chr.9: 133349634-133349634 on Build GRCh38
rs369737214 Chr.9: 133349888-133349888 on Build GRCh38
rs782390032 Chr.9: 133349421-133349421 on Build GRCh38
rs782789907 Chr.9: 133349881-133349881 on Build GRCh38
rs782622493 Chr.9: 133349724-133349724 on Build GRCh38
rs782425715 Chr.9: 133349740-133349740 on Build GRCh38
rs185277497 Chr.9: 133349472-133349472 on Build GRCh38
rs781894254 Chr.9: 133349564-133349564 on Build GRCh38
rs372789873 Chr.9: 133349560-133349560 on Build GRCh38
rs782364309 Chr.9: 133349620-133349620 on Build GRCh38
rs998604604 Chr.9: 133349928-133349928 on Build GRCh38
rs782176287 Chr.9: 133349601-133349601 on Build GRCh38
rs1011791799 Chr.9: 133349843-133349843 on Build GRCh38
rs782060157 Chr.9: 133349862-133349862 on Build GRCh38
rs182515784 Chr.9: 133349782-133349782 on Build GRCh38

View Full Product Details