Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 71121384-71121955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED12
Entrez Gene ID
9968
Gene Name
mediator complex subunit 12
Gene Aliases
ARC240, CAGH45, FGS1, HOPA, MED12S, OHDOX, OKS, OPA1, TNRC11, TRAP230
UniGene
Hs.409226
Gene Chromosome Location
Chr.X: 71118556-71142454 on Build GRCh38
Transcripts
NM_005120.2

SNP Detail

SNP ID Chromosome Location
rs762621629 Chr.X: 71121474-71121474 on Build GRCh38
rs781192327 Chr.X: 71121673-71121673 on Build GRCh38
rs762117732 Chr.X: 71121536-71121536 on Build GRCh38
rs988247503 Chr.X: 71121876-71121876 on Build GRCh38
rs868020528 Chr.X: 71121702-71121702 on Build GRCh38
rs35454896 Chr.X: 71121577-71121578 on Build GRCh38
rs769757456 Chr.X: 71121696-71121696 on Build GRCh38
rs775499343 Chr.X: 71121713-71121713 on Build GRCh38
rs761083268 Chr.X: 71121856-71121856 on Build GRCh38
rs764107388 Chr.X: 71121743-71121743 on Build GRCh38
rs370616416 Chr.X: 71121681-71121681 on Build GRCh38
rs957824537 Chr.X: 71121642-71121642 on Build GRCh38
rs184426039 Chr.X: 71121624-71121624 on Build GRCh38
rs763867883 Chr.X: 71121781-71121781 on Build GRCh38
rs191831783 Chr.X: 71121519-71121519 on Build GRCh38
rs889516860 Chr.X: 71121908-71121908 on Build GRCh38
rs757929935 Chr.X: 71121761-71121761 on Build GRCh38
rs975607823 Chr.X: 71121837-71121837 on Build GRCh38
rs768889545 Chr.X: 71121730-71121730 on Build GRCh38
rs759516228 Chr.X: 71121659-71121659 on Build GRCh38
rs746765381 Chr.X: 71121472-71121472 on Build GRCh38
rs777541463 Chr.X: 71121650-71121650 on Build GRCh38
rs763218462 Chr.X: 71121531-71121531 on Build GRCh38
rs771841092 Chr.X: 71121468-71121468 on Build GRCh38
rs749348331 Chr.X: 71121726-71121726 on Build GRCh38
rs751970722 Chr.X: 71121534-71121534 on Build GRCh38
rs776803467 Chr.X: 71121791-71121791 on Build GRCh38
rs761753527 Chr.X: 71121478-71121479 on Build GRCh38
rs984165957 Chr.X: 71121606-71121606 on Build GRCh38
rs200510424 Chr.X: 71121834-71121834 on Build GRCh38
rs764716429 Chr.X: 71121592-71121592 on Build GRCh38
rs780907010 Chr.X: 71121829-71121829 on Build GRCh38
rs773615925 Chr.X: 71121746-71121746 on Build GRCh38
rs765787995 Chr.X: 71121487-71121487 on Build GRCh38
rs777774078 Chr.X: 71121471-71121471 on Build GRCh38
rs1036703145 Chr.X: 71121842-71121842 on Build GRCh38
rs751750422 Chr.X: 71121888-71121888 on Build GRCh38
rs773484619 Chr.X: 71121853-71121853 on Build GRCh38
rs377403264 Chr.X: 71121649-71121649 on Build GRCh38
rs767398040 Chr.X: 71121697-71121697 on Build GRCh38
rs746820232 Chr.X: 71121651-71121651 on Build GRCh38
rs775793223 Chr.X: 71121521-71121521 on Build GRCh38
rs1020663623 Chr.X: 71121660-71121660 on Build GRCh38
rs776381572 Chr.X: 71121516-71121516 on Build GRCh38
rs751188840 Chr.X: 71121782-71121782 on Build GRCh38
rs990639258 Chr.X: 71121762-71121762 on Build GRCh38
rs766874065 Chr.X: 71121753-71121753 on Build GRCh38
rs770933774 Chr.X: 71121479-71121480 on Build GRCh38
rs762172777 Chr.X: 71121740-71121740 on Build GRCh38
rs752514663 Chr.X: 71121843-71121843 on Build GRCh38
rs946755767 Chr.X: 71121913-71121913 on Build GRCh38
rs1043689054 Chr.X: 71121932-71121932 on Build GRCh38
rs775520886 Chr.X: 71121526-71121526 on Build GRCh38
rs761157306 Chr.X: 71121748-71121748 on Build GRCh38
rs764270048 Chr.X: 71121533-71121533 on Build GRCh38
rs913944670 Chr.X: 71121886-71121886 on Build GRCh38
rs757086557 Chr.X: 71121783-71121783 on Build GRCh38
rs747748109 Chr.X: 71121450-71121450 on Build GRCh38
rs909813295 Chr.X: 71121832-71121832 on Build GRCh38
rs752300879 Chr.X: 71121754-71121754 on Build GRCh38
rs774518546 Chr.X: 71121737-71121737 on Build GRCh38
rs750832089 Chr.X: 71121564-71121564 on Build GRCh38
rs1013999865 Chr.X: 71121482-71121482 on Build GRCh38
rs745671946 Chr.X: 71121830-71121830 on Build GRCh38
rs368175213 Chr.X: 71121845-71121845 on Build GRCh38
rs774257494 Chr.X: 71121466-71121467 on Build GRCh38
rs374744294 Chr.X: 71121549-71121549 on Build GRCh38
rs749399594 Chr.X: 71121835-71121835 on Build GRCh38
rs189170979 Chr.X: 71121879-71121879 on Build GRCh38
rs754533515 Chr.X: 71121587-71121587 on Build GRCh38

View Full Product Details