Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 36494494-36495047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK38
Entrez Gene ID
11329
Gene Name
serine/threonine kinase 38
Gene Aliases
NDR, NDR1
UniGene
Hs.409578
Gene Chromosome Location
Chr.6: 36493882-36547516 on Build GRCh38
Transcripts
NM_001305102.1 NM_007271.3 XM_006714988.3 XM_006714989.3 XM_017010226.1

SNP Detail

SNP ID Chromosome Location
rs116829549 Chr.6: 36494687-36494687 on Build GRCh38
rs563450148 Chr.6: 36494593-36494593 on Build GRCh38
rs949379555 Chr.6: 36494979-36494979 on Build GRCh38
rs765402887 Chr.6: 36494775-36494775 on Build GRCh38
rs1037310090 Chr.6: 36494657-36494657 on Build GRCh38
rs980165672 Chr.6: 36494568-36494568 on Build GRCh38
rs1043602390 Chr.6: 36494966-36494966 on Build GRCh38
rs867703460 Chr.6: 36494524-36494524 on Build GRCh38
rs80033474 Chr.6: 36494541-36494541 on Build GRCh38
rs1029250767 Chr.6: 36494895-36494895 on Build GRCh38
rs1039454358 Chr.6: 36494733-36494733 on Build GRCh38
rs546300326 Chr.6: 36494803-36494803 on Build GRCh38
rs917478621 Chr.6: 36494551-36494552 on Build GRCh38
rs996806757 Chr.6: 36494871-36494871 on Build GRCh38
rs931181575 Chr.6: 36494969-36494969 on Build GRCh38
rs986509139 Chr.6: 36494600-36494600 on Build GRCh38
rs911024170 Chr.6: 36494958-36494958 on Build GRCh38
rs1054930125 Chr.6: 36494973-36494973 on Build GRCh38
rs533250058 Chr.6: 36494953-36494953 on Build GRCh38
rs548832251 Chr.6: 36494890-36494890 on Build GRCh38
rs144312505 Chr.6: 36494560-36494560 on Build GRCh38
rs918436011 Chr.6: 36494630-36494630 on Build GRCh38
rs79817222 Chr.6: 36494936-36494936 on Build GRCh38
rs148729338 Chr.6: 36494823-36494823 on Build GRCh38
rs956238398 Chr.6: 36494829-36494829 on Build GRCh38
rs767360632 Chr.6: 36494616-36494616 on Build GRCh38
rs751209765 Chr.6: 36495028-36495028 on Build GRCh38
rs999697631 Chr.6: 36494834-36494834 on Build GRCh38
rs980338674 Chr.6: 36494855-36494855 on Build GRCh38
rs935991408 Chr.6: 36494985-36494985 on Build GRCh38
rs570290315 Chr.6: 36494933-36494933 on Build GRCh38
rs993109921 Chr.6: 36494542-36494542 on Build GRCh38
rs891871914 Chr.6: 36495004-36495004 on Build GRCh38
rs987032250 Chr.6: 36494939-36494939 on Build GRCh38
rs895130009 Chr.6: 36494711-36494711 on Build GRCh38
rs939859318 Chr.6: 36494613-36494613 on Build GRCh38
rs919834412 Chr.6: 36494671-36494671 on Build GRCh38
rs201810594 Chr.6: 36494640-36494640 on Build GRCh38
rs1014304475 Chr.6: 36495026-36495026 on Build GRCh38
rs35552191 Chr.6: 36494622-36494623 on Build GRCh38
rs760909592 Chr.6: 36494582-36494582 on Build GRCh38
rs145235541 Chr.6: 36494680-36494680 on Build GRCh38
rs559243524 Chr.6: 36494977-36494977 on Build GRCh38
rs941091745 Chr.6: 36494597-36494597 on Build GRCh38
rs1032948973 Chr.6: 36494850-36494850 on Build GRCh38
rs909566192 Chr.6: 36494883-36494883 on Build GRCh38
rs1013544977 Chr.6: 36494714-36494714 on Build GRCh38
rs986496325 Chr.6: 36494919-36494919 on Build GRCh38
rs962342933 Chr.6: 36494898-36494898 on Build GRCh38
rs764543103 Chr.6: 36494624-36494624 on Build GRCh38
rs912309982 Chr.6: 36494924-36494924 on Build GRCh38
rs1046420556 Chr.6: 36494992-36494992 on Build GRCh38
rs970721057 Chr.6: 36494846-36494846 on Build GRCh38
rs1053157309 Chr.6: 36494994-36494994 on Build GRCh38
rs953658946 Chr.6: 36494904-36494904 on Build GRCh38
rs912320034 Chr.6: 36494602-36494602 on Build GRCh38
rs1031689865 Chr.6: 36494786-36494786 on Build GRCh38
rs546679390 Chr.6: 36494961-36494961 on Build GRCh38
rs953710642 Chr.6: 36494598-36494598 on Build GRCh38
rs958243980 Chr.6: 36494869-36494869 on Build GRCh38
rs199968631 Chr.6: 36494526-36494526 on Build GRCh38
rs779619483 Chr.6: 36494940-36494940 on Build GRCh38
rs568001745 Chr.6: 36494806-36494806 on Build GRCh38
rs114299740 Chr.6: 36494785-36494785 on Build GRCh38
rs563833955 Chr.6: 36494765-36494765 on Build GRCh38
rs1014833260 Chr.6: 36494833-36494833 on Build GRCh38
rs1007592835 Chr.6: 36494722-36494722 on Build GRCh38

View Full Product Details