Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89098461-89098959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PM20D2
Entrez Gene ID
135293
Gene Name
peptidase M20 domain containing 2
Gene Aliases
ACY1L2, bA63L7.3
UniGene
Hs.356247
Gene Chromosome Location
Chr.6: 89096879-89165569 on Build GRCh38
Transcripts
XM_011535481.2 NM_080743.4 XM_006715348.2 XM_011535483.2 XM_011535484.1 XM_017010292.1 XM_017010293.1

Gene Symbol
SRSF12
Entrez Gene ID
135295
Gene Name
serine and arginine rich splicing factor 12
Gene Aliases
SFRS13B, SFRS19, SRrp35
UniGene
Hs.254414
Gene Chromosome Location
Chr.6: 89095957-89118081 on Build GRCh38
Transcripts
XM_011535481.2 NM_080743.4 XM_006715348.2 XM_011535483.2 XM_011535484.1 XM_017010292.1 XM_017010293.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1225454 c.767G>A p.R256H Chr.6: 89098597-89098597 on Build GRCh38
4839649 c.736T>G p.F246V Chr.6: 89098628-89098628 on Build GRCh38
4827521 c.533G>A p.G178E Chr.6: 89098831-89098831 on Build GRCh38
1225455 c.470T>G p.L157* Chr.6: 89098894-89098894 on Build GRCh38
5061989 c.535C>T p.R179W Chr.6: 89098829-89098829 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778295171 Chr.6: 89098880-89098880 on Build GRCh38
rs781714886 Chr.6: 89098718-89098718 on Build GRCh38
rs751046994 Chr.6: 89098699-89098699 on Build GRCh38
rs754200150 Chr.6: 89098885-89098885 on Build GRCh38
rs543172142 Chr.6: 89098764-89098764 on Build GRCh38
rs147734410 Chr.6: 89098636-89098636 on Build GRCh38
rs778685186 Chr.6: 89098700-89098700 on Build GRCh38
rs370166331 Chr.6: 89098684-89098684 on Build GRCh38
rs140575590 Chr.6: 89098621-89098621 on Build GRCh38
rs755486839 Chr.6: 89098618-89098618 on Build GRCh38
rs763173864 Chr.6: 89098599-89098599 on Build GRCh38
rs772370071 Chr.6: 89098923-89098923 on Build GRCh38
rs41273303 Chr.6: 89098631-89098631 on Build GRCh38
rs745951507 Chr.6: 89098780-89098780 on Build GRCh38
rs750276762 Chr.6: 89098692-89098692 on Build GRCh38
rs750039910 Chr.6: 89098577-89098577 on Build GRCh38
rs964558584 Chr.6: 89098541-89098541 on Build GRCh38
rs1018358315 Chr.6: 89098906-89098906 on Build GRCh38
rs201226352 Chr.6: 89098771-89098771 on Build GRCh38
rs560814640 Chr.6: 89098861-89098861 on Build GRCh38
rs773348792 Chr.6: 89098935-89098935 on Build GRCh38
rs761804002 Chr.6: 89098800-89098800 on Build GRCh38
rs1048162462 Chr.6: 89098669-89098669 on Build GRCh38
rs377575936 Chr.6: 89098556-89098556 on Build GRCh38
rs192903351 Chr.6: 89098816-89098816 on Build GRCh38
rs188489111 Chr.6: 89098500-89098500 on Build GRCh38
rs913583391 Chr.6: 89098584-89098584 on Build GRCh38
rs773489386 Chr.6: 89098550-89098550 on Build GRCh38
rs374826719 Chr.6: 89098527-89098527 on Build GRCh38
rs373285065 Chr.6: 89098597-89098597 on Build GRCh38
rs764092297 Chr.6: 89098654-89098654 on Build GRCh38
rs201200624 Chr.6: 89098828-89098828 on Build GRCh38
rs17851929 Chr.6: 89098919-89098919 on Build GRCh38
rs748510344 Chr.6: 89098540-89098540 on Build GRCh38
rs774295733 Chr.6: 89098555-89098555 on Build GRCh38
rs556351872 Chr.6: 89098562-89098562 on Build GRCh38
rs745603920 Chr.6: 89098840-89098840 on Build GRCh38
rs553653016 Chr.6: 89098625-89098625 on Build GRCh38
rs774470607 Chr.6: 89098640-89098640 on Build GRCh38
rs554248985 Chr.6: 89098693-89098693 on Build GRCh38
rs76339754 Chr.6: 89098576-89098576 on Build GRCh38
rs377465681 Chr.6: 89098607-89098607 on Build GRCh38
rs768794179 Chr.6: 89098583-89098586 on Build GRCh38
rs756560840 Chr.6: 89098866-89098866 on Build GRCh38
rs375570410 Chr.6: 89098666-89098666 on Build GRCh38
rs960934143 Chr.6: 89098817-89098817 on Build GRCh38
rs762879261 Chr.6: 89098829-89098829 on Build GRCh38
rs368884490 Chr.6: 89098536-89098536 on Build GRCh38
rs533344906 Chr.6: 89098653-89098653 on Build GRCh38
rs779577514 Chr.6: 89098598-89098598 on Build GRCh38
rs1043487845 Chr.6: 89098606-89098606 on Build GRCh38
rs746010685 Chr.6: 89098916-89098916 on Build GRCh38
rs374369058 Chr.6: 89098934-89098934 on Build GRCh38
rs779351857 Chr.6: 89098905-89098905 on Build GRCh38
rs775716564 Chr.6: 89098791-89098791 on Build GRCh38
rs373984221 Chr.6: 89098695-89098695 on Build GRCh38
rs771989086 Chr.6: 89098789-89098789 on Build GRCh38
rs766050040 Chr.6: 89098839-89098839 on Build GRCh38
rs766868282 Chr.6: 89098853-89098853 on Build GRCh38
rs748593398 Chr.6: 89098722-89098722 on Build GRCh38
rs780234250 Chr.6: 89098933-89098933 on Build GRCh38
rs748507578 Chr.6: 89098624-89098624 on Build GRCh38
rs1016310097 Chr.6: 89098725-89098725 on Build GRCh38
rs780489738 Chr.6: 89098542-89098542 on Build GRCh38
rs769970832 Chr.6: 89098542-89098542 on Build GRCh38
rs759236309 Chr.6: 89098852-89098852 on Build GRCh38
rs771155895 Chr.6: 89098770-89098770 on Build GRCh38
rs199535774 Chr.6: 89098927-89098927 on Build GRCh38
rs749507386 Chr.6: 89098558-89098558 on Build GRCh38
rs776493313 Chr.6: 89098655-89098655 on Build GRCh38
rs756613734 Chr.6: 89098729-89098729 on Build GRCh38
rs151294337 Chr.6: 89098553-89098553 on Build GRCh38
rs370679383 Chr.6: 89098821-89098821 on Build GRCh38
rs769875813 Chr.6: 89098823-89098823 on Build GRCh38
rs757624112 Chr.6: 89098886-89098886 on Build GRCh38
rs369074232 Chr.6: 89098762-89098762 on Build GRCh38

View Full Product Details