Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129690239-129690740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NRF1
Entrez Gene ID
4899
Gene Name
nuclear respiratory factor 1
Gene Aliases
ALPHA-PAL
UniGene
Hs.654363
Gene Chromosome Location
Chr.7: 129611702-129757082 on Build GRCh38
Transcripts
NM_001040110.1 NM_001293163.1 NM_001293164.1 NM_005011.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5472350 c.531G>A p.A177A Chr.7: 129690471-129690471 on Build GRCh38
4498572 c.522C>T p.H174H Chr.7: 129690462-129690462 on Build GRCh38
3831875 c.565A>C p.T189P Chr.7: 129690505-129690505 on Build GRCh38
1699351 c.587C>T p.S196F Chr.7: 129690527-129690527 on Build GRCh38
5472351 c.531G>A p.A177A Chr.7: 129690471-129690471 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047622414 Chr.7: 129690682-129690682 on Build GRCh38
rs376438034 Chr.7: 129690378-129690378 on Build GRCh38
rs554680707 Chr.7: 129690714-129690714 on Build GRCh38
rs370334260 Chr.7: 129690574-129690574 on Build GRCh38
rs567614998 Chr.7: 129690260-129690260 on Build GRCh38
rs886464993 Chr.7: 129690553-129690553 on Build GRCh38
rs745494640 Chr.7: 129690465-129690465 on Build GRCh38
rs545104361 Chr.7: 129690330-129690330 on Build GRCh38
rs760886799 Chr.7: 129690557-129690557 on Build GRCh38
rs750103330 Chr.7: 129690590-129690590 on Build GRCh38
rs774042493 Chr.7: 129690514-129690514 on Build GRCh38
rs1037557916 Chr.7: 129690277-129690277 on Build GRCh38
rs544766512 Chr.7: 129690600-129690600 on Build GRCh38
rs1034940135 Chr.7: 129690620-129690620 on Build GRCh38
rs768360011 Chr.7: 129690582-129690582 on Build GRCh38
rs374683493 Chr.7: 129690587-129690587 on Build GRCh38
rs563362343 Chr.7: 129690289-129690289 on Build GRCh38
rs186336338 Chr.7: 129690353-129690353 on Build GRCh38
rs377080587 Chr.7: 129690568-129690568 on Build GRCh38
rs55996206 Chr.7: 129690318-129690318 on Build GRCh38
rs779548887 Chr.7: 129690485-129690485 on Build GRCh38
rs369748385 Chr.7: 129690456-129690456 on Build GRCh38
rs766472419 Chr.7: 129690397-129690397 on Build GRCh38
rs779160827 Chr.7: 129690436-129690436 on Build GRCh38
rs760826845 Chr.7: 129690563-129690563 on Build GRCh38
rs1002866400 Chr.7: 129690345-129690345 on Build GRCh38
rs944591440 Chr.7: 129690376-129690376 on Build GRCh38
rs750609111 Chr.7: 129690383-129690383 on Build GRCh38
rs914460190 Chr.7: 129690334-129690334 on Build GRCh38
rs957532775 Chr.7: 129690638-129690638 on Build GRCh38
rs758884077 Chr.7: 129690585-129690585 on Build GRCh38
rs754373245 Chr.7: 129690408-129690408 on Build GRCh38
rs768338866 Chr.7: 129690364-129690364 on Build GRCh38
rs765278939 Chr.7: 129690568-129690569 on Build GRCh38
rs759985051 Chr.7: 129690567-129690567 on Build GRCh38
rs536244078 Chr.7: 129690697-129690697 on Build GRCh38
rs929151594 Chr.7: 129690646-129690646 on Build GRCh38
rs771829030 Chr.7: 129690543-129690543 on Build GRCh38
rs977747510 Chr.7: 129690704-129690704 on Build GRCh38
rs762043443 Chr.7: 129690371-129690371 on Build GRCh38
rs372860081 Chr.7: 129690560-129690560 on Build GRCh38
rs541992875 Chr.7: 129690275-129690275 on Build GRCh38
rs376710988 Chr.7: 129690462-129690462 on Build GRCh38
rs749194967 Chr.7: 129690495-129690495 on Build GRCh38
rs989247979 Chr.7: 129690659-129690659 on Build GRCh38
rs774136000 Chr.7: 129690370-129690370 on Build GRCh38
rs773477101 Chr.7: 129690550-129690550 on Build GRCh38
rs1022037413 Chr.7: 129690676-129690676 on Build GRCh38
rs530757255 Chr.7: 129690301-129690301 on Build GRCh38
rs755732796 Chr.7: 129690593-129690593 on Build GRCh38
rs747974214 Chr.7: 129690522-129690522 on Build GRCh38
rs556294810 Chr.7: 129690497-129690497 on Build GRCh38
rs980668042 Chr.7: 129690294-129690294 on Build GRCh38
rs1047102969 Chr.7: 129690324-129690324 on Build GRCh38
rs755450023 Chr.7: 129690429-129690429 on Build GRCh38
rs754157680 Chr.7: 129690578-129690578 on Build GRCh38
rs370854166 Chr.7: 129690471-129690471 on Build GRCh38
rs973281133 Chr.7: 129690602-129690602 on Build GRCh38
rs3735006 Chr.7: 129690513-129690513 on Build GRCh38
rs972053490 Chr.7: 129690580-129690580 on Build GRCh38
rs369507589 Chr.7: 129690363-129690363 on Build GRCh38
rs753065116 Chr.7: 129690447-129690447 on Build GRCh38
rs760788740 Chr.7: 129690388-129690388 on Build GRCh38

View Full Product Details