Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108034120-108034669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LAMB4
Entrez Gene ID
22798
Gene Name
laminin subunit beta 4
Gene Aliases
-
UniGene
Hs.62022
Gene Chromosome Location
Chr.7: 108011662-108130356 on Build GRCh38
Transcripts
NM_001318046.1 NM_007356.2 XM_011515975.1 XM_011515978.1 XM_011515979.2 XM_011515980.2 XM_017011879.1 XM_017011880.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3255169 c.4806G>T p.K1602N Chr.7: 108034220-108034220 on Build GRCh38
1131736 c.4756G>A p.A1586T Chr.7: 108034270-108034270 on Build GRCh38
3255172 c.4710C>T p.D1570D Chr.7: 108034316-108034316 on Build GRCh38
6858483 c.4764T>C p.S1588S Chr.7: 108034262-108034262 on Build GRCh38
4811548 c.4738C>G p.Q1580E Chr.7: 108034288-108034288 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs116983773 Chr.7: 108034510-108034510 on Build GRCh38
rs34682332 Chr.7: 108034346-108034346 on Build GRCh38
rs777238295 Chr.7: 108034173-108034173 on Build GRCh38
rs377182839 Chr.7: 108034207-108034207 on Build GRCh38
rs914401848 Chr.7: 108034460-108034460 on Build GRCh38
rs773286123 Chr.7: 108034222-108034222 on Build GRCh38
rs965926168 Chr.7: 108034234-108034234 on Build GRCh38
rs1006328848 Chr.7: 108034538-108034538 on Build GRCh38
rs569347497 Chr.7: 108034226-108034226 on Build GRCh38
rs781503192 Chr.7: 108034286-108034286 on Build GRCh38
rs755573246 Chr.7: 108034283-108034283 on Build GRCh38
rs936984069 Chr.7: 108034544-108034544 on Build GRCh38
rs973288334 Chr.7: 108034477-108034477 on Build GRCh38
rs749501707 Chr.7: 108034312-108034312 on Build GRCh38
rs983759639 Chr.7: 108034259-108034259 on Build GRCh38
rs545324239 Chr.7: 108034473-108034473 on Build GRCh38
rs756884085 Chr.7: 108034269-108034269 on Build GRCh38
rs77263109 Chr.7: 108034518-108034518 on Build GRCh38
rs370387445 Chr.7: 108034236-108034236 on Build GRCh38
rs369664071 Chr.7: 108034171-108034171 on Build GRCh38
rs904341829 Chr.7: 108034634-108034634 on Build GRCh38
rs760582536 Chr.7: 108034364-108034364 on Build GRCh38
rs772092025 Chr.7: 108034357-108034357 on Build GRCh38
rs767293857 Chr.7: 108034250-108034250 on Build GRCh38
rs763845029 Chr.7: 108034265-108034265 on Build GRCh38
rs753533083 Chr.7: 108034268-108034268 on Build GRCh38
rs937192002 Chr.7: 108034376-108034376 on Build GRCh38
rs780771826 Chr.7: 108034196-108034196 on Build GRCh38
rs200506280 Chr.7: 108034214-108034214 on Build GRCh38
rs889764485 Chr.7: 108034385-108034385 on Build GRCh38
rs996436095 Chr.7: 108034531-108034531 on Build GRCh38
rs746337715 Chr.7: 108034587-108034587 on Build GRCh38
rs554793798 Chr.7: 108034384-108034384 on Build GRCh38
rs753627040 Chr.7: 108034388-108034388 on Build GRCh38
rs1019940266 Chr.7: 108034153-108034153 on Build GRCh38
rs140745252 Chr.7: 108034273-108034273 on Build GRCh38
rs902841121 Chr.7: 108034169-108034169 on Build GRCh38
rs774333051 Chr.7: 108034245-108034245 on Build GRCh38
rs1020277537 Chr.7: 108034470-108034470 on Build GRCh38
rs28621880 Chr.7: 108034366-108034366 on Build GRCh38
rs962015753 Chr.7: 108034317-108034317 on Build GRCh38
rs1000833519 Chr.7: 108034379-108034379 on Build GRCh38
rs200072693 Chr.7: 108034249-108034249 on Build GRCh38
rs1056071831 Chr.7: 108034459-108034459 on Build GRCh38
rs573359256 Chr.7: 108034272-108034272 on Build GRCh38
rs936780889 Chr.7: 108034646-108034646 on Build GRCh38
rs945917881 Chr.7: 108034580-108034580 on Build GRCh38
rs952470244 Chr.7: 108034524-108034524 on Build GRCh38
rs147242067 Chr.7: 108034257-108034257 on Build GRCh38
rs78883502 Chr.7: 108034548-108034548 on Build GRCh38
rs765853261 Chr.7: 108034199-108034201 on Build GRCh38
rs1040634276 Chr.7: 108034529-108034529 on Build GRCh38
rs1041427476 Chr.7: 108034507-108034507 on Build GRCh38
rs567868663 Chr.7: 108034141-108034141 on Build GRCh38
rs201807198 Chr.7: 108034213-108034213 on Build GRCh38
rs748595241 Chr.7: 108034290-108034290 on Build GRCh38
rs770026241 Chr.7: 108034305-108034305 on Build GRCh38
rs189650280 Chr.7: 108034531-108034531 on Build GRCh38
rs752199601 Chr.7: 108034215-108034215 on Build GRCh38
rs775255707 Chr.7: 108034363-108034363 on Build GRCh38
rs895686322 Chr.7: 108034340-108034340 on Build GRCh38
rs926987391 Chr.7: 108034539-108034539 on Build GRCh38
rs902134124 Chr.7: 108034530-108034531 on Build GRCh38
rs778124522 Chr.7: 108034311-108034311 on Build GRCh38
rs771087039 Chr.7: 108034332-108034332 on Build GRCh38
rs748875357 Chr.7: 108034174-108034174 on Build GRCh38
rs1007134762 Chr.7: 108034453-108034453 on Build GRCh38
rs760360895 Chr.7: 108034262-108034262 on Build GRCh38
rs571865369 Chr.7: 108034341-108034341 on Build GRCh38
rs769228142 Chr.7: 108034208-108034208 on Build GRCh38
rs917116438 Chr.7: 108034555-108034555 on Build GRCh38
rs550193850 Chr.7: 108034531-108034534 on Build GRCh38
rs372649955 Chr.7: 108034569-108034569 on Build GRCh38

View Full Product Details