Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 2613464-2613663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ1
Entrez Gene ID
3784
Gene Name
potassium voltage-gated channel subfamily Q member 1
Gene Aliases
ATFB1, ATFB3, JLNS1, KCNA8, KCNA9, KVLQT1, Kv1.9, Kv7.1, LQT, LQT1, RWS, SQT2, WRS
UniGene
Hs.95162
Gene Chromosome Location
Chr.11: 2444991-2849110 on Build GRCh38
Transcripts
NM_000218.2 NM_181798.1 NR_002728.3

Gene Symbol
KCNQ1OT1
Entrez Gene ID
10984
Gene Name
KCNQ1 opposite strand/antisense transcript 1 (non-protein coding)
Gene Aliases
KCNQ1-AS2, KCNQ10T1, Kncq1, KvDMR1, KvLQT1-AS, LIT1, NCRNA00012
UniGene
Hs.604823
Gene Chromosome Location
Chr.11: 2608328-2699998 on Build GRCh38
Transcripts
NM_000218.2 NM_181798.1 NR_002728.3

SNP Detail

SNP ID Chromosome Location
rs752401885 Chr.11: 2613493-2613493 on Build GRCh38
rs1000331860 Chr.11: 2613487-2613487 on Build GRCh38
rs1031801335 Chr.11: 2613557-2613557 on Build GRCh38
rs954041875 Chr.11: 2613530-2613530 on Build GRCh38
rs1052734867 Chr.11: 2613489-2613489 on Build GRCh38
rs562874854 Chr.11: 2613539-2613539 on Build GRCh38
rs998062456 Chr.11: 2613518-2613518 on Build GRCh38
rs140152372 Chr.11: 2613597-2613597 on Build GRCh38
rs544927222 Chr.11: 2613529-2613529 on Build GRCh38
rs763680153 Chr.11: 2613626-2613626 on Build GRCh38
rs868355733 Chr.11: 2613592-2613592 on Build GRCh38
rs964580919 Chr.11: 2613584-2613584 on Build GRCh38
rs1046539171 Chr.11: 2613495-2613495 on Build GRCh38
rs957139409 Chr.11: 2613560-2613560 on Build GRCh38
rs902716240 Chr.11: 2613543-2613543 on Build GRCh38
rs185276398 Chr.11: 2613492-2613492 on Build GRCh38
rs541269843 Chr.11: 2613576-2613576 on Build GRCh38
rs562359947 Chr.11: 2613490-2613490 on Build GRCh38
rs1011168426 Chr.11: 2613568-2613568 on Build GRCh38
rs999708858 Chr.11: 2613549-2613549 on Build GRCh38

View Full Product Details