Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2700587-2701078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AMZ1
Entrez Gene ID
155185
Gene Name
archaelysin family metallopeptidase 1
Gene Aliases
-
UniGene
Hs.42221
Gene Chromosome Location
Chr.7: 2679522-2767486 on Build GRCh38
Transcripts
NM_001284355.2 NM_001321766.1 NM_133463.3 XM_011515150.2 XM_011515151.2 XM_017011774.1 XM_017011775.1 XM_017011776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
746293 c.158C>T p.T53M Chr.7: 2700609-2700609 on Build GRCh38
5637141 c.182G>T p.R61L Chr.7: 2700633-2700633 on Build GRCh38
746292 c.302T>C p.I101T Chr.7: 2700753-2700753 on Build GRCh38
3637753 c.204G>A p.L68L Chr.7: 2700655-2700655 on Build GRCh38
6599486 c.181C>T p.R61C Chr.7: 2700632-2700632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754712229 Chr.7: 2700758-2700758 on Build GRCh38
rs147624387 Chr.7: 2700740-2700740 on Build GRCh38
rs745712139 Chr.7: 2700639-2700639 on Build GRCh38
rs748921179 Chr.7: 2700686-2700686 on Build GRCh38
rs761194583 Chr.7: 2700646-2700646 on Build GRCh38
rs1029773426 Chr.7: 2700850-2700850 on Build GRCh38
rs530692397 Chr.7: 2700986-2700986 on Build GRCh38
rs765829705 Chr.7: 2701029-2701029 on Build GRCh38
rs368870586 Chr.7: 2700664-2700664 on Build GRCh38
rs751164383 Chr.7: 2700799-2700799 on Build GRCh38
rs140485254 Chr.7: 2700746-2700746 on Build GRCh38
rs779769298 Chr.7: 2700676-2700676 on Build GRCh38
rs189874698 Chr.7: 2700764-2700764 on Build GRCh38
rs528198788 Chr.7: 2700935-2700935 on Build GRCh38
rs543403758 Chr.7: 2700665-2700665 on Build GRCh38
rs766459246 Chr.7: 2700800-2700801 on Build GRCh38
rs966287802 Chr.7: 2700710-2700710 on Build GRCh38
rs985186411 Chr.7: 2701030-2701030 on Build GRCh38
rs761855465 Chr.7: 2700782-2700782 on Build GRCh38
rs867673857 Chr.7: 2700701-2700701 on Build GRCh38
rs1036589082 Chr.7: 2700817-2700817 on Build GRCh38
rs918406363 Chr.7: 2700917-2700917 on Build GRCh38
rs560557432 Chr.7: 2700962-2700962 on Build GRCh38
rs558749928 Chr.7: 2700773-2700773 on Build GRCh38
rs1006543091 Chr.7: 2701051-2701051 on Build GRCh38
rs114941505 Chr.7: 2700844-2700844 on Build GRCh38
rs1016549556 Chr.7: 2701052-2701052 on Build GRCh38
rs28514648 Chr.7: 2700870-2700870 on Build GRCh38
rs758083347 Chr.7: 2700798-2700798 on Build GRCh38
rs867766709 Chr.7: 2700696-2700696 on Build GRCh38
rs199863314 Chr.7: 2700749-2700749 on Build GRCh38
rs376751656 Chr.7: 2700609-2700609 on Build GRCh38
rs766782779 Chr.7: 2700658-2700658 on Build GRCh38
rs1052628950 Chr.7: 2700975-2700975 on Build GRCh38
rs768068698 Chr.7: 2700644-2700644 on Build GRCh38
rs112599565 Chr.7: 2700759-2700759 on Build GRCh38
rs550493677 Chr.7: 2701016-2701016 on Build GRCh38
rs149751599 Chr.7: 2700751-2700751 on Build GRCh38
rs184451550 Chr.7: 2700950-2700950 on Build GRCh38
rs142280090 Chr.7: 2700636-2700636 on Build GRCh38
rs547984929 Chr.7: 2700739-2700739 on Build GRCh38
rs889330277 Chr.7: 2701031-2701031 on Build GRCh38
rs144711878 Chr.7: 2700697-2700697 on Build GRCh38
rs545804269 Chr.7: 2700914-2700914 on Build GRCh38
rs371746483 Chr.7: 2700715-2700715 on Build GRCh38
rs764881993 Chr.7: 2700791-2700791 on Build GRCh38
rs752419606 Chr.7: 2700793-2700793 on Build GRCh38
rs148513555 Chr.7: 2700643-2700643 on Build GRCh38
rs375324319 Chr.7: 2700760-2700760 on Build GRCh38
rs188324040 Chr.7: 2700961-2700961 on Build GRCh38
rs180672044 Chr.7: 2700818-2700818 on Build GRCh38
rs772751076 Chr.7: 2700648-2700648 on Build GRCh38
rs376501926 Chr.7: 2700720-2700720 on Build GRCh38
rs535183090 Chr.7: 2700783-2700783 on Build GRCh38
rs368298498 Chr.7: 2700728-2700728 on Build GRCh38
rs745639103 Chr.7: 2700616-2700616 on Build GRCh38
rs755015835 Chr.7: 2700628-2700628 on Build GRCh38
rs894672626 Chr.7: 2701026-2701026 on Build GRCh38
rs753711438 Chr.7: 2700784-2700784 on Build GRCh38
rs141821781 Chr.7: 2700660-2700660 on Build GRCh38
rs997762485 Chr.7: 2700836-2700836 on Build GRCh38
rs773859968 Chr.7: 2700709-2700709 on Build GRCh38
rs576798970 Chr.7: 2700904-2700904 on Build GRCh38
rs145132421 Chr.7: 2700610-2700610 on Build GRCh38
rs150760039 Chr.7: 2701028-2701028 on Build GRCh38
rs1028661759 Chr.7: 2700724-2700724 on Build GRCh38
rs754107925 Chr.7: 2700623-2700623 on Build GRCh38
rs757995499 Chr.7: 2700761-2700761 on Build GRCh38
rs1000504464 Chr.7: 2700923-2700923 on Build GRCh38
rs756710822 Chr.7: 2700803-2700803 on Build GRCh38
rs750360131 Chr.7: 2700671-2700671 on Build GRCh38
rs73281171 Chr.7: 2700931-2700931 on Build GRCh38
rs371648045 Chr.7: 2700765-2700765 on Build GRCh38
rs144827365 Chr.7: 2700637-2700637 on Build GRCh38
rs556895537 Chr.7: 2700878-2700878 on Build GRCh38
rs903900939 Chr.7: 2700809-2700809 on Build GRCh38
rs538800719 Chr.7: 2700622-2700622 on Build GRCh38
rs761188501 Chr.7: 2700654-2700654 on Build GRCh38
rs375951352 Chr.7: 2700777-2700777 on Build GRCh38
rs887664993 Chr.7: 2700797-2700797 on Build GRCh38
rs201758008 Chr.7: 2700719-2700719 on Build GRCh38
rs957837911 Chr.7: 2700936-2700936 on Build GRCh38
rs773699105 Chr.7: 2700902-2700902 on Build GRCh38
rs935400009 Chr.7: 2700972-2700972 on Build GRCh38
rs746303742 Chr.7: 2700766-2700766 on Build GRCh38
rs999702885 Chr.7: 2700810-2700810 on Build GRCh38
rs368867595 Chr.7: 2700779-2700779 on Build GRCh38
rs1010524658 Chr.7: 2700944-2700944 on Build GRCh38
rs953751731 Chr.7: 2700855-2700855 on Build GRCh38
rs1050137423 Chr.7: 2700895-2700895 on Build GRCh38
rs778403754 Chr.7: 2700694-2700694 on Build GRCh38
rs753714123 Chr.7: 2700750-2700750 on Build GRCh38
rs925393394 Chr.7: 2700969-2700969 on Build GRCh38
rs937248674 Chr.7: 2700662-2700662 on Build GRCh38
rs528244160 Chr.7: 2700729-2700729 on Build GRCh38
rs901999493 Chr.7: 2700976-2700976 on Build GRCh38
rs541662977 Chr.7: 2700649-2700649 on Build GRCh38
rs140865604 Chr.7: 2700692-2700692 on Build GRCh38
rs569976249 Chr.7: 2700762-2700762 on Build GRCh38
rs754805425 Chr.7: 2700789-2700789 on Build GRCh38
rs1016645887 Chr.7: 2700887-2700887 on Build GRCh38
rs962426658 Chr.7: 2700909-2700909 on Build GRCh38
rs372832452 Chr.7: 2700804-2700804 on Build GRCh38
rs138956355 Chr.7: 2700632-2700632 on Build GRCh38
rs971502993 Chr.7: 2700955-2700955 on Build GRCh38
rs768393104 Chr.7: 2700691-2700691 on Build GRCh38
rs761209850 Chr.7: 2700679-2700687 on Build GRCh38
rs150643476 Chr.7: 2700673-2700673 on Build GRCh38
rs775509838 Chr.7: 2700718-2700718 on Build GRCh38
rs200084826 Chr.7: 2700745-2700745 on Build GRCh38
rs559083782 Chr.7: 2700633-2700633 on Build GRCh38

View Full Product Details