Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42857221-42857732 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AOC3
Entrez Gene ID
8639
Gene Name
amine oxidase, copper containing 3
Gene Aliases
HPAO, SSAO, VAP-1, VAP1
UniGene
Hs.198241
Gene Chromosome Location
Chr.17: 42851123-42858130 on Build GRCh38
Transcripts
NM_001277731.1 NM_001277732.1 NM_003734.3 NR_102422.1 XM_011525419.1 XM_011525420.2

SNP Detail

SNP ID Chromosome Location
rs754275077 Chr.17: 42857279-42857279 on Build GRCh38
rs1046711626 Chr.17: 42857416-42857416 on Build GRCh38
rs35551758 Chr.17: 42857583-42857583 on Build GRCh38
rs34120767 Chr.17: 42857435-42857435 on Build GRCh38
rs957942120 Chr.17: 42857246-42857246 on Build GRCh38
rs540517159 Chr.17: 42857560-42857560 on Build GRCh38
rs977213583 Chr.17: 42857411-42857411 on Build GRCh38
rs990113157 Chr.17: 42857259-42857259 on Build GRCh38
rs771361769 Chr.17: 42857670-42857670 on Build GRCh38
rs1022749417 Chr.17: 42857600-42857600 on Build GRCh38
rs921020875 Chr.17: 42857657-42857657 on Build GRCh38
rs34751259 Chr.17: 42857490-42857490 on Build GRCh38
rs924409581 Chr.17: 42857627-42857627 on Build GRCh38
rs3179583 Chr.17: 42857624-42857624 on Build GRCh38
rs991779661 Chr.17: 42857308-42857308 on Build GRCh38
rs888125176 Chr.17: 42857708-42857708 on Build GRCh38
rs1003587916 Chr.17: 42857557-42857557 on Build GRCh38
rs575205744 Chr.17: 42857446-42857446 on Build GRCh38
rs924461978 Chr.17: 42857420-42857420 on Build GRCh38
rs887372657 Chr.17: 42857677-42857677 on Build GRCh38
rs9914497 Chr.17: 42857428-42857428 on Build GRCh38
rs1013274435 Chr.17: 42857421-42857421 on Build GRCh38
rs959401344 Chr.17: 42857254-42857254 on Build GRCh38
rs912719435 Chr.17: 42857363-42857363 on Build GRCh38
rs1020280423 Chr.17: 42857544-42857544 on Build GRCh38
rs568065772 Chr.17: 42857408-42857408 on Build GRCh38
rs886798712 Chr.17: 42857419-42857419 on Build GRCh38
rs942802019 Chr.17: 42857619-42857619 on Build GRCh38
rs942835386 Chr.17: 42857364-42857364 on Build GRCh38
rs968579766 Chr.17: 42857618-42857618 on Build GRCh38
rs111475752 Chr.17: 42857322-42857322 on Build GRCh38
rs899463058 Chr.17: 42857439-42857439 on Build GRCh38
rs988097983 Chr.17: 42857515-42857515 on Build GRCh38
rs959753115 Chr.17: 42857575-42857575 on Build GRCh38
rs977329108 Chr.17: 42857625-42857625 on Build GRCh38
rs975953377 Chr.17: 42857646-42857646 on Build GRCh38
rs546058622 Chr.17: 42857465-42857465 on Build GRCh38
rs1015740221 Chr.17: 42857545-42857545 on Build GRCh38
rs1032181235 Chr.17: 42857485-42857485 on Build GRCh38
rs977026162 Chr.17: 42857656-42857656 on Build GRCh38
rs181825729 Chr.17: 42857552-42857552 on Build GRCh38
rs112435651 Chr.17: 42857577-42857577 on Build GRCh38
rs914484762 Chr.17: 42857335-42857335 on Build GRCh38
rs1047330331 Chr.17: 42857674-42857674 on Build GRCh38
rs943883188 Chr.17: 42857641-42857641 on Build GRCh38
rs967402142 Chr.17: 42857554-42857554 on Build GRCh38
rs1048045535 Chr.17: 42857705-42857705 on Build GRCh38
rs1039892999 Chr.17: 42857405-42857405 on Build GRCh38
rs1048574367 Chr.17: 42857459-42857459 on Build GRCh38
rs986908138 Chr.17: 42857358-42857358 on Build GRCh38

View Full Product Details