Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 47564940-47565436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARAF
Entrez Gene ID
369
Gene Name
A-Raf proto-oncogene, serine/threonine kinase
Gene Aliases
A-RAF, ARAF1, PKS2, RAFA1
UniGene
Hs.446641
Gene Chromosome Location
Chr.X: 47561100-47571921 on Build GRCh38
Transcripts
NM_001256196.1 NM_001256197.1 NM_001654.4 XM_006724529.2 XM_011543906.2 XM_011543908.2

SNP Detail

SNP ID Chromosome Location
rs936264056 Chr.X: 47565018-47565018 on Build GRCh38
rs774526444 Chr.X: 47565130-47565130 on Build GRCh38
rs869122414 Chr.X: 47565379-47565379 on Build GRCh38
rs761480447 Chr.X: 47565292-47565292 on Build GRCh38
rs184861193 Chr.X: 47565302-47565302 on Build GRCh38
rs151113368 Chr.X: 47565217-47565217 on Build GRCh38
rs763988785 Chr.X: 47564965-47564965 on Build GRCh38
rs775921012 Chr.X: 47565280-47565280 on Build GRCh38
rs146376823 Chr.X: 47565291-47565291 on Build GRCh38
rs947760439 Chr.X: 47565104-47565104 on Build GRCh38
rs779625869 Chr.X: 47565329-47565329 on Build GRCh38
rs957274850 Chr.X: 47565208-47565208 on Build GRCh38
rs749784842 Chr.X: 47565244-47565244 on Build GRCh38
rs746296307 Chr.X: 47564973-47564973 on Build GRCh38
rs764756092 Chr.X: 47565164-47565164 on Build GRCh38
rs745997765 Chr.X: 47565263-47565263 on Build GRCh38
rs761931410 Chr.X: 47565313-47565313 on Build GRCh38
rs757678586 Chr.X: 47565327-47565327 on Build GRCh38
rs770926246 Chr.X: 47565118-47565118 on Build GRCh38
rs887790711 Chr.X: 47564962-47564962 on Build GRCh38
rs958730062 Chr.X: 47565343-47565343 on Build GRCh38
rs538541233 Chr.X: 47565011-47565011 on Build GRCh38
rs371173578 Chr.X: 47565228-47565228 on Build GRCh38
rs922116622 Chr.X: 47565258-47565258 on Build GRCh38
rs756786368 Chr.X: 47564970-47564970 on Build GRCh38
rs2071776 Chr.X: 47565216-47565216 on Build GRCh38
rs780124992 Chr.X: 47565376-47565376 on Build GRCh38
rs62636600 Chr.X: 47565051-47565051 on Build GRCh38
rs1036402369 Chr.X: 47565304-47565304 on Build GRCh38
rs763947223 Chr.X: 47565156-47565156 on Build GRCh38
rs772013716 Chr.X: 47564996-47564998 on Build GRCh38
rs1050828066 Chr.X: 47565240-47565240 on Build GRCh38
rs776439001 Chr.X: 47565161-47565161 on Build GRCh38
rs386825116 Chr.X: 47565216-47565217 on Build GRCh38
rs779536112 Chr.X: 47565252-47565252 on Build GRCh38
rs113512925 Chr.X: 47565259-47565259 on Build GRCh38
rs760446501 Chr.X: 47565155-47565155 on Build GRCh38
rs766074870 Chr.X: 47565210-47565210 on Build GRCh38
rs143159753 Chr.X: 47565335-47565335 on Build GRCh38
rs35192383 Chr.X: 47565167-47565168 on Build GRCh38
rs1014092417 Chr.X: 47565005-47565005 on Build GRCh38
rs375468186 Chr.X: 47565284-47565284 on Build GRCh38
rs758004948 Chr.X: 47564985-47564985 on Build GRCh38
rs371247311 Chr.X: 47565308-47565308 on Build GRCh38
rs139093707 Chr.X: 47565301-47565301 on Build GRCh38
rs756483901 Chr.X: 47565036-47565036 on Build GRCh38
rs754272256 Chr.X: 47565318-47565318 on Build GRCh38
rs747224772 Chr.X: 47565385-47565385 on Build GRCh38
rs869307224 Chr.X: 47565170-47565170 on Build GRCh38
rs749734176 Chr.X: 47565106-47565106 on Build GRCh38
rs200542934 Chr.X: 47565166-47565166 on Build GRCh38
rs768265504 Chr.X: 47565193-47565193 on Build GRCh38
rs779648831 Chr.X: 47565010-47565010 on Build GRCh38
rs896950658 Chr.X: 47564975-47564975 on Build GRCh38
rs1001882020 Chr.X: 47565192-47565192 on Build GRCh38
rs869179262 Chr.X: 47565344-47565344 on Build GRCh38
rs764809054 Chr.X: 47565279-47565279 on Build GRCh38
rs778205718 Chr.X: 47565066-47565066 on Build GRCh38
rs765207392 Chr.X: 47565297-47565297 on Build GRCh38
rs774684341 Chr.X: 47564968-47564968 on Build GRCh38
rs550399272 Chr.X: 47565249-47565249 on Build GRCh38
rs758558184 Chr.X: 47565339-47565339 on Build GRCh38
rs746126878 Chr.X: 47565132-47565132 on Build GRCh38
rs140747108 Chr.X: 47565365-47565365 on Build GRCh38
rs201394917 Chr.X: 47565162-47565162 on Build GRCh38
rs773119164 Chr.X: 47565300-47565300 on Build GRCh38
rs996459358 Chr.X: 47565188-47565188 on Build GRCh38
rs56202823 Chr.X: 47565309-47565309 on Build GRCh38
rs768961381 Chr.X: 47565403-47565403 on Build GRCh38
rs949478395 Chr.X: 47565406-47565406 on Build GRCh38
rs747729615 Chr.X: 47565411-47565411 on Build GRCh38
rs370327383 Chr.X: 47565146-47565146 on Build GRCh38

View Full Product Details