Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228096951-228097459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF1
Entrez Gene ID
375
Gene Name
ADP ribosylation factor 1
Gene Aliases
-
UniGene
Hs.286221
Gene Chromosome Location
Chr.1: 228082660-228099212 on Build GRCh38
Transcripts
NM_001024226.1 NM_001024227.1 NM_001024228.1 NM_001658.3 XM_017001233.1 NR_037415.1

Gene Symbol
MIR3620
Entrez Gene ID
100500810
Gene Name
microRNA 3620
Gene Aliases
mir-3620
UniGene
Gene Chromosome Location
Chr.1: 228097263-228097341 on Build GRCh38
Transcripts
NM_001024226.1 NM_001024227.1 NM_001024228.1 NM_001658.3 XM_017001233.1 NR_037415.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5469978 c.166G>A p.V56M Chr.1: 228097359-228097359 on Build GRCh38
1317574 c.165C>T p.T55T Chr.1: 228097358-228097358 on Build GRCh38
1183326 c.55C>T p.R19C Chr.1: 228097169-228097169 on Build GRCh38
5162053 c.149-9C>T p.? Chr.1: 228097333-228097333 on Build GRCh38
1639743 c.104A>G p.Y35C Chr.1: 228097218-228097218 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759638959 Chr.1: 228097087-228097087 on Build GRCh38
rs762183426 Chr.1: 228097091-228097091 on Build GRCh38
rs778231514 Chr.1: 228097295-228097295 on Build GRCh38
rs41270179 Chr.1: 228097028-228097028 on Build GRCh38
rs981862206 Chr.1: 228096987-228096987 on Build GRCh38
rs768803103 Chr.1: 228097336-228097336 on Build GRCh38
rs376518139 Chr.1: 228097021-228097021 on Build GRCh38
rs758944972 Chr.1: 228097291-228097291 on Build GRCh38
rs970433652 Chr.1: 228096981-228096981 on Build GRCh38
rs747469592 Chr.1: 228097302-228097302 on Build GRCh38
rs764690724 Chr.1: 228097424-228097424 on Build GRCh38
rs776618422 Chr.1: 228097075-228097075 on Build GRCh38
rs751395152 Chr.1: 228097102-228097102 on Build GRCh38
rs771140686 Chr.1: 228097069-228097069 on Build GRCh38
rs11541555 Chr.1: 228097156-228097156 on Build GRCh38
rs368994066 Chr.1: 228097246-228097246 on Build GRCh38
rs11541558 Chr.1: 228097385-228097385 on Build GRCh38
rs771191633 Chr.1: 228097147-228097147 on Build GRCh38
rs754506796 Chr.1: 228097397-228097397 on Build GRCh38
rs369631308 Chr.1: 228096993-228096993 on Build GRCh38
rs966606686 Chr.1: 228097035-228097035 on Build GRCh38
rs752069371 Chr.1: 228097428-228097428 on Build GRCh38
rs769901189 Chr.1: 228097329-228097329 on Build GRCh38
rs369190394 Chr.1: 228097333-228097333 on Build GRCh38
rs2070960 Chr.1: 228097290-228097290 on Build GRCh38
rs757653610 Chr.1: 228097307-228097307 on Build GRCh38
rs775417374 Chr.1: 228097198-228097198 on Build GRCh38
rs765917983 Chr.1: 228097376-228097376 on Build GRCh38
rs201883341 Chr.1: 228097130-228097130 on Build GRCh38
rs764037349 Chr.1: 228097100-228097100 on Build GRCh38
rs942880423 Chr.1: 228097073-228097073 on Build GRCh38
rs781347783 Chr.1: 228097309-228097309 on Build GRCh38
rs375926182 Chr.1: 228097240-228097240 on Build GRCh38
rs772867236 Chr.1: 228097370-228097370 on Build GRCh38
rs368437375 Chr.1: 228097129-228097129 on Build GRCh38
rs769925136 Chr.1: 228097186-228097186 on Build GRCh38
rs760840584 Chr.1: 228097286-228097287 on Build GRCh38
rs997468586 Chr.1: 228097023-228097023 on Build GRCh38
rs775705993 Chr.1: 228097286-228097290 on Build GRCh38
rs372015059 Chr.1: 228097388-228097388 on Build GRCh38
rs760445481 Chr.1: 228097373-228097373 on Build GRCh38
rs552986915 Chr.1: 228097358-228097358 on Build GRCh38
rs376688688 Chr.1: 228097301-228097301 on Build GRCh38
rs747344148 Chr.1: 228097143-228097143 on Build GRCh38
rs755663535 Chr.1: 228097270-228097270 on Build GRCh38
rs774460920 Chr.1: 228097349-228097349 on Build GRCh38
rs746041095 Chr.1: 228097183-228097183 on Build GRCh38
rs774332380 Chr.1: 228097210-228097210 on Build GRCh38
rs377716619 Chr.1: 228097094-228097094 on Build GRCh38
rs11541562 Chr.1: 228097149-228097149 on Build GRCh38
rs879036238 Chr.1: 228097217-228097217 on Build GRCh38
rs934038052 Chr.1: 228097288-228097288 on Build GRCh38
rs750093483 Chr.1: 228097258-228097258 on Build GRCh38
rs375115515 Chr.1: 228097109-228097109 on Build GRCh38
rs765973759 Chr.1: 228097279-228097279 on Build GRCh38
rs749994289 Chr.1: 228097107-228097107 on Build GRCh38
rs202149186 Chr.1: 228097328-228097328 on Build GRCh38
rs746211512 Chr.1: 228097301-228097302 on Build GRCh38
rs778178441 Chr.1: 228097134-228097134 on Build GRCh38
rs989736002 Chr.1: 228097001-228097001 on Build GRCh38
rs537167396 Chr.1: 228097062-228097062 on Build GRCh38
rs965912831 Chr.1: 228097003-228097003 on Build GRCh38
rs776938681 Chr.1: 228097174-228097174 on Build GRCh38
rs753396348 Chr.1: 228097394-228097394 on Build GRCh38
rs879062233 Chr.1: 228097296-228097296 on Build GRCh38
rs923579456 Chr.1: 228096989-228096989 on Build GRCh38
rs746117851 Chr.1: 228097314-228097314 on Build GRCh38
rs749353761 Chr.1: 228097335-228097335 on Build GRCh38
rs141769989 Chr.1: 228097132-228097132 on Build GRCh38
rs767176898 Chr.1: 228097255-228097255 on Build GRCh38
rs763048237 Chr.1: 228097207-228097207 on Build GRCh38
rs575298975 Chr.1: 228097285-228097286 on Build GRCh38
rs766707001 Chr.1: 228097334-228097334 on Build GRCh38
rs371761353 Chr.1: 228097103-228097103 on Build GRCh38

View Full Product Details