Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219535238-219535726 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASIC4
Entrez Gene ID
55515
Gene Name
acid sensing ion channel subunit family member 4
Gene Aliases
ACCN4, BNAC4
UniGene
Hs.87469
Gene Chromosome Location
Chr.2: 219514170-219538772 on Build GRCh38
Transcripts
NM_182847.2 XM_005246671.2 XM_017004439.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6510635 c.1601C>A p.T534N Chr.2: 219535315-219535315 on Build GRCh38
5683710 c.1591C>A p.R531S Chr.2: 219535305-219535305 on Build GRCh38
5353836 c.1610+4_1610+5delTG p.? Chr.2: 219535328-219535329 on Build GRCh38
6510636 c.1602C>A p.T534T Chr.2: 219535316-219535316 on Build GRCh38
73551 c.1539G>T p.M513I Chr.2: 219535253-219535253 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927137965 Chr.2: 219535557-219535558 on Build GRCh38
rs200985286 Chr.2: 219535441-219535441 on Build GRCh38
rs951364011 Chr.2: 219535516-219535517 on Build GRCh38
rs150579390 Chr.2: 219535438-219535441 on Build GRCh38
rs141949099 Chr.2: 219535342-219535342 on Build GRCh38
rs138860330 Chr.2: 219535340-219535340 on Build GRCh38
rs1001288729 Chr.2: 219535424-219535427 on Build GRCh38
rs757037555 Chr.2: 219535338-219535338 on Build GRCh38
rs765381934 Chr.2: 219535320-219535320 on Build GRCh38
rs375945018 Chr.2: 219535549-219535552 on Build GRCh38
rs376923243 Chr.2: 219535372-219535372 on Build GRCh38
rs531834252 Chr.2: 219535548-219535548 on Build GRCh38
rs572878685 Chr.2: 219535328-219535328 on Build GRCh38
rs760053503 Chr.2: 219535327-219535328 on Build GRCh38
rs1027203208 Chr.2: 219535602-219535602 on Build GRCh38
rs13388954 Chr.2: 219535340-219535340 on Build GRCh38
rs148631476 Chr.2: 219535306-219535306 on Build GRCh38
rs775623667 Chr.2: 219535371-219535371 on Build GRCh38
rs982784775 Chr.2: 219535542-219535545 on Build GRCh38
rs757620772 Chr.2: 219535336-219535336 on Build GRCh38
rs747438300 Chr.2: 219535268-219535268 on Build GRCh38
rs369501588 Chr.2: 219535310-219535310 on Build GRCh38
rs772493952 Chr.2: 219535346-219535347 on Build GRCh38
rs777884983 Chr.2: 219535342-219535343 on Build GRCh38
rs1044962684 Chr.2: 219535467-219535467 on Build GRCh38
rs551549729 Chr.2: 219535588-219535588 on Build GRCh38
rs34965338 Chr.2: 219535683-219535683 on Build GRCh38
rs1030956117 Chr.2: 219535502-219535502 on Build GRCh38
rs796174751 Chr.2: 219535343-219535344 on Build GRCh38
rs760199139 Chr.2: 219535333-219535334 on Build GRCh38
rs369231217 Chr.2: 219535549-219535549 on Build GRCh38
rs571640905 Chr.2: 219535648-219535648 on Build GRCh38
rs771291936 Chr.2: 219535584-219535587 on Build GRCh38
rs758615587 Chr.2: 219535323-219535323 on Build GRCh38
rs201444713 Chr.2: 219535324-219535324 on Build GRCh38
rs12477394 Chr.2: 219535292-219535292 on Build GRCh38
rs781507161 Chr.2: 219535341-219535341 on Build GRCh38
rs907805496 Chr.2: 219535384-219535384 on Build GRCh38
rs760629513 Chr.2: 219535361-219535361 on Build GRCh38
rs748818908 Chr.2: 219535347-219535347 on Build GRCh38
rs375907275 Chr.2: 219535649-219535650 on Build GRCh38
rs79379124 Chr.2: 219535379-219535379 on Build GRCh38
rs761761089 Chr.2: 219535305-219535305 on Build GRCh38
rs147146087 Chr.2: 219535648-219535649 on Build GRCh38
rs575467827 Chr.2: 219535344-219535344 on Build GRCh38
rs144993868 Chr.2: 219535328-219535329 on Build GRCh38
rs920037382 Chr.2: 219535511-219535512 on Build GRCh38
rs758233507 Chr.2: 219535341-219535343 on Build GRCh38
rs769374214 Chr.2: 219535282-219535282 on Build GRCh38
rs947986721 Chr.2: 219535463-219535463 on Build GRCh38
rs749524367 Chr.2: 219535269-219535270 on Build GRCh38
rs778080295 Chr.2: 219535267-219535267 on Build GRCh38
rs771095872 Chr.2: 219535344-219535352 on Build GRCh38
rs775806097 Chr.2: 219535351-219535362 on Build GRCh38
rs763537185 Chr.2: 219535311-219535311 on Build GRCh38
rs903872881 Chr.2: 219535470-219535470 on Build GRCh38
rs919944218 Chr.2: 219535374-219535374 on Build GRCh38
rs747886920 Chr.2: 219535352-219535352 on Build GRCh38
rs553265071 Chr.2: 219535517-219535517 on Build GRCh38
rs140725401 Chr.2: 219535684-219535684 on Build GRCh38
rs1000784527 Chr.2: 219535489-219535489 on Build GRCh38
rs60623120 Chr.2: 219535342-219535342 on Build GRCh38
rs940414356 Chr.2: 219535401-219535401 on Build GRCh38
rs781671132 Chr.2: 219535272-219535272 on Build GRCh38
rs974139505 Chr.2: 219535507-219535510 on Build GRCh38
rs386655314 Chr.2: 219535340-219535342 on Build GRCh38
rs549025967 Chr.2: 219535474-219535474 on Build GRCh38
rs745307628 Chr.2: 219535281-219535281 on Build GRCh38
rs1020518384 Chr.2: 219535465-219535465 on Build GRCh38
rs529385503 Chr.2: 219535461-219535461 on Build GRCh38
rs548417454 Chr.2: 219535516-219535516 on Build GRCh38
rs756129176 Chr.2: 219535343-219535343 on Build GRCh38
rs746180761 Chr.2: 219535345-219535379 on Build GRCh38
rs111281614 Chr.2: 219535322-219535322 on Build GRCh38
rs544528470 Chr.2: 219535446-219535451 on Build GRCh38
rs772974923 Chr.2: 219535360-219535360 on Build GRCh38
rs753528619 Chr.2: 219535338-219535340 on Build GRCh38
rs950015953 Chr.2: 219535550-219535551 on Build GRCh38
rs866848955 Chr.2: 219535409-219535409 on Build GRCh38
rs767956288 Chr.2: 219535328-219535331 on Build GRCh38
rs984490636 Chr.2: 219535667-219535667 on Build GRCh38
rs757747405 Chr.2: 219535271-219535271 on Build GRCh38
rs776099576 Chr.2: 219535333-219535371 on Build GRCh38
rs749503740 Chr.2: 219535343-219535344 on Build GRCh38
rs371583964 Chr.2: 219535400-219535400 on Build GRCh38
rs759494381 Chr.2: 219535316-219535316 on Build GRCh38
rs991901557 Chr.2: 219535687-219535687 on Build GRCh38
rs774286941 Chr.2: 219535301-219535301 on Build GRCh38
rs774161844 Chr.2: 219535351-219535351 on Build GRCh38
rs564415781 Chr.2: 219535381-219535381 on Build GRCh38
rs552375695 Chr.2: 219535555-219535558 on Build GRCh38
rs766294390 Chr.2: 219535362-219535362 on Build GRCh38
rs768413143 Chr.2: 219535348-219535348 on Build GRCh38
rs1018859390 Chr.2: 219535499-219535499 on Build GRCh38
rs892820888 Chr.2: 219535432-219535439 on Build GRCh38
rs1020223525 Chr.2: 219535545-219535545 on Build GRCh38
rs756631747 Chr.2: 219535343-219535343 on Build GRCh38
rs774953081 Chr.2: 219535291-219535291 on Build GRCh38
rs771267404 Chr.2: 219535304-219535305 on Build GRCh38
rs905648343 Chr.2: 219535273-219535273 on Build GRCh38
rs560513582 Chr.2: 219535448-219535448 on Build GRCh38
rs77649813 Chr.2: 219535380-219535380 on Build GRCh38
rs1054194511 Chr.2: 219535639-219535639 on Build GRCh38
rs1056730369 Chr.2: 219535408-219535408 on Build GRCh38
rs572143533 Chr.2: 219535434-219535437 on Build GRCh38
rs754129154 Chr.2: 219535545-219535546 on Build GRCh38
rs779589710 Chr.2: 219535346-219535346 on Build GRCh38
rs1054100307 Chr.2: 219535428-219535428 on Build GRCh38
rs1045529199 Chr.2: 219535553-219535558 on Build GRCh38
rs72523909 Chr.2: 219535546-219535549 on Build GRCh38
rs751809155 Chr.2: 219535376-219535376 on Build GRCh38
rs1008296347 Chr.2: 219535521-219535521 on Build GRCh38
rs896302605 Chr.2: 219535459-219535459 on Build GRCh38
rs777351667 Chr.2: 219535558-219535559 on Build GRCh38
rs796187539 Chr.2: 219535340-219535342 on Build GRCh38
rs867150440 Chr.2: 219535339-219535340 on Build GRCh38
rs763714275 Chr.2: 219535335-219535371 on Build GRCh38
rs369741202 Chr.2: 219535441-219535444 on Build GRCh38
rs1001192475 Chr.2: 219535274-219535274 on Build GRCh38
rs772024765 Chr.2: 219535359-219535359 on Build GRCh38
rs542849490 Chr.2: 219535505-219535505 on Build GRCh38
rs767014390 Chr.2: 219535363-219535374 on Build GRCh38
rs376514033 Chr.2: 219535342-219535379 on Build GRCh38

View Full Product Details