Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 50780140-50780686 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATF1
Entrez Gene ID
466
Gene Name
activating transcription factor 1
Gene Aliases
EWS-ATF1, FUS/ATF-1, TREB36
UniGene
Hs.648565
Gene Chromosome Location
Chr.12: 50763682-50821162 on Build GRCh38
Transcripts
NM_005171.4 XM_011538386.2 XM_011538387.2 XM_011538388.2 XM_017019331.1 XM_017019332.1 XM_017019333.1 XM_017019334.1 XM_017019335.1 XM_017019336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4570635 c.2T>A p.M1K Chr.12: 50780147-50780147 on Build GRCh38
6606504 c.48T>G p.P16P Chr.12: 50780193-50780193 on Build GRCh38
5481060 c.54A>G p.S18S Chr.12: 50780199-50780199 on Build GRCh38
289563 c.88C>A p.Q30K Chr.12: 50780233-50780233 on Build GRCh38
3461829 c.65G>A p.G22E Chr.12: 50780210-50780210 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779950819 Chr.12: 50780232-50780232 on Build GRCh38
rs112410474 Chr.12: 50780653-50780653 on Build GRCh38
rs760384514 Chr.12: 50780172-50780172 on Build GRCh38
rs770276530 Chr.12: 50780185-50780185 on Build GRCh38
rs745886354 Chr.12: 50780251-50780251 on Build GRCh38
rs779392910 Chr.12: 50780243-50780243 on Build GRCh38
rs142947977 Chr.12: 50780652-50780653 on Build GRCh38
rs571683633 Chr.12: 50780460-50780460 on Build GRCh38
rs771849855 Chr.12: 50780379-50780379 on Build GRCh38
rs1038086519 Chr.12: 50780359-50780359 on Build GRCh38
rs138386001 Chr.12: 50780486-50780486 on Build GRCh38
rs1011919821 Chr.12: 50780341-50780349 on Build GRCh38
rs891473999 Chr.12: 50780622-50780622 on Build GRCh38
rs886673604 Chr.12: 50780517-50780517 on Build GRCh38
rs774206374 Chr.12: 50780320-50780320 on Build GRCh38
rs942371846 Chr.12: 50780314-50780314 on Build GRCh38
rs774198657 Chr.12: 50780208-50780209 on Build GRCh38
rs772833813 Chr.12: 50780168-50780168 on Build GRCh38
rs772360905 Chr.12: 50780263-50780263 on Build GRCh38
rs770636828 Chr.12: 50780288-50780288 on Build GRCh38
rs375093984 Chr.12: 50780193-50780193 on Build GRCh38
rs574489007 Chr.12: 50780362-50780362 on Build GRCh38
rs752721786 Chr.12: 50780202-50780202 on Build GRCh38
rs898280827 Chr.12: 50780485-50780485 on Build GRCh38
rs546059040 Chr.12: 50780449-50780449 on Build GRCh38
rs563040678 Chr.12: 50780646-50780646 on Build GRCh38
rs545001764 Chr.12: 50780285-50780285 on Build GRCh38
rs762836758 Chr.12: 50780205-50780205 on Build GRCh38
rs756391850 Chr.12: 50780228-50780228 on Build GRCh38
rs1021390721 Chr.12: 50780341-50780342 on Build GRCh38
rs540208072 Chr.12: 50780411-50780411 on Build GRCh38
rs201817307 Chr.12: 50780651-50780651 on Build GRCh38
rs371703036 Chr.12: 50780165-50780165 on Build GRCh38
rs181605386 Chr.12: 50780413-50780413 on Build GRCh38
rs560638582 Chr.12: 50780501-50780501 on Build GRCh38
rs1044153043 Chr.12: 50780206-50780206 on Build GRCh38
rs773031365 Chr.12: 50780400-50780400 on Build GRCh38
rs537986294 Chr.12: 50780302-50780302 on Build GRCh38
rs767709933 Chr.12: 50780651-50780652 on Build GRCh38
rs938437087 Chr.12: 50780576-50780576 on Build GRCh38
rs769945843 Chr.12: 50780301-50780301 on Build GRCh38
rs61735520 Chr.12: 50780190-50780190 on Build GRCh38
rs753935869 Chr.12: 50780233-50780233 on Build GRCh38
rs1050698904 Chr.12: 50780258-50780258 on Build GRCh38
rs928327801 Chr.12: 50780510-50780510 on Build GRCh38
rs933936833 Chr.12: 50780227-50780227 on Build GRCh38
rs10876088 Chr.12: 50780446-50780446 on Build GRCh38
rs778112866 Chr.12: 50780266-50780266 on Build GRCh38
rs375740066 Chr.12: 50780292-50780292 on Build GRCh38
rs750545129 Chr.12: 50780215-50780215 on Build GRCh38
rs115717108 Chr.12: 50780347-50780347 on Build GRCh38
rs868360734 Chr.12: 50780652-50780652 on Build GRCh38
rs141797982 Chr.12: 50780247-50780247 on Build GRCh38
rs763994606 Chr.12: 50780210-50780210 on Build GRCh38
rs947096006 Chr.12: 50780627-50780627 on Build GRCh38
rs10876089 Chr.12: 50780507-50780507 on Build GRCh38
rs11169556 Chr.12: 50780577-50780577 on Build GRCh38
rs1014491409 Chr.12: 50780518-50780518 on Build GRCh38
rs755094795 Chr.12: 50780235-50780235 on Build GRCh38

View Full Product Details