Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 51598954-51599481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP9A
Entrez Gene ID
10079
Gene Name
ATPase phospholipid transporting 9A (putative)
Gene Aliases
ATPIIA
UniGene
Hs.649234
Gene Chromosome Location
Chr.20: 51596775-51768411 on Build GRCh38
Transcripts
NM_006045.2 XM_011528481.2

SNP Detail

SNP ID Chromosome Location
rs528940544 Chr.20: 51599222-51599222 on Build GRCh38
rs890121780 Chr.20: 51599141-51599141 on Build GRCh38
rs1021110380 Chr.20: 51598981-51598981 on Build GRCh38
rs575808878 Chr.20: 51599150-51599150 on Build GRCh38
rs987472271 Chr.20: 51599416-51599416 on Build GRCh38
rs144778262 Chr.20: 51599149-51599149 on Build GRCh38
rs111560708 Chr.20: 51599356-51599356 on Build GRCh38
rs902284167 Chr.20: 51598997-51598997 on Build GRCh38
rs954200528 Chr.20: 51599411-51599411 on Build GRCh38
rs1003446904 Chr.20: 51599330-51599330 on Build GRCh38
rs1015656947 Chr.20: 51599305-51599305 on Build GRCh38
rs192389312 Chr.20: 51599076-51599076 on Build GRCh38
rs1023239853 Chr.20: 51599054-51599054 on Build GRCh38
rs148131170 Chr.20: 51599199-51599199 on Build GRCh38
rs112580990 Chr.20: 51599399-51599399 on Build GRCh38
rs529025900 Chr.20: 51599185-51599185 on Build GRCh38
rs1023207254 Chr.20: 51599266-51599266 on Build GRCh38
rs1050137954 Chr.20: 51599137-51599137 on Build GRCh38
rs545682728 Chr.20: 51599138-51599138 on Build GRCh38
rs959033872 Chr.20: 51599022-51599022 on Build GRCh38
rs961848206 Chr.20: 51599361-51599361 on Build GRCh38
rs777083133 Chr.20: 51599447-51599447 on Build GRCh38
rs773958951 Chr.20: 51599201-51599201 on Build GRCh38
rs891071853 Chr.20: 51599184-51599184 on Build GRCh38
rs961006141 Chr.20: 51599263-51599263 on Build GRCh38
rs932889933 Chr.20: 51599121-51599121 on Build GRCh38
rs1042262609 Chr.20: 51599192-51599192 on Build GRCh38
rs184611899 Chr.20: 51599176-51599176 on Build GRCh38
rs912789741 Chr.20: 51599420-51599420 on Build GRCh38
rs41274694 Chr.20: 51599015-51599015 on Build GRCh38
rs3827043 Chr.20: 51599281-51599281 on Build GRCh38
rs981659792 Chr.20: 51599084-51599084 on Build GRCh38
rs201103316 Chr.20: 51599087-51599087 on Build GRCh38
rs561717927 Chr.20: 51599151-51599151 on Build GRCh38
rs1002531446 Chr.20: 51599217-51599217 on Build GRCh38
rs971942134 Chr.20: 51599072-51599072 on Build GRCh38
rs770366212 Chr.20: 51599177-51599177 on Build GRCh38
rs543969982 Chr.20: 51599258-51599258 on Build GRCh38
rs985450352 Chr.20: 51599445-51599445 on Build GRCh38
rs147634257 Chr.20: 51599395-51599395 on Build GRCh38
rs187912145 Chr.20: 51599027-51599027 on Build GRCh38
rs954033200 Chr.20: 51599402-51599402 on Build GRCh38
rs941551320 Chr.20: 51599006-51599006 on Build GRCh38
rs1043569686 Chr.20: 51599198-51599198 on Build GRCh38
rs974247437 Chr.20: 51599387-51599387 on Build GRCh38
rs1052202062 Chr.20: 51599169-51599169 on Build GRCh38

View Full Product Details