Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 42112571-42113079 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEND4
Entrez Gene ID
389206
Gene Name
BEN domain containing 4
Gene Aliases
CCDC4
UniGene
Hs.120591
Gene Chromosome Location
Chr.4: 42110853-42152878 on Build GRCh38
Transcripts
NM_001159547.1 NM_207406.3 XM_017008185.1

SNP Detail

SNP ID Chromosome Location
rs770741940 Chr.4: 42112607-42112607 on Build GRCh38
rs981418906 Chr.4: 42112844-42112844 on Build GRCh38
rs1048614111 Chr.4: 42113011-42113011 on Build GRCh38
rs936163182 Chr.4: 42112925-42112925 on Build GRCh38
rs983675542 Chr.4: 42112735-42112735 on Build GRCh38
rs886873067 Chr.4: 42113016-42113016 on Build GRCh38
rs1016077716 Chr.4: 42112977-42112977 on Build GRCh38
rs764777912 Chr.4: 42112999-42112999 on Build GRCh38
rs755146612 Chr.4: 42112954-42112954 on Build GRCh38
rs973958727 Chr.4: 42112785-42112785 on Build GRCh38
rs761307251 Chr.4: 42112909-42112909 on Build GRCh38
rs1056311094 Chr.4: 42112861-42112861 on Build GRCh38
rs549280411 Chr.4: 42112885-42112885 on Build GRCh38
rs1015152874 Chr.4: 42112801-42112801 on Build GRCh38
rs747815085 Chr.4: 42112751-42112752 on Build GRCh38
rs768090262 Chr.4: 42112803-42112803 on Build GRCh38
rs566654265 Chr.4: 42112775-42112775 on Build GRCh38
rs529201030 Chr.4: 42112840-42112840 on Build GRCh38
rs937388306 Chr.4: 42112887-42112887 on Build GRCh38
rs557283518 Chr.4: 42112948-42112948 on Build GRCh38
rs974307715 Chr.4: 42113033-42113033 on Build GRCh38
rs891104271 Chr.4: 42112952-42112952 on Build GRCh38
rs571050276 Chr.4: 42112986-42112986 on Build GRCh38
rs76086470 Chr.4: 42112937-42112937 on Build GRCh38
rs908638713 Chr.4: 42112768-42112768 on Build GRCh38
rs1054662669 Chr.4: 42112926-42112926 on Build GRCh38
rs55811816 Chr.4: 42112704-42112704 on Build GRCh38
rs996841596 Chr.4: 42112612-42112612 on Build GRCh38
rs7677472 Chr.4: 42112654-42112654 on Build GRCh38
rs762568350 Chr.4: 42112757-42112757 on Build GRCh38
rs990633026 Chr.4: 42112888-42112888 on Build GRCh38
rs56050241 Chr.4: 42112927-42112927 on Build GRCh38
rs952302062 Chr.4: 42112710-42112710 on Build GRCh38
rs1042106333 Chr.4: 42112890-42112890 on Build GRCh38
rs926532303 Chr.4: 42112800-42112800 on Build GRCh38
rs914702256 Chr.4: 42112891-42112891 on Build GRCh38
rs995645069 Chr.4: 42112691-42112691 on Build GRCh38
rs745485341 Chr.4: 42112698-42112698 on Build GRCh38
rs937966640 Chr.4: 42112841-42112841 on Build GRCh38
rs140146503 Chr.4: 42112840-42112840 on Build GRCh38
rs945055091 Chr.4: 42112886-42112886 on Build GRCh38
rs542475048 Chr.4: 42112802-42112802 on Build GRCh38
rs533627181 Chr.4: 42113036-42113036 on Build GRCh38
rs535841233 Chr.4: 42112711-42112711 on Build GRCh38
rs971654799 Chr.4: 42112830-42112830 on Build GRCh38
rs945830638 Chr.4: 42112907-42112907 on Build GRCh38

View Full Product Details