Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7882394-7882942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S5-TXNDC5
Entrez Gene ID
100526836
Gene Name
BLOC1S5-TXNDC5 readthrough (NMD candidate)
Gene Aliases
MUTED-TXNDC5
UniGene
Hs.150837
Gene Chromosome Location
Chr.6: 7881250-8064414 on Build GRCh38
Transcripts
NR_037616.1 NM_001145549.2 NM_030810.3

Gene Symbol
TXNDC5
Entrez Gene ID
81567
Gene Name
thioredoxin domain containing 5
Gene Aliases
ENDOPDI, ERP46, HCC-2, HCC2, PDIA15, STRF8, UNQ364
UniGene
Hs.150837
Gene Chromosome Location
Chr.6: 7881250-7910814 on Build GRCh38
Transcripts
NR_037616.1 NM_001145549.2 NM_030810.3

SNP Detail

SNP ID Chromosome Location
rs552268749 Chr.6: 7882483-7882483 on Build GRCh38
rs961604844 Chr.6: 7882631-7882631 on Build GRCh38
rs1027980685 Chr.6: 7882889-7882889 on Build GRCh38
rs981019478 Chr.6: 7882903-7882903 on Build GRCh38
rs1026611226 Chr.6: 7882657-7882657 on Build GRCh38
rs559308544 Chr.6: 7882497-7882497 on Build GRCh38
rs1014564426 Chr.6: 7882867-7882867 on Build GRCh38
rs961940676 Chr.6: 7882609-7882609 on Build GRCh38
rs973272402 Chr.6: 7882615-7882615 on Build GRCh38
rs377347877 Chr.6: 7882778-7882778 on Build GRCh38
rs112273125 Chr.6: 7882841-7882841 on Build GRCh38
rs777615307 Chr.6: 7882482-7882482 on Build GRCh38
rs940564049 Chr.6: 7882426-7882426 on Build GRCh38
rs370179919 Chr.6: 7882519-7882519 on Build GRCh38
rs749993468 Chr.6: 7882559-7882560 on Build GRCh38
rs900468269 Chr.6: 7882603-7882603 on Build GRCh38
rs574919872 Chr.6: 7882749-7882749 on Build GRCh38
rs571120732 Chr.6: 7882663-7882663 on Build GRCh38
rs987535047 Chr.6: 7882593-7882593 on Build GRCh38
rs541940577 Chr.6: 7882764-7882764 on Build GRCh38
rs374697218 Chr.6: 7882499-7882499 on Build GRCh38
rs765594405 Chr.6: 7882797-7882797 on Build GRCh38
rs954442056 Chr.6: 7882590-7882590 on Build GRCh38
rs940757430 Chr.6: 7882873-7882873 on Build GRCh38
rs995402156 Chr.6: 7882643-7882643 on Build GRCh38
rs920390817 Chr.6: 7882470-7882470 on Build GRCh38
rs181032688 Chr.6: 7882489-7882489 on Build GRCh38
rs769254895 Chr.6: 7882656-7882656 on Build GRCh38
rs557351718 Chr.6: 7882796-7882796 on Build GRCh38
rs187157283 Chr.6: 7882710-7882710 on Build GRCh38
rs8643 Chr.6: 7882840-7882840 on Build GRCh38
rs1029142249 Chr.6: 7882569-7882570 on Build GRCh38
rs928387616 Chr.6: 7882502-7882502 on Build GRCh38
rs557736326 Chr.6: 7882478-7882478 on Build GRCh38
rs373039433 Chr.6: 7882526-7882526 on Build GRCh38
rs1053279997 Chr.6: 7882524-7882524 on Build GRCh38
rs568880503 Chr.6: 7882551-7882551 on Build GRCh38
rs995969908 Chr.6: 7882565-7882565 on Build GRCh38
rs896947709 Chr.6: 7882899-7882899 on Build GRCh38
rs938745541 Chr.6: 7882675-7882675 on Build GRCh38
rs904660066 Chr.6: 7882560-7882560 on Build GRCh38
rs913499602 Chr.6: 7882525-7882525 on Build GRCh38
rs1015405331 Chr.6: 7882629-7882629 on Build GRCh38
rs974165368 Chr.6: 7882485-7882485 on Build GRCh38
rs996746209 Chr.6: 7882827-7882827 on Build GRCh38
rs746980547 Chr.6: 7882521-7882521 on Build GRCh38
rs947638761 Chr.6: 7882538-7882538 on Build GRCh38
rs913369083 Chr.6: 7882715-7882715 on Build GRCh38
rs891776753 Chr.6: 7882757-7882757 on Build GRCh38
rs535238557 Chr.6: 7882664-7882664 on Build GRCh38
rs564845338 Chr.6: 7882890-7882890 on Build GRCh38
rs1051889194 Chr.6: 7882605-7882605 on Build GRCh38
rs995744940 Chr.6: 7882904-7882904 on Build GRCh38
rs928418489 Chr.6: 7882680-7882680 on Build GRCh38
rs927779758 Chr.6: 7882909-7882909 on Build GRCh38
rs904713500 Chr.6: 7882802-7882802 on Build GRCh38
rs978967941 Chr.6: 7882772-7882772 on Build GRCh38
rs536912595 Chr.6: 7882845-7882845 on Build GRCh38
rs1008506107 Chr.6: 7882611-7882611 on Build GRCh38
rs1043090483 Chr.6: 7882575-7882575 on Build GRCh38
rs979731897 Chr.6: 7882673-7882673 on Build GRCh38
rs955151401 Chr.6: 7882423-7882423 on Build GRCh38
rs11541846 Chr.6: 7882893-7882893 on Build GRCh38
rs1057414583 Chr.6: 7882522-7882522 on Build GRCh38
rs867612134 Chr.6: 7882654-7882654 on Build GRCh38
rs771714345 Chr.6: 7882544-7882544 on Build GRCh38
rs776463103 Chr.6: 7882711-7882711 on Build GRCh38
rs959800373 Chr.6: 7882704-7882704 on Build GRCh38
rs775084189 Chr.6: 7882608-7882608 on Build GRCh38
rs569529930 Chr.6: 7882447-7882447 on Build GRCh38
rs563783394 Chr.6: 7882879-7882880 on Build GRCh38
rs996263741 Chr.6: 7882604-7882604 on Build GRCh38
rs756028998 Chr.6: 7882481-7882481 on Build GRCh38
rs545755988 Chr.6: 7882862-7882862 on Build GRCh38
rs761331592 Chr.6: 7882634-7882634 on Build GRCh38

View Full Product Details