Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 3800619-3801111 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDC25B
Entrez Gene ID
994
Gene Name
cell division cycle 25B
Gene Aliases
-
UniGene
Hs.153752
Gene Chromosome Location
Chr.20: 3786772-3806121 on Build GRCh38
Transcripts
NM_001287516.1 NM_001287517.1 NM_001287518.1 NM_001287519.1 NM_001287520.1 NM_001287522.1 NM_001287524.1 NM_004358.4 NM_021872.3 NM_021873.3 NR_136335.1 NR_136336.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6624181 c.332G>A p.R111Q Chr.20: 3800807-3800807 on Build GRCh38
1307381 c.600G>A p.K200K Chr.20: 3800988-3800988 on Build GRCh38
5857855 c.508C>T p.Q170* Chr.20: 3800791-3800791 on Build GRCh38
7091132 c.408G>A p.K136K Chr.20: 3800988-3800988 on Build GRCh38
6300448 c.484G>C p.V162L Chr.20: 3800767-3800767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777091404 Chr.20: 3800706-3800707 on Build GRCh38
rs748880746 Chr.20: 3801037-3801037 on Build GRCh38
rs755182399 Chr.20: 3800853-3800853 on Build GRCh38
rs370909880 Chr.20: 3800958-3800958 on Build GRCh38
rs746114677 Chr.20: 3800699-3800699 on Build GRCh38
rs573830932 Chr.20: 3801077-3801077 on Build GRCh38
rs751526183 Chr.20: 3800840-3800840 on Build GRCh38
rs1028022871 Chr.20: 3800886-3800886 on Build GRCh38
rs549552103 Chr.20: 3800723-3800723 on Build GRCh38
rs765711564 Chr.20: 3800857-3800859 on Build GRCh38
rs939335268 Chr.20: 3800865-3800865 on Build GRCh38
rs769049251 Chr.20: 3800729-3800729 on Build GRCh38
rs759162155 Chr.20: 3800825-3800825 on Build GRCh38
rs904266009 Chr.20: 3800683-3800683 on Build GRCh38
rs371350767 Chr.20: 3800773-3800773 on Build GRCh38
rs780196030 Chr.20: 3800800-3800800 on Build GRCh38
rs997793868 Chr.20: 3800884-3800884 on Build GRCh38
rs373068861 Chr.20: 3800795-3800795 on Build GRCh38
rs767534464 Chr.20: 3801047-3801047 on Build GRCh38
rs920383476 Chr.20: 3800822-3800822 on Build GRCh38
rs773393529 Chr.20: 3800821-3800821 on Build GRCh38
rs764466123 Chr.20: 3801070-3801070 on Build GRCh38
rs368042199 Chr.20: 3801019-3801019 on Build GRCh38
rs11552106 Chr.20: 3800796-3800796 on Build GRCh38
rs894752608 Chr.20: 3800670-3800670 on Build GRCh38
rs757831012 Chr.20: 3801018-3801018 on Build GRCh38
rs781519537 Chr.20: 3801023-3801023 on Build GRCh38
rs776161634 Chr.20: 3800823-3800823 on Build GRCh38
rs868511318 Chr.20: 3800739-3800739 on Build GRCh38
rs749465321 Chr.20: 3800802-3800802 on Build GRCh38
rs758489417 Chr.20: 3800910-3800910 on Build GRCh38
rs754035549 Chr.20: 3800851-3800851 on Build GRCh38
rs758361574 Chr.20: 3800789-3800789 on Build GRCh38
rs199988612 Chr.20: 3800812-3800812 on Build GRCh38
rs761766290 Chr.20: 3800943-3800943 on Build GRCh38
rs1057375613 Chr.20: 3800663-3800663 on Build GRCh38
rs773681532 Chr.20: 3800809-3800809 on Build GRCh38
rs113619811 Chr.20: 3800867-3800867 on Build GRCh38
rs961178536 Chr.20: 3800970-3800970 on Build GRCh38
rs762236479 Chr.20: 3800748-3800748 on Build GRCh38
rs765024374 Chr.20: 3800776-3800776 on Build GRCh38
rs202155757 Chr.20: 3800975-3800975 on Build GRCh38
rs779089853 Chr.20: 3800868-3800868 on Build GRCh38
rs776157040 Chr.20: 3800909-3800909 on Build GRCh38
rs201911534 Chr.20: 3800770-3800770 on Build GRCh38
rs753214978 Chr.20: 3800884-3800884 on Build GRCh38
rs761494790 Chr.20: 3801046-3801046 on Build GRCh38
rs771606389 Chr.20: 3800765-3800765 on Build GRCh38
rs540104946 Chr.20: 3801025-3801025 on Build GRCh38
rs766394983 Chr.20: 3800953-3800953 on Build GRCh38
rs532926530 Chr.20: 3800681-3800681 on Build GRCh38
rs536795359 Chr.20: 3800842-3800842 on Build GRCh38
rs373883850 Chr.20: 3800883-3800883 on Build GRCh38
rs148574309 Chr.20: 3800715-3800726 on Build GRCh38
rs754985027 Chr.20: 3800965-3800965 on Build GRCh38
rs749680272 Chr.20: 3800724-3800724 on Build GRCh38
rs775293101 Chr.20: 3801035-3801035 on Build GRCh38
rs577638878 Chr.20: 3800831-3800831 on Build GRCh38
rs758579174 Chr.20: 3801065-3801065 on Build GRCh38
rs376072567 Chr.20: 3800817-3800817 on Build GRCh38
rs538158719 Chr.20: 3800774-3800774 on Build GRCh38
rs370507593 Chr.20: 3800873-3800873 on Build GRCh38
rs765244196 Chr.20: 3801056-3801056 on Build GRCh38
rs538614130 Chr.20: 3800839-3800839 on Build GRCh38
rs375465916 Chr.20: 3800767-3800767 on Build GRCh38
rs775276750 Chr.20: 3800914-3800914 on Build GRCh38
rs571020625 Chr.20: 3800781-3800781 on Build GRCh38
rs554011160 Chr.20: 3800933-3800933 on Build GRCh38
rs772664176 Chr.20: 3800766-3800766 on Build GRCh38
rs746300504 Chr.20: 3801085-3801085 on Build GRCh38
rs1011064003 Chr.20: 3800751-3800751 on Build GRCh38
rs142459784 Chr.20: 3801040-3801040 on Build GRCh38
rs111526979 Chr.20: 3800686-3800686 on Build GRCh38
rs748554755 Chr.20: 3800872-3800872 on Build GRCh38
rs984352715 Chr.20: 3800737-3800737 on Build GRCh38
rs200968688 Chr.20: 3801058-3801058 on Build GRCh38
rs1052778969 Chr.20: 3800836-3800836 on Build GRCh38
rs770189227 Chr.20: 3800702-3800702 on Build GRCh38
rs373954063 Chr.20: 3800740-3800740 on Build GRCh38
rs144359318 Chr.20: 3800807-3800807 on Build GRCh38
rs778311397 Chr.20: 3800885-3800885 on Build GRCh38
rs762895681 Chr.20: 3800922-3800922 on Build GRCh38
rs747493600 Chr.20: 3800887-3800887 on Build GRCh38
rs772425364 Chr.20: 3800882-3800882 on Build GRCh38
rs769525432 Chr.20: 3800913-3800913 on Build GRCh38
rs778045635 Chr.20: 3800994-3800994 on Build GRCh38
rs779300536 Chr.20: 3800806-3800806 on Build GRCh38
rs367730244 Chr.20: 3800908-3800908 on Build GRCh38
rs781751180 Chr.20: 3801078-3801078 on Build GRCh38
rs11570002 Chr.20: 3800687-3800687 on Build GRCh38
rs756159379 Chr.20: 3800841-3800841 on Build GRCh38
rs745347347 Chr.20: 3800911-3800911 on Build GRCh38
rs770463028 Chr.20: 3800818-3800818 on Build GRCh38
rs765071084 Chr.20: 3801007-3801007 on Build GRCh38
rs1013491251 Chr.20: 3800677-3800677 on Build GRCh38
rs367573455 Chr.20: 3800803-3800803 on Build GRCh38
rs752588927 Chr.20: 3800835-3800835 on Build GRCh38
rs759722205 Chr.20: 3801051-3801051 on Build GRCh38
rs1037966855 Chr.20: 3800673-3800673 on Build GRCh38
rs534156738 Chr.20: 3800692-3800692 on Build GRCh38
rs919329789 Chr.20: 3801017-3801017 on Build GRCh38
rs780110600 Chr.20: 3800846-3800846 on Build GRCh38
rs41279400 Chr.20: 3800691-3800691 on Build GRCh38
rs774430425 Chr.20: 3800939-3800939 on Build GRCh38
rs751994016 Chr.20: 3801071-3801071 on Build GRCh38
rs11570003 Chr.20: 3800722-3800733 on Build GRCh38
rs752711886 Chr.20: 3800786-3800786 on Build GRCh38
rs148769904 Chr.20: 3800969-3800969 on Build GRCh38
rs762808228 Chr.20: 3800838-3800838 on Build GRCh38
rs752923615 Chr.20: 3801059-3801059 on Build GRCh38

View Full Product Details