Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 1676374-1676897 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLGAP2
Entrez Gene ID
9228
Gene Name
DLG associated protein 2
Gene Aliases
DAP2, SAPAP2
UniGene
Hs.113287
Gene Chromosome Location
Chr.8: 1501366-1708476 on Build GRCh38
Transcripts
NM_001277161.1 NM_004745.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1552245 c.2071C>T p.R691* Chr.8: 1676617-1676617 on Build GRCh38
3646823 c.2032G>T p.E678* Chr.8: 1676578-1676578 on Build GRCh38
6644937 c.2130C>T p.R710R Chr.8: 1676568-1676568 on Build GRCh38
3646825 c.2046C>T p.V682V Chr.8: 1676592-1676592 on Build GRCh38
1097421 c.2170G>A p.D724N Chr.8: 1676608-1676608 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374871089 Chr.8: 1676547-1676547 on Build GRCh38
rs978825293 Chr.8: 1676401-1676401 on Build GRCh38
rs567157430 Chr.8: 1676541-1676541 on Build GRCh38
rs950328504 Chr.8: 1676405-1676405 on Build GRCh38
rs1020971009 Chr.8: 1676783-1676783 on Build GRCh38
rs557905499 Chr.8: 1676721-1676721 on Build GRCh38
rs931984294 Chr.8: 1676419-1676419 on Build GRCh38
rs752051468 Chr.8: 1676598-1676598 on Build GRCh38
rs765472321 Chr.8: 1676501-1676501 on Build GRCh38
rs764755107 Chr.8: 1676517-1676517 on Build GRCh38
rs372528299 Chr.8: 1676664-1676664 on Build GRCh38
rs750554089 Chr.8: 1676628-1676628 on Build GRCh38
rs115508527 Chr.8: 1676791-1676791 on Build GRCh38
rs564014359 Chr.8: 1676788-1676788 on Build GRCh38
rs555422589 Chr.8: 1676576-1676576 on Build GRCh38
rs73547759 Chr.8: 1676415-1676415 on Build GRCh38
rs940910705 Chr.8: 1676784-1676784 on Build GRCh38
rs758252726 Chr.8: 1676811-1676811 on Build GRCh38
rs973854776 Chr.8: 1676540-1676540 on Build GRCh38
rs987403087 Chr.8: 1676486-1676486 on Build GRCh38
rs775703416 Chr.8: 1676592-1676592 on Build GRCh38
rs776279542 Chr.8: 1676657-1676657 on Build GRCh38
rs543622259 Chr.8: 1676702-1676702 on Build GRCh38
rs1044837486 Chr.8: 1676736-1676736 on Build GRCh38
rs1046026354 Chr.8: 1676409-1676409 on Build GRCh38
rs371349976 Chr.8: 1676542-1676542 on Build GRCh38
rs769421865 Chr.8: 1676506-1676507 on Build GRCh38
rs377217252 Chr.8: 1676641-1676641 on Build GRCh38
rs954592036 Chr.8: 1676472-1676472 on Build GRCh38
rs79682431 Chr.8: 1676437-1676437 on Build GRCh38
rs749158458 Chr.8: 1676566-1676566 on Build GRCh38
rs749443269 Chr.8: 1676414-1676414 on Build GRCh38
rs372939448 Chr.8: 1676589-1676589 on Build GRCh38
rs922032049 Chr.8: 1676687-1676687 on Build GRCh38
rs904926499 Chr.8: 1676575-1676575 on Build GRCh38
rs994838440 Chr.8: 1676716-1676716 on Build GRCh38
rs560048519 Chr.8: 1676732-1676732 on Build GRCh38
rs914908637 Chr.8: 1676767-1676768 on Build GRCh38
rs551013637 Chr.8: 1676421-1676421 on Build GRCh38
rs200431414 Chr.8: 1676533-1676533 on Build GRCh38
rs987478041 Chr.8: 1676701-1676701 on Build GRCh38
rs191116349 Chr.8: 1676869-1676869 on Build GRCh38
rs767793627 Chr.8: 1676595-1676595 on Build GRCh38
rs898393571 Chr.8: 1676639-1676657 on Build GRCh38
rs750156612 Chr.8: 1676735-1676735 on Build GRCh38
rs763441363 Chr.8: 1676594-1676594 on Build GRCh38
rs550844699 Chr.8: 1676868-1676868 on Build GRCh38
rs759889581 Chr.8: 1676640-1676640 on Build GRCh38
rs936279335 Chr.8: 1676758-1676758 on Build GRCh38
rs770484892 Chr.8: 1676656-1676656 on Build GRCh38
rs987959525 Chr.8: 1676820-1676820 on Build GRCh38
rs772031461 Chr.8: 1676647-1676647 on Build GRCh38
rs756271753 Chr.8: 1676632-1676632 on Build GRCh38
rs1028096779 Chr.8: 1676719-1676719 on Build GRCh38
rs1010518098 Chr.8: 1676765-1676765 on Build GRCh38
rs750012072 Chr.8: 1676495-1676495 on Build GRCh38
rs570998898 Chr.8: 1676436-1676436 on Build GRCh38
rs368932884 Chr.8: 1676652-1676652 on Build GRCh38
rs767662562 Chr.8: 1676627-1676627 on Build GRCh38
rs760388653 Chr.8: 1676499-1676499 on Build GRCh38
rs753329856 Chr.8: 1676526-1676527 on Build GRCh38
rs757856736 Chr.8: 1676613-1676613 on Build GRCh38
rs529173105 Chr.8: 1676734-1676734 on Build GRCh38
rs746026598 Chr.8: 1676552-1676552 on Build GRCh38
rs748082745 Chr.8: 1676569-1676569 on Build GRCh38
rs779434573 Chr.8: 1676662-1676663 on Build GRCh38
rs778693739 Chr.8: 1676642-1676642 on Build GRCh38
rs752726801 Chr.8: 1676643-1676643 on Build GRCh38
rs780384355 Chr.8: 1676638-1676638 on Build GRCh38
rs141701208 Chr.8: 1676682-1676682 on Build GRCh38
rs758807381 Chr.8: 1676516-1676516 on Build GRCh38
rs908148974 Chr.8: 1676743-1676743 on Build GRCh38
rs145320852 Chr.8: 1676416-1676416 on Build GRCh38
rs755444890 Chr.8: 1676648-1676648 on Build GRCh38
rs139486417 Chr.8: 1676753-1676753 on Build GRCh38
rs867490767 Chr.8: 1676534-1676534 on Build GRCh38
rs994574630 Chr.8: 1676553-1676553 on Build GRCh38
rs904957512 Chr.8: 1676742-1676742 on Build GRCh38
rs751684188 Chr.8: 1676521-1676521 on Build GRCh38
rs940659012 Chr.8: 1676705-1676705 on Build GRCh38
rs940570129 Chr.8: 1676467-1676467 on Build GRCh38
rs767156984 Chr.8: 1676492-1676492 on Build GRCh38
rs750340739 Chr.8: 1676787-1676787 on Build GRCh38
rs375426065 Chr.8: 1676567-1676567 on Build GRCh38
rs890253849 Chr.8: 1676794-1676794 on Build GRCh38
rs372484845 Chr.8: 1676559-1676559 on Build GRCh38
rs369578407 Chr.8: 1676570-1676570 on Build GRCh38
rs575128513 Chr.8: 1676582-1676582 on Build GRCh38
rs1056170689 Chr.8: 1676750-1676750 on Build GRCh38
rs73672920 Chr.8: 1676748-1676748 on Build GRCh38
rs1036674723 Chr.8: 1676710-1676710 on Build GRCh38
rs890306301 Chr.8: 1676682-1676683 on Build GRCh38
rs150506542 Chr.8: 1676689-1676689 on Build GRCh38
rs187398862 Chr.8: 1676515-1676515 on Build GRCh38
rs774343202 Chr.8: 1676568-1676568 on Build GRCh38
rs1035062906 Chr.8: 1676605-1676605 on Build GRCh38
rs367964215 Chr.8: 1676558-1676558 on Build GRCh38
rs759119836 Chr.8: 1676661-1676661 on Build GRCh38
rs953476655 Chr.8: 1676812-1676812 on Build GRCh38

View Full Product Details