Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3652937-3653431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1
Entrez Gene ID
1773
Gene Name
deoxyribonuclease I
Gene Aliases
DNL1, DRNI
UniGene
Hs.629638
Gene Chromosome Location
Chr.16: 3611705-3665472 on Build GRCh38
Transcripts
NM_005223.3 XM_005255148.3 XM_006720854.3 XM_011522393.2 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022996.1 XM_017022997.1 XM_017022998.1 XM_017023001.1 XM_017023004.1 XM_017023005.1 XM_017023007.1

SNP Detail

SNP ID Chromosome Location
rs998157597 Chr.16: 3653190-3653190 on Build GRCh38
rs192125454 Chr.16: 3653253-3653253 on Build GRCh38
rs1016356455 Chr.16: 3653407-3653407 on Build GRCh38
rs1021769492 Chr.16: 3653231-3653231 on Build GRCh38
rs903397322 Chr.16: 3653394-3653394 on Build GRCh38
rs751909335 Chr.16: 3653211-3653211 on Build GRCh38
rs911639537 Chr.16: 3653315-3653315 on Build GRCh38
rs886652099 Chr.16: 3653355-3653355 on Build GRCh38
rs563439329 Chr.16: 3653071-3653071 on Build GRCh38
rs530042900 Chr.16: 3653226-3653226 on Build GRCh38
rs535357402 Chr.16: 3652963-3652963 on Build GRCh38
rs977455534 Chr.16: 3653034-3653034 on Build GRCh38
rs1043278005 Chr.16: 3653393-3653393 on Build GRCh38
rs554579561 Chr.16: 3652993-3652993 on Build GRCh38
rs182709234 Chr.16: 3653115-3653115 on Build GRCh38
rs191253928 Chr.16: 3653040-3653040 on Build GRCh38
rs559368881 Chr.16: 3653150-3653150 on Build GRCh38
rs1035730639 Chr.16: 3653252-3653252 on Build GRCh38
rs758859304 Chr.16: 3653002-3653002 on Build GRCh38
rs973292580 Chr.16: 3653284-3653284 on Build GRCh38
rs998977499 Chr.16: 3653408-3653408 on Build GRCh38
rs756777261 Chr.16: 3653174-3653174 on Build GRCh38
rs749702789 Chr.16: 3652980-3652980 on Build GRCh38
rs1007240691 Chr.16: 3653158-3653158 on Build GRCh38
rs1054205585 Chr.16: 3653152-3653152 on Build GRCh38
rs146695354 Chr.16: 3653143-3653143 on Build GRCh38
rs567204918 Chr.16: 3653362-3653362 on Build GRCh38
rs1054257072 Chr.16: 3653265-3653265 on Build GRCh38
rs139048434 Chr.16: 3653235-3653235 on Build GRCh38
rs563022240 Chr.16: 3653227-3653227 on Build GRCh38
rs1051491379 Chr.16: 3653198-3653198 on Build GRCh38
rs973653095 Chr.16: 3653157-3653157 on Build GRCh38
rs1039117110 Chr.16: 3653333-3653333 on Build GRCh38
rs984826857 Chr.16: 3652999-3652999 on Build GRCh38
rs567240803 Chr.16: 3653151-3653153 on Build GRCh38
rs892852470 Chr.16: 3653156-3653156 on Build GRCh38
rs568054081 Chr.16: 3653307-3653307 on Build GRCh38
rs560898746 Chr.16: 3653209-3653209 on Build GRCh38
rs74005841 Chr.16: 3653197-3653197 on Build GRCh38
rs752108172 Chr.16: 3653020-3653020 on Build GRCh38
rs960301544 Chr.16: 3653112-3653112 on Build GRCh38
rs892905034 Chr.16: 3653269-3653269 on Build GRCh38
rs900030411 Chr.16: 3653011-3653011 on Build GRCh38
rs903344727 Chr.16: 3653232-3653232 on Build GRCh38
rs1021707363 Chr.16: 3653015-3653015 on Build GRCh38
rs556759674 Chr.16: 3653409-3653409 on Build GRCh38
rs374703381 Chr.16: 3653158-3653160 on Build GRCh38
rs537092067 Chr.16: 3653331-3653331 on Build GRCh38
rs532583247 Chr.16: 3653250-3653250 on Build GRCh38
rs552478639 Chr.16: 3653125-3653125 on Build GRCh38
rs967450761 Chr.16: 3653033-3653033 on Build GRCh38
rs1011804680 Chr.16: 3653229-3653229 on Build GRCh38
rs950517248 Chr.16: 3653313-3653313 on Build GRCh38
rs901492694 Chr.16: 3653309-3653309 on Build GRCh38
rs528334818 Chr.16: 3653160-3653160 on Build GRCh38
rs919062915 Chr.16: 3653183-3653183 on Build GRCh38
rs34767593 Chr.16: 3653399-3653400 on Build GRCh38
rs890101517 Chr.16: 3652960-3652960 on Build GRCh38
rs1035678533 Chr.16: 3653080-3653080 on Build GRCh38
rs915835605 Chr.16: 3653077-3653078 on Build GRCh38
rs903004234 Chr.16: 3653142-3653142 on Build GRCh38
rs1008503455 Chr.16: 3652972-3652972 on Build GRCh38
rs902112405 Chr.16: 3653184-3653184 on Build GRCh38
rs914890970 Chr.16: 3653352-3653352 on Build GRCh38
rs143982736 Chr.16: 3653256-3653256 on Build GRCh38
rs987252225 Chr.16: 3653314-3653314 on Build GRCh38
rs1022931657 Chr.16: 3653272-3653272 on Build GRCh38
rs934486381 Chr.16: 3653259-3653259 on Build GRCh38
rs185414501 Chr.16: 3653014-3653014 on Build GRCh38
rs949977439 Chr.16: 3653089-3653089 on Build GRCh38
rs183334678 Chr.16: 3653304-3653304 on Build GRCh38
rs890153929 Chr.16: 3653204-3653204 on Build GRCh38
rs775799368 Chr.16: 3652996-3652996 on Build GRCh38
rs45564535 Chr.16: 3653249-3653249 on Build GRCh38
rs185968354 Chr.16: 3653144-3653144 on Build GRCh38
rs990792329 Chr.16: 3653244-3653245 on Build GRCh38
rs112878652 Chr.16: 3653275-3653275 on Build GRCh38
rs369554733 Chr.16: 3653154-3653154 on Build GRCh38

View Full Product Details