Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83166764-83167308 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DOPEY1
Entrez Gene ID
23033
Gene Name
dopey family member 1
Gene Aliases
DOP1, KIAA1117, dJ202D23.2
UniGene
Hs.520246
Gene Chromosome Location
Chr.6: 83067650-83171351 on Build GRCh38
Transcripts
NM_001199942.1 NM_015018.3 XM_011535619.2 XM_017010559.1 XM_017010560.1 XM_017010561.1 XM_017010562.1 XM_017010563.1 XM_017010564.1 XM_017010565.1 XM_017010566.1 XM_017010567.1 XM_017010568.1 XM_017010569.1 XM_017010570.1 XM_017010571.1 XM_017010572.1 NM_001199917.1 NM_001199918.1 NM_001199919.1 NM_015599.2 XM_017010935.1 XM_017010936.1 XM_017010937.1 XM_017010938.1

Gene Symbol
PGM3
Entrez Gene ID
5238
Gene Name
phosphoglucomutase 3
Gene Aliases
AGM1, IMD23, PAGM, PGM 3
UniGene
Hs.661665
Gene Chromosome Location
Chr.6: 83150728-83193936 on Build GRCh38
Transcripts
NM_001199942.1 NM_015018.3 XM_011535619.2 XM_017010559.1 XM_017010560.1 XM_017010561.1 XM_017010562.1 XM_017010563.1 XM_017010564.1 XM_017010565.1 XM_017010566.1 XM_017010567.1 XM_017010568.1 XM_017010569.1 XM_017010570.1 XM_017010571.1 XM_017010572.1 NM_001199917.1 NM_001199918.1 NM_001199919.1 NM_015599.2 XM_017010935.1 XM_017010936.1 XM_017010937.1 XM_017010938.1

SNP Detail

SNP ID Chromosome Location
rs969678269 Chr.6: 83167125-83167125 on Build GRCh38
rs996134186 Chr.6: 83167271-83167271 on Build GRCh38
rs551001119 Chr.6: 83167139-83167139 on Build GRCh38
rs558706567 Chr.6: 83166962-83166963 on Build GRCh38
rs963254193 Chr.6: 83166993-83166993 on Build GRCh38
rs917059876 Chr.6: 83166803-83166803 on Build GRCh38
rs899514371 Chr.6: 83166892-83166892 on Build GRCh38
rs890897875 Chr.6: 83166967-83166967 on Build GRCh38
rs992263452 Chr.6: 83166792-83166792 on Build GRCh38
rs1037568254 Chr.6: 83167242-83167242 on Build GRCh38
rs549011878 Chr.6: 83166995-83166995 on Build GRCh38
rs557782883 Chr.6: 83167122-83167122 on Build GRCh38
rs777881270 Chr.6: 83166818-83166818 on Build GRCh38
rs998408838 Chr.6: 83166818-83166818 on Build GRCh38
rs777771220 Chr.6: 83167048-83167048 on Build GRCh38
rs959415682 Chr.6: 83167036-83167036 on Build GRCh38
rs569824362 Chr.6: 83166869-83166869 on Build GRCh38
rs1054487973 Chr.6: 83166873-83166874 on Build GRCh38
rs1017028601 Chr.6: 83166977-83166977 on Build GRCh38
rs765369533 Chr.6: 83166969-83166969 on Build GRCh38
rs995087550 Chr.6: 83167025-83167025 on Build GRCh38
rs573502256 Chr.6: 83166911-83166911 on Build GRCh38
rs535447952 Chr.6: 83166956-83166956 on Build GRCh38
rs1026108309 Chr.6: 83167031-83167031 on Build GRCh38
rs935890605 Chr.6: 83167105-83167105 on Build GRCh38
rs916932490 Chr.6: 83167124-83167124 on Build GRCh38
rs1050756450 Chr.6: 83167155-83167155 on Build GRCh38
rs992189957 Chr.6: 83167037-83167040 on Build GRCh38
rs779807652 Chr.6: 83167002-83167002 on Build GRCh38
rs569311282 Chr.6: 83167183-83167183 on Build GRCh38
rs565617048 Chr.6: 83167040-83167040 on Build GRCh38
rs1007006858 Chr.6: 83166950-83166950 on Build GRCh38
rs1015748619 Chr.6: 83166990-83166990 on Build GRCh38
rs536853186 Chr.6: 83167252-83167252 on Build GRCh38
rs959915660 Chr.6: 83167280-83167280 on Build GRCh38
rs1034073589 Chr.6: 83167012-83167012 on Build GRCh38
rs989388062 Chr.6: 83167276-83167276 on Build GRCh38
rs903098592 Chr.6: 83166858-83166858 on Build GRCh38
rs1004335010 Chr.6: 83166968-83166968 on Build GRCh38
rs761754362 Chr.6: 83166829-83166829 on Build GRCh38
rs925695116 Chr.6: 83167211-83167211 on Build GRCh38
rs1054390053 Chr.6: 83167121-83167121 on Build GRCh38
rs45477797 Chr.6: 83167062-83167062 on Build GRCh38
rs892047649 Chr.6: 83166815-83166815 on Build GRCh38
rs1033924101 Chr.6: 83167274-83167275 on Build GRCh38
rs935969568 Chr.6: 83166867-83166867 on Build GRCh38
rs561460483 Chr.6: 83166901-83166901 on Build GRCh38
rs924479483 Chr.6: 83167093-83167093 on Build GRCh38
rs765371586 Chr.6: 83166876-83166876 on Build GRCh38
rs979859352 Chr.6: 83167185-83167185 on Build GRCh38
rs532589992 Chr.6: 83166817-83166817 on Build GRCh38
rs889761613 Chr.6: 83166926-83166926 on Build GRCh38
rs141200751 Chr.6: 83167215-83167215 on Build GRCh38
rs932299726 Chr.6: 83167123-83167123 on Build GRCh38
rs181180825 Chr.6: 83167052-83167052 on Build GRCh38

View Full Product Details