Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 74712877-74713401 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FA2H
Entrez Gene ID
79152
Gene Name
fatty acid 2-hydroxylase
Gene Aliases
FAAH, FAH1, FAXDC1, SCS7, SPG35
UniGene
Hs.461329
Gene Chromosome Location
Chr.16: 74712955-74774831 on Build GRCh38
Transcripts
NM_024306.4 XM_011523319.2

SNP Detail

SNP ID Chromosome Location
rs971625894 Chr.16: 74713080-74713080 on Build GRCh38
rs145615295 Chr.16: 74712931-74712931 on Build GRCh38
rs529743145 Chr.16: 74713305-74713305 on Build GRCh38
rs568964501 Chr.16: 74713377-74713377 on Build GRCh38
rs764667334 Chr.16: 74712997-74712997 on Build GRCh38
rs754403053 Chr.16: 74713002-74713002 on Build GRCh38
rs569182380 Chr.16: 74712966-74712966 on Build GRCh38
rs911147412 Chr.16: 74713158-74713158 on Build GRCh38
rs985284448 Chr.16: 74713154-74713154 on Build GRCh38
rs1036391048 Chr.16: 74713304-74713304 on Build GRCh38
rs962072121 Chr.16: 74713146-74713146 on Build GRCh38
rs1055932712 Chr.16: 74713266-74713266 on Build GRCh38
rs1001709575 Chr.16: 74713084-74713084 on Build GRCh38
rs1034013374 Chr.16: 74713098-74713098 on Build GRCh38
rs1056076259 Chr.16: 74713072-74713072 on Build GRCh38
rs369101225 Chr.16: 74713171-74713171 on Build GRCh38
rs141862106 Chr.16: 74713353-74713353 on Build GRCh38
rs537756842 Chr.16: 74713022-74713022 on Build GRCh38
rs952668796 Chr.16: 74713134-74713134 on Build GRCh38
rs985760745 Chr.16: 74712944-74712944 on Build GRCh38
rs543103467 Chr.16: 74713132-74713132 on Build GRCh38
rs962184603 Chr.16: 74713127-74713127 on Build GRCh38
rs896037051 Chr.16: 74713116-74713116 on Build GRCh38
rs1015023559 Chr.16: 74712912-74712912 on Build GRCh38
rs994822334 Chr.16: 74713352-74713352 on Build GRCh38
rs976428288 Chr.16: 74713178-74713178 on Build GRCh38
rs962081687 Chr.16: 74712917-74712917 on Build GRCh38
rs925702120 Chr.16: 74713227-74713227 on Build GRCh38
rs1046371 Chr.16: 74713126-74713126 on Build GRCh38
rs992799153 Chr.16: 74712902-74712902 on Build GRCh38
rs965229492 Chr.16: 74713168-74713168 on Build GRCh38
rs945846097 Chr.16: 74713203-74713203 on Build GRCh38
rs546605325 Chr.16: 74713323-74713323 on Build GRCh38
rs747070392 Chr.16: 74713330-74713330 on Build GRCh38
rs1018063118 Chr.16: 74713375-74713375 on Build GRCh38
rs891980427 Chr.16: 74713050-74713050 on Build GRCh38
rs755731623 Chr.16: 74712935-74712935 on Build GRCh38
rs182769519 Chr.16: 74713271-74713271 on Build GRCh38
rs747950593 Chr.16: 74713089-74713089 on Build GRCh38
rs1128935 Chr.16: 74712976-74712976 on Build GRCh38
rs1040656018 Chr.16: 74713000-74713000 on Build GRCh38
rs953456535 Chr.16: 74713361-74713361 on Build GRCh38
rs532244283 Chr.16: 74713360-74713360 on Build GRCh38
rs1054974994 Chr.16: 74713263-74713263 on Build GRCh38
rs1014506049 Chr.16: 74713275-74713275 on Build GRCh38
rs946084703 Chr.16: 74713279-74713279 on Build GRCh38
rs546416499 Chr.16: 74713231-74713231 on Build GRCh38
rs886052285 Chr.16: 74713303-74713303 on Build GRCh38
rs1014982032 Chr.16: 74713378-74713378 on Build GRCh38
rs1001818788 Chr.16: 74713071-74713071 on Build GRCh38
rs995316246 Chr.16: 74713128-74713128 on Build GRCh38
rs780315219 Chr.16: 74713316-74713316 on Build GRCh38
rs923057107 Chr.16: 74713186-74713186 on Build GRCh38
rs7189731 Chr.16: 74713059-74713059 on Build GRCh38
rs1040724613 Chr.16: 74712905-74712905 on Build GRCh38
rs911111476 Chr.16: 74712993-74712993 on Build GRCh38
rs576534015 Chr.16: 74713248-74713248 on Build GRCh38
rs886052284 Chr.16: 74713279-74713279 on Build GRCh38
rs562341602 Chr.16: 74713295-74713295 on Build GRCh38
rs912891901 Chr.16: 74713179-74713179 on Build GRCh38
rs573058036 Chr.16: 74713369-74713369 on Build GRCh38
rs1043180387 Chr.16: 74713218-74713218 on Build GRCh38
rs1045909184 Chr.16: 74713294-74713294 on Build GRCh38
rs115575599 Chr.16: 74713232-74713232 on Build GRCh38
rs533902177 Chr.16: 74713090-74713090 on Build GRCh38
rs148090175 Chr.16: 74713123-74713123 on Build GRCh38
rs931272931 Chr.16: 74712940-74712940 on Build GRCh38
rs553167415 Chr.16: 74713272-74713272 on Build GRCh38
rs7184172 Chr.16: 74713061-74713061 on Build GRCh38
rs4247497 Chr.16: 74712899-74712899 on Build GRCh38
rs951714742 Chr.16: 74713110-74713110 on Build GRCh38

View Full Product Details