Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151268493-151269041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GM2A
Entrez Gene ID
2760
Gene Name
GM2 ganglioside activator
Gene Aliases
GM2-AP, SAP-3
UniGene
Hs.483873
Gene Chromosome Location
Chr.5: 151253052-151270394 on Build GRCh38
Transcripts
NM_000405.4 NM_001167607.1

SNP Detail

SNP ID Chromosome Location
rs543560957 Chr.5: 151268527-151268527 on Build GRCh38
rs11262 Chr.5: 151268932-151268932 on Build GRCh38
rs886060273 Chr.5: 151268813-151268813 on Build GRCh38
rs185065661 Chr.5: 151269013-151269013 on Build GRCh38
rs995485542 Chr.5: 151268789-151268789 on Build GRCh38
rs911747681 Chr.5: 151268588-151268588 on Build GRCh38
rs913637180 Chr.5: 151268624-151268624 on Build GRCh38
rs968310896 Chr.5: 151268984-151268984 on Build GRCh38
rs760209124 Chr.5: 151268697-151268697 on Build GRCh38
rs201288211 Chr.5: 151268681-151268681 on Build GRCh38
rs1022511710 Chr.5: 151268949-151268949 on Build GRCh38
rs896687610 Chr.5: 151268749-151268749 on Build GRCh38
rs1052635263 Chr.5: 151268613-151268613 on Build GRCh38
rs980871303 Chr.5: 151268811-151268811 on Build GRCh38
rs992578142 Chr.5: 151268834-151268834 on Build GRCh38
rs966299714 Chr.5: 151268884-151268884 on Build GRCh38
rs153450 Chr.5: 151268896-151268896 on Build GRCh38
rs944552070 Chr.5: 151268607-151268607 on Build GRCh38
rs770712692 Chr.5: 151268831-151268831 on Build GRCh38
rs926791102 Chr.5: 151268818-151268818 on Build GRCh38
rs541345989 Chr.5: 151268763-151268763 on Build GRCh38
rs530626896 Chr.5: 151268925-151268925 on Build GRCh38
rs200856540 Chr.5: 151268722-151268722 on Build GRCh38
rs1002393492 Chr.5: 151268996-151268996 on Build GRCh38
rs552596465 Chr.5: 151268931-151268931 on Build GRCh38
rs562785409 Chr.5: 151268643-151268643 on Build GRCh38
rs940346735 Chr.5: 151268856-151268856 on Build GRCh38
rs954037942 Chr.5: 151268825-151268825 on Build GRCh38
rs879332294 Chr.5: 151268658-151268658 on Build GRCh38
rs181014044 Chr.5: 151268861-151268861 on Build GRCh38
rs1016123303 Chr.5: 151268765-151268765 on Build GRCh38
rs397716595 Chr.5: 151268725-151268726 on Build GRCh38
rs958473388 Chr.5: 151268829-151268829 on Build GRCh38
rs888931335 Chr.5: 151268902-151268902 on Build GRCh38
rs1056711686 Chr.5: 151268734-151268734 on Build GRCh38
rs986491729 Chr.5: 151268840-151268840 on Build GRCh38
rs532825835 Chr.5: 151268859-151268859 on Build GRCh38
rs1022520631 Chr.5: 151268704-151268704 on Build GRCh38
rs576574973 Chr.5: 151268519-151268519 on Build GRCh38
rs1009423626 Chr.5: 151268913-151268913 on Build GRCh38
rs1028246461 Chr.5: 151268808-151268808 on Build GRCh38
rs889236166 Chr.5: 151268556-151268556 on Build GRCh38
rs146782771 Chr.5: 151268853-151268853 on Build GRCh38
rs974031985 Chr.5: 151268520-151268520 on Build GRCh38
rs541818620 Chr.5: 151268692-151268692 on Build GRCh38
rs1009530448 Chr.5: 151268941-151268941 on Build GRCh38
rs1033838529 Chr.5: 151269017-151269017 on Build GRCh38
rs1036692772 Chr.5: 151268857-151268857 on Build GRCh38
rs397883774 Chr.5: 151268690-151268691 on Build GRCh38
rs1008978233 Chr.5: 151268557-151268557 on Build GRCh38
rs1029092038 Chr.5: 151268550-151268550 on Build GRCh38
rs916916097 Chr.5: 151268838-151268838 on Build GRCh38
rs962740601 Chr.5: 151268780-151268780 on Build GRCh38
rs201799049 Chr.5: 151268723-151268723 on Build GRCh38
rs1043423169 Chr.5: 151268930-151268930 on Build GRCh38

View Full Product Details