Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39332622-39333168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMFG
Entrez Gene ID
9535
Gene Name
glia maturation factor gamma
Gene Aliases
GMF-GAMMA
UniGene
Hs.5210
Gene Chromosome Location
Chr.19: 39328357-39336086 on Build GRCh38
Transcripts
NM_001301008.1 NM_004877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6587638 c.200G>C p.R67T Chr.19: 39333077-39333077 on Build GRCh38
4473441 c.184C>T p.P62S Chr.19: 39333093-39333093 on Build GRCh38
4533246 c.195G>A p.Q65Q Chr.19: 39333082-39333082 on Build GRCh38
3533686 c.165G>A p.E55E Chr.19: 39333112-39333112 on Build GRCh38
5933815 c.187G>A p.E63K Chr.19: 39333090-39333090 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs71856435 Chr.19: 39332657-39332658 on Build GRCh38
rs1035702843 Chr.19: 39332924-39332924 on Build GRCh38
rs888292000 Chr.19: 39332777-39332777 on Build GRCh38
rs768089333 Chr.19: 39333036-39333036 on Build GRCh38
rs767399179 Chr.19: 39332855-39332855 on Build GRCh38
rs888387282 Chr.19: 39333138-39333138 on Build GRCh38
rs376843898 Chr.19: 39333090-39333090 on Build GRCh38
rs768542738 Chr.19: 39332954-39332954 on Build GRCh38
rs993980543 Chr.19: 39332902-39332902 on Build GRCh38
rs1005416134 Chr.19: 39332801-39332801 on Build GRCh38
rs1056493497 Chr.19: 39332851-39332851 on Build GRCh38
rs985231926 Chr.19: 39333008-39333008 on Build GRCh38
rs747706667 Chr.19: 39333088-39333088 on Build GRCh38
rs781165561 Chr.19: 39332656-39332657 on Build GRCh38
rs75165044 Chr.19: 39332645-39332645 on Build GRCh38
rs76869068 Chr.19: 39332676-39332676 on Build GRCh38
rs915089903 Chr.19: 39332789-39332789 on Build GRCh38
rs932282367 Chr.19: 39333101-39333101 on Build GRCh38
rs749165340 Chr.19: 39332848-39332848 on Build GRCh38
rs765625547 Chr.19: 39333067-39333067 on Build GRCh38
rs1015416104 Chr.19: 39332847-39332847 on Build GRCh38
rs771805964 Chr.19: 39333112-39333113 on Build GRCh38
rs199816793 Chr.19: 39333110-39333110 on Build GRCh38
rs767566629 Chr.19: 39333091-39333091 on Build GRCh38
rs773879884 Chr.19: 39333041-39333041 on Build GRCh38
rs377027256 Chr.19: 39332734-39332734 on Build GRCh38
rs779866264 Chr.19: 39333114-39333114 on Build GRCh38
rs202194573 Chr.19: 39332656-39332656 on Build GRCh38
rs773985100 Chr.19: 39333010-39333010 on Build GRCh38
rs868470137 Chr.19: 39332950-39332950 on Build GRCh38
rs188870098 Chr.19: 39332769-39332769 on Build GRCh38
rs922246166 Chr.19: 39332680-39332680 on Build GRCh38
rs747604476 Chr.19: 39333046-39333046 on Build GRCh38
rs771437043 Chr.19: 39333048-39333048 on Build GRCh38
rs1029347471 Chr.19: 39332980-39332980 on Build GRCh38
rs567684996 Chr.19: 39332817-39332817 on Build GRCh38
rs777051753 Chr.19: 39333049-39333049 on Build GRCh38
rs751712734 Chr.19: 39333079-39333079 on Build GRCh38
rs780598066 Chr.19: 39333046-39333047 on Build GRCh38
rs953812567 Chr.19: 39332996-39332996 on Build GRCh38
rs533301042 Chr.19: 39332944-39332944 on Build GRCh38
rs755909808 Chr.19: 39333096-39333096 on Build GRCh38
rs551580183 Chr.19: 39333056-39333056 on Build GRCh38
rs900803067 Chr.19: 39332650-39332650 on Build GRCh38
rs1023203370 Chr.19: 39333014-39333014 on Build GRCh38
rs967784605 Chr.19: 39333039-39333039 on Build GRCh38
rs948176838 Chr.19: 39332863-39332863 on Build GRCh38
rs748946918 Chr.19: 39333031-39333031 on Build GRCh38
rs1049390911 Chr.19: 39333125-39333125 on Build GRCh38
rs78456185 Chr.19: 39332677-39332677 on Build GRCh38
rs369173001 Chr.19: 39333071-39333071 on Build GRCh38
rs953840852 Chr.19: 39332721-39332721 on Build GRCh38
rs59277772 Chr.19: 39332676-39332677 on Build GRCh38
rs968925437 Chr.19: 39333019-39333019 on Build GRCh38
rs985587586 Chr.19: 39332761-39332761 on Build GRCh38
rs1015038483 Chr.19: 39332942-39332942 on Build GRCh38
rs150080208 Chr.19: 39333092-39333092 on Build GRCh38
rs796794840 Chr.19: 39332820-39332820 on Build GRCh38
rs941254073 Chr.19: 39333143-39333143 on Build GRCh38
rs748994750 Chr.19: 39333135-39333135 on Build GRCh38
rs544288960 Chr.19: 39332755-39332755 on Build GRCh38
rs924774986 Chr.19: 39333044-39333044 on Build GRCh38
rs764372926 Chr.19: 39333078-39333078 on Build GRCh38
rs557951854 Chr.19: 39332819-39332820 on Build GRCh38

View Full Product Details