Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 70817395-70817944 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRSF1
Entrez Gene ID
2926
Gene Name
G-rich RNA sequence binding factor 1
Gene Aliases
-
UniGene
Hs.309763
Gene Chromosome Location
Chr.4: 70815782-70843274 on Build GRCh38
Transcripts
NM_001098477.1 NM_002092.3

SNP Detail

SNP ID Chromosome Location
rs1055660427 Chr.4: 70817579-70817579 on Build GRCh38
rs144576218 Chr.4: 70817536-70817536 on Build GRCh38
rs971356747 Chr.4: 70817647-70817647 on Build GRCh38
rs982402094 Chr.4: 70817649-70817649 on Build GRCh38
rs938052210 Chr.4: 70817430-70817430 on Build GRCh38
rs867907039 Chr.4: 70817563-70817563 on Build GRCh38
rs748183285 Chr.4: 70817852-70817856 on Build GRCh38
rs757050324 Chr.4: 70817562-70817567 on Build GRCh38
rs1053872534 Chr.4: 70817463-70817463 on Build GRCh38
rs549399302 Chr.4: 70817695-70817695 on Build GRCh38
rs983080026 Chr.4: 70817838-70817838 on Build GRCh38
rs896885956 Chr.4: 70817911-70817911 on Build GRCh38
rs572471287 Chr.4: 70817539-70817539 on Build GRCh38
rs922916274 Chr.4: 70817481-70817481 on Build GRCh38
rs1049839458 Chr.4: 70817523-70817523 on Build GRCh38
rs999454587 Chr.4: 70817685-70817685 on Build GRCh38
rs189027080 Chr.4: 70817511-70817511 on Build GRCh38
rs949558434 Chr.4: 70817908-70817908 on Build GRCh38
rs976384930 Chr.4: 70817782-70817782 on Build GRCh38
rs763223842 Chr.4: 70817610-70817610 on Build GRCh38
rs895436013 Chr.4: 70817608-70817608 on Build GRCh38
rs762231479 Chr.4: 70817557-70817557 on Build GRCh38
rs765235374 Chr.4: 70817761-70817762 on Build GRCh38
rs780714753 Chr.4: 70817920-70817920 on Build GRCh38
rs533177310 Chr.4: 70817604-70817604 on Build GRCh38
rs367789706 Chr.4: 70817723-70817724 on Build GRCh38
rs565076513 Chr.4: 70817703-70817703 on Build GRCh38
rs557412391 Chr.4: 70817895-70817895 on Build GRCh38
rs945504061 Chr.4: 70817766-70817766 on Build GRCh38
rs113120280 Chr.4: 70817542-70817542 on Build GRCh38
rs1046418134 Chr.4: 70817629-70817629 on Build GRCh38
rs1030223822 Chr.4: 70817785-70817785 on Build GRCh38
rs543435278 Chr.4: 70817471-70817471 on Build GRCh38
rs929063478 Chr.4: 70817483-70817483 on Build GRCh38
rs79923263 Chr.4: 70817615-70817615 on Build GRCh38
rs902542043 Chr.4: 70817650-70817650 on Build GRCh38
rs537048213 Chr.4: 70817689-70817689 on Build GRCh38
rs576125431 Chr.4: 70817912-70817912 on Build GRCh38
rs893607453 Chr.4: 70817486-70817486 on Build GRCh38
rs766968335 Chr.4: 70817661-70817661 on Build GRCh38
rs554192822 Chr.4: 70817512-70817512 on Build GRCh38
rs926621455 Chr.4: 70817427-70817427 on Build GRCh38
rs758433475 Chr.4: 70817841-70817842 on Build GRCh38
rs961152920 Chr.4: 70817552-70817552 on Build GRCh38
rs901066099 Chr.4: 70817522-70817522 on Build GRCh38
rs975807063 Chr.4: 70817464-70817464 on Build GRCh38
rs116536521 Chr.4: 70817888-70817888 on Build GRCh38
rs915325064 Chr.4: 70817734-70817734 on Build GRCh38
rs181298345 Chr.4: 70817749-70817749 on Build GRCh38
rs908962720 Chr.4: 70817862-70817862 on Build GRCh38
rs567372293 Chr.4: 70817842-70817842 on Build GRCh38
rs945195638 Chr.4: 70817503-70817503 on Build GRCh38
rs754114404 Chr.4: 70817482-70817482 on Build GRCh38
rs553276352 Chr.4: 70817762-70817762 on Build GRCh38
rs561823812 Chr.4: 70817522-70817522 on Build GRCh38
rs148480711 Chr.4: 70817571-70817571 on Build GRCh38
rs1026513930 Chr.4: 70817632-70817632 on Build GRCh38
rs950373874 Chr.4: 70817788-70817788 on Build GRCh38
rs982509386 Chr.4: 70817425-70817425 on Build GRCh38
rs575619278 Chr.4: 70817454-70817454 on Build GRCh38
rs1005225818 Chr.4: 70817537-70817537 on Build GRCh38
rs1032398067 Chr.4: 70817687-70817687 on Build GRCh38
rs577784484 Chr.4: 70817786-70817786 on Build GRCh38
rs567498506 Chr.4: 70817698-70817698 on Build GRCh38
rs889735090 Chr.4: 70817875-70817875 on Build GRCh38
rs570190570 Chr.4: 70817623-70817623 on Build GRCh38
rs1017847252 Chr.4: 70817750-70817750 on Build GRCh38
rs535277894 Chr.4: 70817760-70817760 on Build GRCh38
rs763718819 Chr.4: 70817473-70817476 on Build GRCh38
rs758795114 Chr.4: 70817906-70817906 on Build GRCh38
rs991034811 Chr.4: 70817903-70817903 on Build GRCh38

View Full Product Details