Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 72888013-72888602 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GXYLT2
Entrez Gene ID
727936
Gene Name
glucoside xylosyltransferase 2
Gene Aliases
GLT8D4
UniGene
Hs.710275
Gene Chromosome Location
Chr.3: 72848522-72975374 on Build GRCh38
Transcripts
NM_001080393.1 XM_011534067.2 XM_011534068.1 XM_017007123.1 XM_017007124.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7429673 c.214C>A p.P72T Chr.3: 72888447-72888447 on Build GRCh38
5744783 c.214C>T p.P72S Chr.3: 72888447-72888447 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs529400999 Chr.3: 72888447-72888447 on Build GRCh38
rs960217789 Chr.3: 72888259-72888259 on Build GRCh38
rs926204165 Chr.3: 72888446-72888446 on Build GRCh38
rs927412212 Chr.3: 72888120-72888121 on Build GRCh38
rs945731190 Chr.3: 72888151-72888151 on Build GRCh38
rs980217305 Chr.3: 72888234-72888234 on Build GRCh38
rs993423692 Chr.3: 72888377-72888377 on Build GRCh38
rs989442622 Chr.3: 72888290-72888311 on Build GRCh38
rs930470254 Chr.3: 72888527-72888527 on Build GRCh38
rs978834366 Chr.3: 72888279-72888279 on Build GRCh38
rs1008733191 Chr.3: 72888072-72888072 on Build GRCh38
rs1028758407 Chr.3: 72888055-72888055 on Build GRCh38
rs992997204 Chr.3: 72888262-72888262 on Build GRCh38
rs953079287 Chr.3: 72888448-72888448 on Build GRCh38
rs1008714710 Chr.3: 72888205-72888205 on Build GRCh38
rs748906404 Chr.3: 72888518-72888518 on Build GRCh38
rs146974723 Chr.3: 72888548-72888548 on Build GRCh38
rs890320265 Chr.3: 72888193-72888193 on Build GRCh38
rs565548280 Chr.3: 72888497-72888497 on Build GRCh38
rs925586153 Chr.3: 72888281-72888281 on Build GRCh38
rs868115901 Chr.3: 72888041-72888041 on Build GRCh38
rs551431157 Chr.3: 72888278-72888278 on Build GRCh38
rs1034462309 Chr.3: 72888362-72888362 on Build GRCh38
rs1001516087 Chr.3: 72888358-72888358 on Build GRCh38
rs138863184 Chr.3: 72888069-72888069 on Build GRCh38
rs972747641 Chr.3: 72888081-72888081 on Build GRCh38
rs1007285679 Chr.3: 72888579-72888579 on Build GRCh38
rs566749857 Chr.3: 72888305-72888305 on Build GRCh38
rs913486589 Chr.3: 72888275-72888275 on Build GRCh38
rs994596477 Chr.3: 72888217-72888217 on Build GRCh38
rs897741867 Chr.3: 72888330-72888330 on Build GRCh38
rs564923305 Chr.3: 72888064-72888065 on Build GRCh38
rs904258754 Chr.3: 72888042-72888042 on Build GRCh38
rs1034432949 Chr.3: 72888235-72888235 on Build GRCh38
rs931691190 Chr.3: 72888184-72888184 on Build GRCh38
rs944434283 Chr.3: 72888490-72888490 on Build GRCh38
rs985733281 Chr.3: 72888461-72888461 on Build GRCh38
rs890192550 Chr.3: 72888308-72888308 on Build GRCh38
rs913465534 Chr.3: 72888144-72888144 on Build GRCh38
rs993524924 Chr.3: 72888128-72888128 on Build GRCh38
rs938825908 Chr.3: 72888121-72888121 on Build GRCh38
rs911663362 Chr.3: 72888474-72888474 on Build GRCh38
rs1014834371 Chr.3: 72888075-72888075 on Build GRCh38
rs189757959 Chr.3: 72888092-72888092 on Build GRCh38
rs888488893 Chr.3: 72888352-72888352 on Build GRCh38
rs555295462 Chr.3: 72888399-72888399 on Build GRCh38
rs995979124 Chr.3: 72888051-72888052 on Build GRCh38
rs1036612362 Chr.3: 72888506-72888506 on Build GRCh38
rs897715287 Chr.3: 72888211-72888211 on Build GRCh38
rs890213367 Chr.3: 72888567-72888567 on Build GRCh38
rs952781205 Chr.3: 72888230-72888230 on Build GRCh38
rs1015286524 Chr.3: 72888208-72888209 on Build GRCh38
rs745383897 Chr.3: 72888319-72888319 on Build GRCh38
rs1042700484 Chr.3: 72888157-72888157 on Build GRCh38
rs750468771 Chr.3: 72888570-72888570 on Build GRCh38
rs920020182 Chr.3: 72888088-72888088 on Build GRCh38
rs765424859 Chr.3: 72888532-72888532 on Build GRCh38
rs994729885 Chr.3: 72888348-72888348 on Build GRCh38
rs905096888 Chr.3: 72888542-72888542 on Build GRCh38
rs961700590 Chr.3: 72888079-72888088 on Build GRCh38
rs944552494 Chr.3: 72888318-72888318 on Build GRCh38
rs980268130 Chr.3: 72888105-72888105 on Build GRCh38
rs113295663 Chr.3: 72888148-72888148 on Build GRCh38
rs952751685 Chr.3: 72888094-72888094 on Build GRCh38
rs1020409025 Chr.3: 72888379-72888379 on Build GRCh38
rs533124440 Chr.3: 72888191-72888191 on Build GRCh38
rs979068907 Chr.3: 72888417-72888417 on Build GRCh38
rs892344228 Chr.3: 72888535-72888535 on Build GRCh38
rs904297353 Chr.3: 72888182-72888182 on Build GRCh38
rs1027565271 Chr.3: 72888350-72888350 on Build GRCh38
rs779750574 Chr.3: 72888493-72888493 on Build GRCh38
rs182416999 Chr.3: 72888374-72888374 on Build GRCh38
rs919131793 Chr.3: 72888526-72888526 on Build GRCh38
rs1009384985 Chr.3: 72888546-72888546 on Build GRCh38

View Full Product Details