Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44842819-44843368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
H2AFV
Entrez Gene ID
94239
Gene Name
H2A histone family member V
Gene Aliases
H2A.Z-2, H2AV
UniGene
Hs.488189
Gene Chromosome Location
Chr.7: 44826889-44848126 on Build GRCh38
Transcripts
NM_012412.4 NM_138635.3 NM_201436.2 NM_201516.2 NM_201517.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
333481 c.36G>A p.K12K Chr.7: 44843322-44843322 on Build GRCh38
4689411 c.19G>T p.G7* Chr.7: 44843339-44843339 on Build GRCh38
5884555 c.5C>T p.A2V Chr.7: 44843353-44843353 on Build GRCh38
1450961 c.42G>A p.K14K Chr.7: 44843316-44843316 on Build GRCh38
3881299 c.39C>T p.A13A Chr.7: 44843319-44843319 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027258685 Chr.7: 44843181-44843181 on Build GRCh38
rs749690202 Chr.7: 44843340-44843340 on Build GRCh38
rs781235556 Chr.7: 44842844-44842844 on Build GRCh38
rs965663130 Chr.7: 44843183-44843184 on Build GRCh38
rs749920657 Chr.7: 44843018-44843018 on Build GRCh38
rs945311498 Chr.7: 44843053-44843053 on Build GRCh38
rs185405989 Chr.7: 44842999-44842999 on Build GRCh38
rs779291349 Chr.7: 44843343-44843343 on Build GRCh38
rs778327341 Chr.7: 44843331-44843331 on Build GRCh38
rs932353212 Chr.7: 44843087-44843087 on Build GRCh38
rs996051699 Chr.7: 44843175-44843175 on Build GRCh38
rs191415881 Chr.7: 44843217-44843217 on Build GRCh38
rs896998491 Chr.7: 44843135-44843135 on Build GRCh38
rs913738648 Chr.7: 44842917-44842917 on Build GRCh38
rs567731448 Chr.7: 44842957-44842957 on Build GRCh38
rs762026910 Chr.7: 44842853-44842853 on Build GRCh38
rs554473164 Chr.7: 44843125-44843125 on Build GRCh38
rs897309782 Chr.7: 44843170-44843170 on Build GRCh38
rs1043635856 Chr.7: 44843068-44843068 on Build GRCh38
rs751593237 Chr.7: 44843270-44843270 on Build GRCh38
rs938715119 Chr.7: 44843012-44843012 on Build GRCh38
rs367901145 Chr.7: 44843023-44843023 on Build GRCh38
rs775158767 Chr.7: 44843147-44843157 on Build GRCh38
rs756512163 Chr.7: 44843315-44843315 on Build GRCh38
rs750339562 Chr.7: 44843265-44843265 on Build GRCh38
rs974347937 Chr.7: 44842967-44842967 on Build GRCh38
rs540419981 Chr.7: 44843154-44843181 on Build GRCh38
rs55941046 Chr.7: 44843157-44843175 on Build GRCh38
rs890851978 Chr.7: 44842868-44842868 on Build GRCh38
rs765057806 Chr.7: 44843262-44843262 on Build GRCh38
rs371956322 Chr.7: 44843258-44843258 on Build GRCh38
rs761581898 Chr.7: 44843260-44843260 on Build GRCh38
rs898668828 Chr.7: 44843047-44843047 on Build GRCh38
rs775459818 Chr.7: 44843007-44843007 on Build GRCh38
rs760132152 Chr.7: 44843229-44843229 on Build GRCh38
rs768286930 Chr.7: 44843230-44843230 on Build GRCh38
rs1035685653 Chr.7: 44843255-44843255 on Build GRCh38
rs940294551 Chr.7: 44843011-44843011 on Build GRCh38
rs1007288175 Chr.7: 44843105-44843105 on Build GRCh38
rs147999975 Chr.7: 44843300-44843300 on Build GRCh38
rs188056740 Chr.7: 44843124-44843124 on Build GRCh38
rs967990333 Chr.7: 44843259-44843259 on Build GRCh38
rs571824851 Chr.7: 44843097-44843097 on Build GRCh38
rs537294145 Chr.7: 44842993-44842993 on Build GRCh38
rs1037489122 Chr.7: 44843115-44843115 on Build GRCh38
rs754804881 Chr.7: 44843138-44843157 on Build GRCh38
rs1037199383 Chr.7: 44843139-44843139 on Build GRCh38
rs1008353775 Chr.7: 44842869-44842869 on Build GRCh38
rs901082599 Chr.7: 44843156-44843181 on Build GRCh38
rs1007923084 Chr.7: 44843116-44843116 on Build GRCh38
rs996111561 Chr.7: 44843057-44843057 on Build GRCh38
rs374028878 Chr.7: 44842925-44842925 on Build GRCh38
rs781554792 Chr.7: 44843290-44843290 on Build GRCh38
rs368107823 Chr.7: 44843231-44843231 on Build GRCh38
rs997027298 Chr.7: 44842865-44842865 on Build GRCh38
rs888885195 Chr.7: 44843093-44843093 on Build GRCh38
rs749062057 Chr.7: 44843092-44843092 on Build GRCh38
rs913735744 Chr.7: 44843030-44843030 on Build GRCh38
rs766771912 Chr.7: 44842867-44842867 on Build GRCh38
rs757740920 Chr.7: 44843341-44843341 on Build GRCh38
rs750736632 Chr.7: 44842866-44842866 on Build GRCh38
rs118118848 Chr.7: 44843202-44843202 on Build GRCh38
rs745866459 Chr.7: 44842934-44842934 on Build GRCh38
rs755030031 Chr.7: 44843275-44843275 on Build GRCh38
rs980489419 Chr.7: 44843004-44843004 on Build GRCh38
rs979283193 Chr.7: 44842941-44842941 on Build GRCh38
rs778257383 Chr.7: 44843244-44843246 on Build GRCh38
rs925163192 Chr.7: 44843080-44843080 on Build GRCh38
rs913766975 Chr.7: 44843321-44843321 on Build GRCh38
rs926465930 Chr.7: 44843299-44843299 on Build GRCh38
rs961637659 Chr.7: 44842935-44842935 on Build GRCh38
rs767851241 Chr.7: 44842859-44842859 on Build GRCh38
rs988034067 Chr.7: 44842876-44842876 on Build GRCh38

View Full Product Details