Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 149810563-149811110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST2H2BF
Entrez Gene ID
440689
Gene Name
histone cluster 2, H2bf
Gene Aliases
-
UniGene
Hs.632451
Gene Chromosome Location
Chr.1: 149782689-149812373 on Build GRCh38
Transcripts
NM_001161334.1

SNP Detail

SNP ID Chromosome Location
rs147523682 Chr.1: 149810981-149810981 on Build GRCh38
rs370468189 Chr.1: 149810874-149810874 on Build GRCh38
rs184494504 Chr.1: 149810929-149810929 on Build GRCh38
rs193023699 Chr.1: 149810915-149810915 on Build GRCh38
rs10888479 Chr.1: 149810854-149810854 on Build GRCh38
rs587666340 Chr.1: 149810928-149810928 on Build GRCh38
rs142701235 Chr.1: 149810779-149810779 on Build GRCh38
rs3005609 Chr.1: 149810878-149810878 on Build GRCh38
rs6698340 Chr.1: 149810898-149810898 on Build GRCh38
rs587750531 Chr.1: 149810977-149810977 on Build GRCh38
rs587676437 Chr.1: 149810609-149810609 on Build GRCh38
rs587739353 Chr.1: 149810652-149810652 on Build GRCh38
rs2457471 Chr.1: 149810749-149810749 on Build GRCh38
rs187588166 Chr.1: 149811010-149811010 on Build GRCh38
rs782749340 Chr.1: 149810960-149810960 on Build GRCh38
rs629346 Chr.1: 149810654-149810654 on Build GRCh38
rs2791058 Chr.1: 149811085-149811085 on Build GRCh38

View Full Product Details