Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30460062-30460595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMGB1
Entrez Gene ID
3146
Gene Name
high mobility group box 1
Gene Aliases
HMG-1, HMG1, HMG3, SBP-1
UniGene
Hs.434102
Gene Chromosome Location
Chr.13: 30457916-30617597 on Build GRCh38
Transcripts
NM_001313892.1 NM_001313893.1 NM_002128.5

SNP Detail

SNP ID Chromosome Location
rs980623192 Chr.13: 30460258-30460258 on Build GRCh38
rs933568072 Chr.13: 30460566-30460566 on Build GRCh38
rs970557322 Chr.13: 30460117-30460120 on Build GRCh38
rs961983598 Chr.13: 30460204-30460204 on Build GRCh38
rs557515158 Chr.13: 30460181-30460183 on Build GRCh38
rs539371459 Chr.13: 30460170-30460170 on Build GRCh38
rs749264897 Chr.13: 30460110-30460110 on Build GRCh38
rs572869891 Chr.13: 30460480-30460480 on Build GRCh38
rs958764787 Chr.13: 30460274-30460274 on Build GRCh38
rs967362184 Chr.13: 30460526-30460526 on Build GRCh38
rs570096954 Chr.13: 30460185-30460192 on Build GRCh38
rs927367449 Chr.13: 30460133-30460133 on Build GRCh38
rs1025346318 Chr.13: 30460465-30460465 on Build GRCh38
rs4097504 Chr.13: 30460190-30460190 on Build GRCh38
rs1016221423 Chr.13: 30460093-30460093 on Build GRCh38
rs914850775 Chr.13: 30460150-30460150 on Build GRCh38
rs957840455 Chr.13: 30460495-30460495 on Build GRCh38
rs59858203 Chr.13: 30460140-30460143 on Build GRCh38
rs977438640 Chr.13: 30460534-30460534 on Build GRCh38
rs1033461990 Chr.13: 30460490-30460490 on Build GRCh38
rs548144896 Chr.13: 30460183-30460186 on Build GRCh38
rs558540478 Chr.13: 30460108-30460111 on Build GRCh38
rs1042366453 Chr.13: 30460344-30460344 on Build GRCh38
rs933516478 Chr.13: 30460373-30460373 on Build GRCh38
rs1050610717 Chr.13: 30460376-30460376 on Build GRCh38
rs4097505 Chr.13: 30460087-30460087 on Build GRCh38
rs539970699 Chr.13: 30460557-30460557 on Build GRCh38
rs946028559 Chr.13: 30460316-30460316 on Build GRCh38
rs914589251 Chr.13: 30460308-30460309 on Build GRCh38
rs200308321 Chr.13: 30460262-30460263 on Build GRCh38
rs993834697 Chr.13: 30460210-30460210 on Build GRCh38
rs61338778 Chr.13: 30460265-30460266 on Build GRCh38
rs941745373 Chr.13: 30460408-30460408 on Build GRCh38
rs146076135 Chr.13: 30460306-30460306 on Build GRCh38
rs560751642 Chr.13: 30460384-30460384 on Build GRCh38
rs1037425789 Chr.13: 30460413-30460413 on Build GRCh38
rs149637108 Chr.13: 30460189-30460192 on Build GRCh38
rs937377629 Chr.13: 30460135-30460135 on Build GRCh38
rs182881863 Chr.13: 30460196-30460196 on Build GRCh38
rs371108527 Chr.13: 30460361-30460361 on Build GRCh38
rs990214266 Chr.13: 30460513-30460513 on Build GRCh38
rs970969697 Chr.13: 30460232-30460232 on Build GRCh38
rs923515152 Chr.13: 30460347-30460347 on Build GRCh38
rs759996394 Chr.13: 30460114-30460117 on Build GRCh38
rs981951397 Chr.13: 30460131-30460131 on Build GRCh38
rs538493533 Chr.13: 30460237-30460237 on Build GRCh38
rs111892138 Chr.13: 30460267-30460267 on Build GRCh38
rs542594948 Chr.13: 30460383-30460383 on Build GRCh38
rs574608092 Chr.13: 30460137-30460140 on Build GRCh38
rs990138808 Chr.13: 30460307-30460307 on Build GRCh38
rs779093822 Chr.13: 30460337-30460340 on Build GRCh38
rs375561440 Chr.13: 30460188-30460191 on Build GRCh38
rs1005929241 Chr.13: 30460194-30460194 on Build GRCh38
rs577524260 Chr.13: 30460223-30460223 on Build GRCh38
rs55642413 Chr.13: 30460307-30460307 on Build GRCh38
rs146618114 Chr.13: 30460171-30460171 on Build GRCh38
rs1002472355 Chr.13: 30460483-30460483 on Build GRCh38
rs1021627653 Chr.13: 30460525-30460525 on Build GRCh38
rs897512531 Chr.13: 30460425-30460425 on Build GRCh38
rs201945336 Chr.13: 30460287-30460287 on Build GRCh38
rs993139495 Chr.13: 30460447-30460447 on Build GRCh38
rs373653401 Chr.13: 30460191-30460194 on Build GRCh38
rs559717140 Chr.13: 30460118-30460118 on Build GRCh38

View Full Product Details