Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4495964-4496479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HMOX2
Entrez Gene ID
3163
Gene Name
heme oxygenase 2
Gene Aliases
HO-2
UniGene
Hs.284279
Gene Chromosome Location
Chr.16: 4474697-4510347 on Build GRCh38
Transcripts
NM_001127204.1 NM_001127205.1 NM_001127206.2 NM_001286267.1 NM_001286268.1 NM_001286269.1 NM_001286270.1 NM_001286271.1 NM_002134.3 XM_011522473.1 XM_017023196.1 XM_017023197.1

SNP Detail

SNP ID Chromosome Location
rs35152645 Chr.16: 4496143-4496143 on Build GRCh38
rs916927520 Chr.16: 4496227-4496227 on Build GRCh38
rs796689055 Chr.16: 4496127-4496127 on Build GRCh38
rs541297357 Chr.16: 4496273-4496273 on Build GRCh38
rs1018934194 Chr.16: 4496038-4496038 on Build GRCh38
rs539178097 Chr.16: 4496452-4496452 on Build GRCh38
rs905585399 Chr.16: 4496267-4496267 on Build GRCh38
rs112357236 Chr.16: 4496316-4496317 on Build GRCh38
rs998454474 Chr.16: 4496161-4496161 on Build GRCh38
rs915707107 Chr.16: 4496022-4496022 on Build GRCh38
rs978646547 Chr.16: 4496107-4496107 on Build GRCh38
rs567351396 Chr.16: 4496255-4496255 on Build GRCh38
rs760085587 Chr.16: 4496375-4496375 on Build GRCh38
rs1029919213 Chr.16: 4496224-4496224 on Build GRCh38
rs757090571 Chr.16: 4496037-4496037 on Build GRCh38
rs1028337350 Chr.16: 4495991-4495991 on Build GRCh38
rs771331405 Chr.16: 4496335-4496335 on Build GRCh38
rs550055636 Chr.16: 4496293-4496293 on Build GRCh38
rs550546524 Chr.16: 4496072-4496072 on Build GRCh38
rs1029390583 Chr.16: 4496373-4496373 on Build GRCh38
rs191348149 Chr.16: 4496145-4496145 on Build GRCh38
rs149371705 Chr.16: 4496281-4496281 on Build GRCh38
rs746623077 Chr.16: 4496117-4496117 on Build GRCh38
rs547241362 Chr.16: 4496371-4496371 on Build GRCh38
rs869269848 Chr.16: 4496127-4496128 on Build GRCh38
rs1053017557 Chr.16: 4496209-4496209 on Build GRCh38
rs182840102 Chr.16: 4496272-4496272 on Build GRCh38
rs1004204854 Chr.16: 4496249-4496249 on Build GRCh38
rs922014937 Chr.16: 4496061-4496061 on Build GRCh38
rs200779344 Chr.16: 4496423-4496423 on Build GRCh38
rs996169732 Chr.16: 4496359-4496359 on Build GRCh38
rs991322615 Chr.16: 4496015-4496018 on Build GRCh38
rs532362879 Chr.16: 4496312-4496313 on Build GRCh38
rs988734123 Chr.16: 4496424-4496424 on Build GRCh38
rs934356802 Chr.16: 4496346-4496346 on Build GRCh38
rs538325242 Chr.16: 4496142-4496143 on Build GRCh38
rs561430435 Chr.16: 4496370-4496370 on Build GRCh38
rs974799949 Chr.16: 4496060-4496060 on Build GRCh38
rs576963255 Chr.16: 4496079-4496079 on Build GRCh38
rs748794597 Chr.16: 4496127-4496129 on Build GRCh38
rs962894045 Chr.16: 4495998-4495999 on Build GRCh38
rs541042043 Chr.16: 4496258-4496258 on Build GRCh38
rs563566468 Chr.16: 4496344-4496344 on Build GRCh38
rs1051377388 Chr.16: 4496448-4496448 on Build GRCh38
rs1044089760 Chr.16: 4496454-4496454 on Build GRCh38
rs917141215 Chr.16: 4496439-4496439 on Build GRCh38
rs185972558 Chr.16: 4496376-4496376 on Build GRCh38
rs766944355 Chr.16: 4496246-4496246 on Build GRCh38
rs769596926 Chr.16: 4496035-4496036 on Build GRCh38
rs762438566 Chr.16: 4496385-4496385 on Build GRCh38
rs892873400 Chr.16: 4496071-4496071 on Build GRCh38
rs138917406 Chr.16: 4496231-4496231 on Build GRCh38
rs567063222 Chr.16: 4496379-4496379 on Build GRCh38
rs1019115055 Chr.16: 4496325-4496325 on Build GRCh38
rs780788397 Chr.16: 4496058-4496058 on Build GRCh38
rs186443131 Chr.16: 4496044-4496044 on Build GRCh38
rs925848063 Chr.16: 4496382-4496382 on Build GRCh38
rs372403965 Chr.16: 4496034-4496034 on Build GRCh38
rs955817859 Chr.16: 4496383-4496383 on Build GRCh38
rs565609526 Chr.16: 4496397-4496397 on Build GRCh38
rs758317776 Chr.16: 4496128-4496129 on Build GRCh38
rs565221201 Chr.16: 4496156-4496156 on Build GRCh38

View Full Product Details