Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112143575-112144078 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL18
Entrez Gene ID
3606
Gene Name
interleukin 18
Gene Aliases
IGIF, IL-18, IL-1g, IL1F4
UniGene
Hs.83077
Gene Chromosome Location
Chr.11: 112143251-112164117 on Build GRCh38
Transcripts
NM_001243211.1 NM_001562.3 XM_011542805.1 XM_011542806.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371652 c.436G>A p.D146N Chr.11: 112143742-112143742 on Build GRCh38
5360627 c.406A>G p.I136V Chr.11: 112143772-112143772 on Build GRCh38
1351280 c.472G>T p.G158* Chr.11: 112143706-112143706 on Build GRCh38
2102781 c.548G>T p.R183I Chr.11: 112143630-112143630 on Build GRCh38
6242886 c.500G>A p.R167K Chr.11: 112143678-112143678 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs919469601 Chr.11: 112143889-112143889 on Build GRCh38
rs568313590 Chr.11: 112143986-112143986 on Build GRCh38
rs767542035 Chr.11: 112143863-112143863 on Build GRCh38
rs267602693 Chr.11: 112143646-112143646 on Build GRCh38
rs751394806 Chr.11: 112143742-112143742 on Build GRCh38
rs1009370485 Chr.11: 112144007-112144007 on Build GRCh38
rs762538166 Chr.11: 112143831-112143831 on Build GRCh38
rs199644300 Chr.11: 112143678-112143678 on Build GRCh38
rs377220185 Chr.11: 112143688-112143688 on Build GRCh38
rs930931157 Chr.11: 112143924-112143924 on Build GRCh38
rs774720131 Chr.11: 112143687-112143687 on Build GRCh38
rs781094787 Chr.11: 112143768-112143768 on Build GRCh38
rs750475139 Chr.11: 112144003-112144003 on Build GRCh38
rs1048717233 Chr.11: 112143936-112143936 on Build GRCh38
rs748404347 Chr.11: 112143770-112143770 on Build GRCh38
rs764934507 Chr.11: 112143823-112143823 on Build GRCh38
rs768324029 Chr.11: 112143790-112143790 on Build GRCh38
rs967939118 Chr.11: 112143716-112143716 on Build GRCh38
rs535317056 Chr.11: 112143975-112143977 on Build GRCh38
rs772444123 Chr.11: 112143812-112143812 on Build GRCh38
rs183621789 Chr.11: 112143947-112143947 on Build GRCh38
rs1020804039 Chr.11: 112144016-112144016 on Build GRCh38
rs200708638 Chr.11: 112143631-112143631 on Build GRCh38
rs1036495457 Chr.11: 112143870-112143870 on Build GRCh38
rs201729952 Chr.11: 112143767-112143767 on Build GRCh38
rs764844182 Chr.11: 112143710-112143710 on Build GRCh38
rs756287389 Chr.11: 112144014-112144014 on Build GRCh38
rs3882891 Chr.11: 112144038-112144038 on Build GRCh38
rs201277302 Chr.11: 112143830-112143830 on Build GRCh38
rs1056202626 Chr.11: 112143987-112143987 on Build GRCh38
rs760993192 Chr.11: 112143820-112143820 on Build GRCh38
rs761403415 Chr.11: 112143701-112143701 on Build GRCh38
rs944845576 Chr.11: 112143839-112143839 on Build GRCh38
rs201855432 Chr.11: 112143709-112143709 on Build GRCh38
rs751547710 Chr.11: 112143858-112143858 on Build GRCh38
rs775880271 Chr.11: 112143814-112143814 on Build GRCh38
rs532583543 Chr.11: 112143937-112143938 on Build GRCh38
rs199505901 Chr.11: 112143604-112143604 on Build GRCh38
rs754825008 Chr.11: 112143743-112143743 on Build GRCh38
rs761553581 Chr.11: 112143951-112143951 on Build GRCh38
rs200887271 Chr.11: 112143734-112143734 on Build GRCh38
rs756454973 Chr.11: 112143774-112143774 on Build GRCh38
rs767815296 Chr.11: 112143694-112143694 on Build GRCh38
rs774005090 Chr.11: 112143741-112143741 on Build GRCh38
rs554823341 Chr.11: 112144047-112144047 on Build GRCh38
rs568928137 Chr.11: 112143860-112143860 on Build GRCh38
rs200850775 Chr.11: 112143698-112143698 on Build GRCh38
rs201530260 Chr.11: 112143600-112143600 on Build GRCh38
rs922360136 Chr.11: 112143898-112143898 on Build GRCh38
rs762921309 Chr.11: 112143647-112143647 on Build GRCh38
rs556943721 Chr.11: 112144039-112144039 on Build GRCh38
rs201898660 Chr.11: 112143676-112143676 on Build GRCh38
rs111632191 Chr.11: 112144024-112144024 on Build GRCh38
rs911992503 Chr.11: 112143838-112143838 on Build GRCh38
rs201170165 Chr.11: 112143613-112143613 on Build GRCh38
rs752581080 Chr.11: 112143865-112143865 on Build GRCh38
rs751985093 Chr.11: 112143649-112143650 on Build GRCh38
rs749454902 Chr.11: 112143786-112143786 on Build GRCh38
rs746330842 Chr.11: 112143810-112143810 on Build GRCh38
rs757840888 Chr.11: 112143730-112143730 on Build GRCh38
rs574538646 Chr.11: 112144048-112144048 on Build GRCh38
rs200076894 Chr.11: 112143822-112143822 on Build GRCh38
rs199758158 Chr.11: 112143625-112143625 on Build GRCh38
rs770906130 Chr.11: 112143797-112143797 on Build GRCh38
rs576890715 Chr.11: 112144044-112144044 on Build GRCh38
rs141853304 Chr.11: 112143798-112143798 on Build GRCh38
rs202091683 Chr.11: 112143833-112143833 on Build GRCh38
rs368708524 Chr.11: 112143861-112143861 on Build GRCh38
rs749986913 Chr.11: 112143721-112143721 on Build GRCh38
rs199869197 Chr.11: 112143732-112143732 on Build GRCh38
rs199498017 Chr.11: 112143836-112143836 on Build GRCh38
rs895862739 Chr.11: 112143637-112143637 on Build GRCh38
rs5744290 Chr.11: 112143605-112143605 on Build GRCh38
rs908406301 Chr.11: 112143954-112143954 on Build GRCh38
rs760069850 Chr.11: 112143833-112143833 on Build GRCh38
rs545913818 Chr.11: 112144051-112144051 on Build GRCh38

View Full Product Details