Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 3068248-3068774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL5RA
Entrez Gene ID
3568
Gene Name
interleukin 5 receptor subunit alpha
Gene Aliases
CD125, CDw125, HSIL5R3, IL5R
UniGene
Hs.68876
Gene Chromosome Location
Chr.3: 3066324-3110414 on Build GRCh38
Transcripts
NM_000564.4 NM_001243099.1 NM_175726.3 XM_011533677.2 XM_011533678.2

SNP Detail

SNP ID Chromosome Location
rs892423815 Chr.3: 3068725-3068725 on Build GRCh38
rs899323086 Chr.3: 3068633-3068633 on Build GRCh38
rs988583198 Chr.3: 3068566-3068566 on Build GRCh38
rs980196891 Chr.3: 3068516-3068516 on Build GRCh38
rs533838922 Chr.3: 3068660-3068660 on Build GRCh38
rs1038907511 Chr.3: 3068661-3068661 on Build GRCh38
rs567207930 Chr.3: 3068688-3068688 on Build GRCh38
rs9821233 Chr.3: 3068601-3068601 on Build GRCh38
rs532622897 Chr.3: 3068343-3068343 on Build GRCh38
rs1009611891 Chr.3: 3068734-3068734 on Build GRCh38
rs760670295 Chr.3: 3068588-3068595 on Build GRCh38
rs750572577 Chr.3: 3068742-3068742 on Build GRCh38
rs780399499 Chr.3: 3068595-3068596 on Build GRCh38
rs1038963152 Chr.3: 3068369-3068369 on Build GRCh38
rs6768065 Chr.3: 3068553-3068553 on Build GRCh38
rs988654514 Chr.3: 3068587-3068587 on Build GRCh38
rs571676434 Chr.3: 3068408-3068409 on Build GRCh38
rs1042999300 Chr.3: 3068342-3068342 on Build GRCh38
rs57131247 Chr.3: 3068608-3068609 on Build GRCh38
rs375621201 Chr.3: 3068609-3068609 on Build GRCh38
rs202020785 Chr.3: 3068600-3068600 on Build GRCh38
rs995041501 Chr.3: 3068649-3068649 on Build GRCh38
rs961076723 Chr.3: 3068459-3068459 on Build GRCh38
rs190105041 Chr.3: 3068344-3068344 on Build GRCh38
rs918884380 Chr.3: 3068595-3068609 on Build GRCh38
rs532608636 Chr.3: 3068536-3068536 on Build GRCh38
rs138591145 Chr.3: 3068600-3068601 on Build GRCh38
rs928554337 Chr.3: 3068524-3068524 on Build GRCh38
rs559634547 Chr.3: 3068352-3068352 on Build GRCh38
rs186292449 Chr.3: 3068449-3068449 on Build GRCh38
rs112253246 Chr.3: 3068517-3068517 on Build GRCh38
rs151100168 Chr.3: 3068514-3068514 on Build GRCh38
rs7615194 Chr.3: 3068586-3068586 on Build GRCh38
rs988699323 Chr.3: 3068720-3068720 on Build GRCh38
rs769208542 Chr.3: 3068589-3068595 on Build GRCh38
rs7615192 Chr.3: 3068581-3068581 on Build GRCh38
rs959911473 Chr.3: 3068706-3068706 on Build GRCh38
rs558754874 Chr.3: 3068597-3068597 on Build GRCh38
rs572022327 Chr.3: 3068280-3068281 on Build GRCh38
rs68121623 Chr.3: 3068601-3068602 on Build GRCh38
rs7617502 Chr.3: 3068652-3068652 on Build GRCh38
rs141075353 Chr.3: 3068549-3068549 on Build GRCh38
rs647154 Chr.3: 3068609-3068609 on Build GRCh38
rs544990780 Chr.3: 3068607-3068614 on Build GRCh38
rs143760788 Chr.3: 3068466-3068466 on Build GRCh38
rs1005041754 Chr.3: 3068426-3068426 on Build GRCh38
rs530926244 Chr.3: 3068470-3068470 on Build GRCh38
rs1032323271 Chr.3: 3068441-3068441 on Build GRCh38
rs973639163 Chr.3: 3068547-3068547 on Build GRCh38
rs531979293 Chr.3: 3068311-3068311 on Build GRCh38
rs181948902 Chr.3: 3068445-3068445 on Build GRCh38
rs190035781 Chr.3: 3068650-3068650 on Build GRCh38
rs532639027 Chr.3: 3068420-3068421 on Build GRCh38
rs1005140088 Chr.3: 3068604-3068604 on Build GRCh38
rs947494840 Chr.3: 3068336-3068336 on Build GRCh38
rs373023205 Chr.3: 3068409-3068409 on Build GRCh38
rs980938477 Chr.3: 3068503-3068503 on Build GRCh38
rs1010030146 Chr.3: 3068456-3068456 on Build GRCh38
rs564865287 Chr.3: 3068608-3068614 on Build GRCh38
rs749095632 Chr.3: 3068678-3068679 on Build GRCh38
rs36113235 Chr.3: 3068683-3068684 on Build GRCh38
rs1053034963 Chr.3: 3068689-3068689 on Build GRCh38
rs1052228213 Chr.3: 3068602-3068602 on Build GRCh38
rs963621610 Chr.3: 3068539-3068539 on Build GRCh38
rs543192232 Chr.3: 3068324-3068324 on Build GRCh38
rs960245258 Chr.3: 3068529-3068529 on Build GRCh38
rs1043116336 Chr.3: 3068599-3068599 on Build GRCh38
rs550844525 Chr.3: 3068505-3068505 on Build GRCh38
rs114513243 Chr.3: 3068738-3068738 on Build GRCh38
rs200496802 Chr.3: 3068603-3068603 on Build GRCh38
rs1036497569 Chr.3: 3068618-3068618 on Build GRCh38
rs941691800 Chr.3: 3068274-3068274 on Build GRCh38
rs1052020138 Chr.3: 3068345-3068345 on Build GRCh38
rs1052910298 Chr.3: 3068288-3068288 on Build GRCh38
rs563392356 Chr.3: 3068335-3068335 on Build GRCh38
rs913071756 Chr.3: 3068576-3068576 on Build GRCh38
rs555288372 Chr.3: 3068673-3068673 on Build GRCh38
rs528733499 Chr.3: 3068427-3068427 on Build GRCh38

View Full Product Details