Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 129087405-129087895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISY1-RAB43
Entrez Gene ID
100534599
Gene Name
ISY1-RAB43 readthrough
Gene Aliases
-
UniGene
Hs.512661
Gene Chromosome Location
Chr.3: 129087569-129161230 on Build GRCh38
Transcripts
NM_001204890.1 NM_001204883.1 NM_001204884.1 NM_001204885.1 NM_001204886.1 NM_001204887.1 NM_001204888.1 NM_198490.2

Gene Symbol
RAB43
Entrez Gene ID
339122
Gene Name
RAB43, member RAS oncogene family
Gene Aliases
RAB11B, RAB41
UniGene
Hs.381132
Gene Chromosome Location
Chr.3: 129087569-129122150 on Build GRCh38
Transcripts
NM_001204890.1 NM_001204883.1 NM_001204884.1 NM_001204885.1 NM_001204886.1 NM_001204887.1 NM_001204888.1 NM_198490.2

SNP Detail

SNP ID Chromosome Location
rs557965059 Chr.3: 129087483-129087483 on Build GRCh38
rs982143963 Chr.3: 129087679-129087679 on Build GRCh38
rs981447279 Chr.3: 129087565-129087565 on Build GRCh38
rs566208340 Chr.3: 129087564-129087564 on Build GRCh38
rs7635746 Chr.3: 129087475-129087475 on Build GRCh38
rs989509699 Chr.3: 129087648-129087648 on Build GRCh38
rs928168606 Chr.3: 129087699-129087699 on Build GRCh38
rs909007737 Chr.3: 129087639-129087639 on Build GRCh38
rs938296784 Chr.3: 129087700-129087700 on Build GRCh38
rs998224659 Chr.3: 129087809-129087809 on Build GRCh38
rs1055272662 Chr.3: 129087724-129087724 on Build GRCh38
rs569003161 Chr.3: 129087448-129087448 on Build GRCh38
rs914027609 Chr.3: 129087664-129087664 on Build GRCh38
rs958086279 Chr.3: 129087629-129087629 on Build GRCh38
rs963724880 Chr.3: 129087489-129087489 on Build GRCh38
rs906820753 Chr.3: 129087427-129087427 on Build GRCh38
rs529158666 Chr.3: 129087437-129087437 on Build GRCh38
rs893277396 Chr.3: 129087441-129087441 on Build GRCh38
rs995226637 Chr.3: 129087518-129087518 on Build GRCh38
rs1029331745 Chr.3: 129087813-129087813 on Build GRCh38
rs1029984605 Chr.3: 129087527-129087527 on Build GRCh38
rs917834762 Chr.3: 129087539-129087539 on Build GRCh38
rs1032919303 Chr.3: 129087824-129087824 on Build GRCh38
rs1032824359 Chr.3: 129087627-129087627 on Build GRCh38
rs550602416 Chr.3: 129087445-129087445 on Build GRCh38
rs553686645 Chr.3: 129087738-129087738 on Build GRCh38
rs573311299 Chr.3: 129087738-129087738 on Build GRCh38
rs985657695 Chr.3: 129087566-129087566 on Build GRCh38
rs556602387 Chr.3: 129087786-129087786 on Build GRCh38
rs113299362 Chr.3: 129087488-129087488 on Build GRCh38
rs891405168 Chr.3: 129087470-129087470 on Build GRCh38
rs957199499 Chr.3: 129087829-129087829 on Build GRCh38
rs988638953 Chr.3: 129087853-129087853 on Build GRCh38
rs951153988 Chr.3: 129087523-129087523 on Build GRCh38
rs145026762 Chr.3: 129087466-129087466 on Build GRCh38
rs888175680 Chr.3: 129087482-129087482 on Build GRCh38
rs544457349 Chr.3: 129087739-129087739 on Build GRCh38
rs889676839 Chr.3: 129087816-129087816 on Build GRCh38
rs896804179 Chr.3: 129087495-129087495 on Build GRCh38
rs888604574 Chr.3: 129087735-129087735 on Build GRCh38
rs1026742590 Chr.3: 129087520-129087520 on Build GRCh38
rs971082119 Chr.3: 129087554-129087554 on Build GRCh38
rs1006821120 Chr.3: 129087817-129087817 on Build GRCh38
rs945362194 Chr.3: 129087673-129087673 on Build GRCh38
rs533686362 Chr.3: 129087573-129087573 on Build GRCh38
rs7626531 Chr.3: 129087703-129087703 on Build GRCh38
rs982653437 Chr.3: 129087537-129087537 on Build GRCh38

View Full Product Details